Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  30  40  50  60  70  80  90  100  110  120  121  122  123  124  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1622 
4021 - 4050 of 48655 products displayed
  • BCL2A1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519386
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Protein GRS; Protein BFL-1; Hemopoietic-specific early response protein; Bcl-2-like protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519386 349.00
Check your price
  • BCL2A1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520268
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Protein GRS; Protein BFL-1; Hemopoietic-specific early response protein; Bcl-2-like protein 5; A1-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520268 349.00
Check your price
  • BCL2A1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520269
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20µg/ml), Immunohistochemistry (2µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Protein GRS; Protein BFL-1; Hemopoietic-specific early response protein; Bcl-2-like protein 5; A1-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520269 349.00
Check your price
  • BCL2L10 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520234
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Apoptosis regulator Bcl-B; Anti-apoptotic protein NrH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520234 349.00
Check your price
  • BCL7A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517933
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
BCL7A; BCL7; B-cell CLL/lymphoma 7A; B-cell CLL/lymphoma-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517933 349.00
Check your price
  • BCL9L Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521111
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Protein BCL9-2; BCL9-related beta-catenin-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521111 349.00
Check your price
  • BCMA Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519973
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (Paraffin) (10 ug/ml), Western Blot (5 ug/ml)
Alternate names:
B-cell maturation protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519973 349.00
Check your price
  • BDH2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518866
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Short chain dehydrogenase/reductase family 15C member 1; R-beta-hydroxybutyrate dehydrogenase; Oxidoreductase UCPA; Dehydrogenase/reductase SDR family member 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518866 349.00
Check your price
  • Beclin 1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520171
  • Size 100 ug
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Coiled-coil myosin-like BCL2-interacting protein; Protein GT197
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520171 349.00
Check your price
  • Beclin 1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520172
  • Size 100 ug
Applications
Immunohistochemistry (2 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Protein GT197; Coiled-coil myosin-like BCL2-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520172 349.00
Check your price
  • Benzoylecgonine Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA514691
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514691 349.00
Check your price
  • Benzoylecgonine Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA514673
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514673 349.00
Check your price
  • BERP Polyclonal Antibody for Western Blot and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518136
  • Size 100 ug
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
RING finger protein HAC1; RING finger protein 22; RING finger protein 97; Brain-expressed RING finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518136 349.00
Check your price
  • beta Actin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1140
  • Size 100 ug
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunoprecipitation (2ug), Western Blot (1:5000-1:20,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1140 219.00
Check your price
  • beta Amyloid Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519922
  • Size 100 ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519922 349.00
Check your price
  • beta Amyloid Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519923
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (2 ug/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519923 349.00
Check your price
  • beta Amyloid Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520711
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Amyloidogenic glycoprotein; Alzheimer disease amyloid A4 protein homolog; ABPP; Protease nexin-II; PreA4; Cerebral vascular amyloid peptide; Alzheimer disease amyloid protein; APPI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520711 349.00
Check your price
  • beta Arrestin 1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA523342
  • Size 100 ug
Applications
Western Blot (2.0-3 µg/ml)
Alternate names:
Arrestin beta-1; Arrestin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523342 349.00
Check your price
  • beta Catenin Monoclonal Antibody for Western Blot, ICC, Flow and IP
  • Host Mouse
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA110056
  • Size 100 ug
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110056 249.00
Check your price
  • beta Galactosidase Monoclonal Antibody for Western Blot and ICC
  • Host Mouse
  • Target Species All
  • Conjugate Unconjugated
  • Catalog # MA110057
  • Size 100 ug
Applications
Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110057 225.00
Check your price
  • beta-1 Adrenergic Receptor Polyclonal Antibody for Western Blot and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518482
  • Size 100 ug
Applications
Western Blot (2-6 µg/ml)
Alternate names:
Beta-1 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518482 349.00
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA518423
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518423 349.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Biotin
  • Catalog # MA110054
  • Size 100 ug
Applications
Flow Cytometry (1:200)
Alternate names:
CDABP0092, beta chain of MHC class I molecules, beta-2-microglobin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110054 249.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, APC conjugate for Western Blot and Flow
  • Host Rat
  • Target Species Human
  • Conjugate APC
  • Catalog # MA517683
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Western Blot (Assay Dependent)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517683 325.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, Biotin conjugate for Western Blot and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA517684
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Western Blot (Assay Dependent)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517684 225.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, FITC conjugate for Western Blot and Flow
  • Host Rat
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA517685
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Western Blot (Assay Dependent)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517685 225.00
Check your price
  • beta-3 Adrenergic Receptor Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518793
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Beta-3 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518793 349.00
Check your price
  • beta-Arrestin 2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523296
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.25-0.5 µg/ml)
Alternate names:
Arrestin beta-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523296 349.00
Check your price
  • BFAR Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520894
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
RING finger protein 47
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520894 349.00
Check your price
  • BHMT Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518772
  • Size 100 ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.03-0.1 ug/ml)
Alternate names:
BHMT; betaine-homocysteine methyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518772 349.00
Check your price