Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  40  50  60  70  80  90  100  110  120  125  126  127  128  129  130  131  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  160  170  180  190  200  210  220  230  300  400  500  600  700  800  900  1000  1100  1630 
4171 - 4200 of 48877 products displayed
  • NDRG1 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518109
  • Size 100ug
Applications
Immunohistochemistry (2-5µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Rit42; Reducing agents and tunicamycin-responsive protein; Nickel-specific induction protein Cap43; N-myc downstream-regulated gene 1 protein; Differentiation-related gene 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518109 349.00
Check your price
  • NDUFAF2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520831
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
NDUFA12-like protein; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2; Myc-induced mitochondrial protein; B17.2-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520831 349.00
Check your price
  • NDUFB9 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520922
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase B22 subunit; Complex I-B22; LYR motif-containing protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520922 349.00
Check your price
  • NDUFS1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519341
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Complex I-75kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519341 349.00
Check your price
  • NDUFS2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519342
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 49 kDa subunit; Complex I-49kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519342 349.00
Check your price
  • NDUFS3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519118
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 30 kDa subunit; Complex I-30kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519118 349.00
Check your price
  • NDUFS6 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519238
  • Size 100ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 13 kDa-A subunit; Complex I-13kD-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519238 349.00
Check your price
  • NDUFS7 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519343
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
PSST subunit; NADH-ubiquinone oxidoreductase 20 kDa subunit; Complex I-20kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519343 349.00
Check your price
  • NDUFS8 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519344
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 23 kDa subunit; Complex I-23kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519344 349.00
Check your price
  • NEDD1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518626
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Neural precursor cell expressed developmentally down-regulated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518626 349.00
Check your price
  • NEDD4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523492
  • Size 100ug
Applications
Western Blot (4-6 µg/ml)
Alternate names:
Neural precursor cell expressed developmentally down-regulated protein 4; Cell proliferation-inducing gene 53 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523492 349.00
Check your price
  • NEDD4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523462
  • Size 100ug
Applications
Western Blot (4-6 µg/ml)
Alternate names:
Neural precursor cell expressed developmentally down-regulated protein 4; Cell proliferation-inducing gene 53 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523462 349.00
Check your price
  • NEDD8 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517133
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Ubiquitin-like protein Nedd8; Neural precursor cell expressed developmentally down-regulated protein 8; Neddylin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517133 309.00
Check your price
  • NEFH Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517134
  • Size 100ug
Applications
Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517134 309.00
Check your price
  • NEFH Monoclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Mouse
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA110441
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (1 ug/ml)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110441 225.00
Check your price
  • NEFL Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517135
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Neurofilament triplet L protein; 68 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517135 309.00
Check your price
  • NEFM Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519200
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Neurofilament triplet M protein; Neurofilament 3; 160 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519200 349.00
Check your price
  • NEIL1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518134
  • Size 100ug
Applications
Immunohistochemistry (5-10µg/ml)
Alternate names:
Nei-like protein 1; Nei homolog 1; FPG1; Endonuclease VIII-like 1; DNA-(apurinic or apyrimidinic site) lyase Neil1; DNA glycosylase/AP lyase Neil1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518134 349.00
Check your price
  • NEK7 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519410
  • Size 100ug
Applications
Western Blot (0.05-0.2 ug/ml)
Alternate names:
Never in mitosis A-related kinase 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519410 349.00
Check your price
  • NENF Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520577
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Secreted protein of unknown function; Neuron-derived neurotrophic factor; Spinal cord injury-related protein 10; SCIRP10-related protein; Protein GIG47; Cell immortalization-related protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520577 349.00
Check your price
  • Nestin Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517136
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517136 309.00
Check your price
  • Nestin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523386
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (3-5 µg/ml)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523386 349.00
Check your price
  • NET1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518526
  • Size 100ug
Applications
Immunohistochemistry (2.5µg/ml)
Alternate names:
Rho guanine nucleotide exchange factor 8; Proto-oncogene p65 Net1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518526 349.00
Check your price
  • NET1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518217
  • Size 100ug
Applications
Immunohistochemistry (2.5µg/ml)
Alternate names:
Rho guanine nucleotide exchange factor 8; Proto-oncogene p65 Net1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518217 349.00
Check your price
  • Neuritin Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520368
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (5-10 ug/ml)
Alternate names:
Candidate plasticity gene 15 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520368 349.00
Check your price
  • NeuroD1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517137
  • Size 100ug
Applications
Western Blot (1:500-1:2000)
Alternate names:
Beta-cell E-box transcriptional activator 2; Class A basic helix-loop-helix protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517137 309.00
Check your price
  • NeuroG2 Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534472
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Protein atonal homolog 4; Class A basic helix-loop-helix protein 8; Helix-loop-helix protein mATH-4A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534472 349.00
Check your price
  • NeuroG3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA523481
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Protein atonal homolog 5; Helix-loop-helix protein mATH-4B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523481 349.00
Check your price
  • Neuropilin 1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523306
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Vascular endothelial cell growth factor 165 receptor; A5 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523306 349.00
Check your price
  • Neuroserpin Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518359
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Serpin I1; Peptidase inhibitor 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518359 349.00
Check your price