Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  40  50  60  70  80  90  100  110  120  132  133  134  135  136  137  138  139  140  141  142  143  144  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  170  180  190  200  210  220  230  240  300  400  500  600  700  800  900  1000  1100  1622 
4381 - 4410 of 48655 products displayed
  • CD28 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517471
  • Size 100 ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
MGC138290, Tp44, CD28 antigen, T-cell-specific surface glycoprotein CD28
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517471 159.00
Check your price
  • CD2BP2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518286
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 ug/ml), Western Blot (0.3-1.0 ug/ml)
Alternate names:
U5 snRNP 52K protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518286 349.00
Check your price
  • CD3 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA516460
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516460 225.00
Check your price
  • CD3 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA516461
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516461 275.00
Check your price
  • CD3 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17640
  • Size 100 ug
Applications
Flow Cytometry (1:1000)
Alternate names:
T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17640 275.00
Check your price
  • CD300a Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110190
  • Size 100 ug
Applications
Flow Cytometry (2 ug/ml), Western Blot (Assay Dependent)
Alternate names:
NK inhibitory receptor; Inhibitory receptor protein 60; Immunoglobulin superfamily member 12; IRC1/IRC2; CMRF35-H; CMRF-35-H9; CD300 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110190 225.00
Check your price
  • CD31 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Biotin
  • Catalog # MA516770
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:200)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516770 475.00
Check your price
  • CD31 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Non-human primate, Rat
  • Conjugate Biotin
  • Catalog # MA516950
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516950 225.00
Check your price
  • CD31 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Non-human primate, Rat
  • Conjugate FITC
  • Catalog # MA516952
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516952 225.00
Check your price
  • CD33 Monoclonal Antibody for Western Blot, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516982
  • Size 100 ug
Applications
Flow Cytometry (1:10 -1:50), Immunohistochemistry (Frozen) (1:50-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
gp67; Sialic acid-binding Ig-like lectin 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516982 159.00
Check your price
  • CD33 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516981
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
gp67; Sialic acid-binding Ig-like lectin 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516981 325.00
Check your price
  • CD34 Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA110203
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml), Immunohistochemistry (Frozen) (Assay-Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110203 159.00
Check your price
  • CD34 Monoclonal Antibody for IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17652
  • Size 100 ug
Applications
Flow Cytometry (1:1000), Immunohistochemistry (Assay dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17652 275.00
Check your price
  • CD34 Monoclonal Antibody, Biotin conjugate for Western Blot, IHC (F), IHC (P), Flow and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517827
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517827 129.00
Check your price
  • CD34 Monoclonal Antibody, FITC conjugate for Western Blot, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517829
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517829 149.00
Check your price
  • CD34 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow, IP and FN
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA110202
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml), Functional Assay (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1-2µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110202 159.00
Check your price
  • CD35 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate FITC
  • Catalog # MA516649
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
C3b/C4b receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516649 325.00
Check your price
  • CD35 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA517712
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
C3b/C4b receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517712 309.00
Check your price
  • CD35 Monoclonal Antibody, Biotin conjugate for Western Blot, IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA517713
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
C3b/C4b receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517713 225.00
Check your price
  • CD35 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA517714
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
C3b/C4b receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517714 225.00
Check your price
  • CD357 Monoclonal Antibody, Biotin conjugate for IHC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517932
  • Size 100 ug
Applications
Flow Cytometry (1.0 ug/10^6 cells), Immunohistochemistry (Assay Dependent)
Alternate names:
Glucocorticoid-induced TNFR-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517932 159.00
Check your price
  • CD357 Monoclonal Antibody, FITC conjugate for IHC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517933
  • Size 100 ug
Applications
Flow Cytometry (1.0 ug/10^6 cells), Immunohistochemistry (Assay Dependent)
Alternate names:
Glucocorticoid-induced TNFR-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517933 179.00
Check your price
  • CD36 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110209
  • Size 100 ug
Applications
Flow Cytometry (1 ug/ml)
Alternate names:
Thrombospondin receptor; Platelet glycoprotein IV; Platelet collagen receptor; PAS-4; PAS IV; Leukocyte differentiation antigen CD36; Glycoprotein IIIb; Fatty acid translocase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110209 159.00
Check your price
  • CD36 Monoclonal Antibody for IF and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516833
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:25), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Platelet glycoprotein IV; PAS-4; PAS IV; Glycoprotein IIIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516833 225.00
Check your price
  • CD36 Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA516832
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
Platelet glycoprotein IV; PAS-4; PAS IV; Glycoprotein IIIb
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516832 325.00
Check your price
  • CD36 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516940
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
Thrombospondin receptor; Platelet glycoprotein IV; Platelet collagen receptor; PAS-4; PAS IV; Leukocyte differentiation antigen CD36; Glycoprotein IIIb; Fatty acid translocase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516940 325.00
Check your price
  • CD37 Monoclonal Antibody for IHC, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17627
  • Size 100 ug
Applications
ELISA (1:1000), Flow Cytometry (1:1000), Immunohistochemistry (1:1000), Immunoprecipitation (1:1000)
Alternate names:
Tetraspanin-26
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17627 275.00
Check your price
  • CD38 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17653
  • Size 100 ug
Applications
Flow Cytometry (1:1000)
Alternate names:
T10; Cyclic ADP-ribose hydrolase 1; ADP-ribosyl cyclase 1; 2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase; 2'-phospho-ADP-ribosyl cyclase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17653 275.00
Check your price
  • CD3e Monoclonal Antibody for ICC, IHC (F), Flow, IP and FN
  • Host Armenian Hamster
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA110184
  • Size 100 ug
Applications
Flow Cytometry (1-2 ug/ml), Functional Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (1-2 ug/ml)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110184 119.00
Check your price
  • CD3e Monoclonal Antibody, APC conjugate for Flow
  • Host Armenian Hamster
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA110186
  • Size 100 ug
Applications
Flow Cytometry (2 ug/ml)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110186 275.00
Check your price