Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  145  146  147  148  149  150  151  152  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  180  190  200  210  220  230  240  250  300  400  500  600  700  800  900  1000  1100  1605 
4771 - 4800 of 48135 products displayed
  • CPNE1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519137
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Copine I; Chromobindin 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519137 349.00
Check your price
  • CPT1A Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518578
  • Size 100 ug
Applications
Immunohistochemistry (4-6µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Carnitine palmitoyltransferase 1A; Carnitine O-palmitoyltransferase I, liver isoform
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518578 349.00
Check your price
  • CPT1B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518062
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Carnitine palmitoyltransferase I-like protein; Carnitine palmitoyltransferase 1B; Carnitine O-palmitoyltransferase I, muscle isoform
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518062 349.00
Check your price
  • CPT1C Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519021
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Carnitine palmitoyltransferase 1C; Carnitine O-palmitoyltransferase I, brain isoform; CPT IC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519021 349.00
Check your price
  • CPT2 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519222
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Carnitine palmitoyltransferase II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519222 349.00
Check your price
  • CPT2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519223
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Carnitine palmitoyltransferase II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519223 349.00
Check your price
  • CRABP2 Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523213
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Cellular retinoic acid-binding protein II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523213 349.00
Check your price
  • CRB1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534491
  • Size 100 ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
CRB1, Crumbs homolog 1, LCA8, RP12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534491 349.00
Check your price
  • CRB2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534492
  • Size 100 ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
Crumbs-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534492 349.00
Check your price
  • Creatine Kinase BB Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518490
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Creatine kinase B chain; B-CK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518490 349.00
Check your price
  • CREB3L1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518417
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Old astrocyte specifically-induced substance
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518417 349.00
Check your price
  • CREB3L4 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518028
  • Size 100 ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
hJAL; Transcript induced in spermiogenesis protein 40; Cyclic AMP-responsive element-binding protein 4; Attaching to CRE-like 1; Androgen-induced basic leucine zipper protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518028 349.00
Check your price
  • CREBZF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521106
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Host cell factor-binding transcription factor Zhangfei; Tyrosine kinase-associated leucine zipper protein LAZip
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521106 349.00
Check your price
  • CREM Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518313
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Inducible cAMP early repressor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518313 349.00
Check your price
  • CRFR1 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518625
  • Size 100 ug
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Corticotropin-releasing hormone receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518625 349.00
Check your price
  • CRFR1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518801
  • Size 100 ug
Applications
Immunocytochemistry (0.1-0.5ug/ml), Immunofluorescence (2ug/ml), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Corticotropin-releasing hormone receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518801 349.00
Check your price
  • CRHR2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523129
  • Size 100 ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Corticotropin-releasing hormone receptor 2; CRF-RB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523129 349.00
Check your price
  • CRIM1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534410
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/mL), Western Blot (1 ug/mL)
Alternate names:
Cysteine-rich repeat-containing protein S52
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534410 349.00
Check your price
  • CRIM2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534411
  • Size 100 ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
Kielin/chordin-like protein 1; Cysteine-rich motor neuron 2 protein; Cysteine-rich BMP regulator 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534411 349.00
Check your price
  • Cripto Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518951
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Epidermal growth factor-like cripto protein CR1; Cripto-1 growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518951 349.00
Check your price
  • Cripto Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534476
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/mL), Western Blot (1-2 ug/mL)
Alternate names:
Epidermal growth factor-like cripto protein CR1; Cripto-1 growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534476 349.00
Check your price
  • CRISP2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519266
  • Size 100 ug
Applications
Western Blot (0.05-0.1 ug/ml)
Alternate names:
Testis-specific protein TPX-1; Cancer/testis antigen 36
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519266 349.00
Check your price
  • CRISP2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519265
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
Testis-specific protein TPX-1; Cancer/testis antigen 36
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519265 349.00
Check your price
  • CRISP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521007
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Testis-specific protein TPX-1; Cancer/testis antigen 36
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521007 349.00
Check your price
  • CrkL Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19033
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.01-0.03 µg/ml)
Alternate names:
CRKL; v-crk sarcoma virus CT10 oncogene homolog (avian)-like; v-crk avian sarcoma virus CT10 oncogene homolog-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19033 309.00
Check your price
  • CRP Antibody (Monoclonal, C2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110321
  • Size 100 ug
Applications
ELISA (Assay Dependent)
Alternate names:
RP11-419N10.4, MGC149895, MGC88244, PTX1, C-reactive protein, pentraxin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110321 249.00
Check your price
  • CRP Antibody (Monoclonal, C5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110319
  • Size 100 ug
Applications
ELISA (Assay Dependent)
Alternate names:
RP11-419N10.4, MGC149895, MGC88244, PTX1, C-reactive protein, pentraxin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110319 249.00
Check your price
  • CRP Antibody (Monoclonal, C6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110322
  • Size 100 ug
Applications
ELISA (Assay Dependent)
Alternate names:
RP11-419N10.4, MGC149895, MGC88244, PTX1, C-reactive protein, pentraxin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110322 249.00
Check your price
  • CRP Antibody (Monoclonal, C7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110320
  • Size 100 ug
Applications
ELISA (Assay Dependent)
Alternate names:
RP11-419N10.4, MGC149895, MGC88244, PTX1, C-reactive protein, pentraxin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110320 249.00
Check your price
  • CRP Antibody, Biotin (Monoclonal, C7)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110323
  • Size 100 ug
Applications
ELISA (Assay Dependent)
Alternate names:
RP11-419N10.4, MGC149895, MGC88244, PTX1, C-reactive protein, pentraxin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110323 349.00
Check your price