Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  60  70  80  90  100  110  120  130  140  153  154  155  156  157  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  190  200  210  220  230  240  250  260  300  400  500  600  700  800  900  1000  1100  1622 
5011 - 5040 of 48655 products displayed
  • EDG3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523225
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1.0-2 µg/ml)
Alternate names:
Sphingosine 1-phosphate receptor Edg-3; Endothelial differentiation G-protein coupled receptor 3; Lysophospholipid receptor B3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523225 349.00
Check your price
  • EDG6 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523221
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (3.0-5 µg/ml)
Alternate names:
Sphingosine 1-phosphate receptor Edg-6; Lysophospholipid receptor C1; Endothelial differentiation G-protein coupled receptor 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523221 349.00
Check your price
  • EDG8 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523195
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Sphingosine 1-phosphate receptor Edg-8; Lysophospholipid receptor B4; Endothelial differentiation G-protein-coupled receptor 8; Nerve growth factor-regulated G-protein-coupled receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523195 349.00
Check your price
  • EED Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534430
  • Size 100 ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1 ug/mL)
Alternate names:
WD protein associating with integrin cytoplasmic tails 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534430 349.00
Check your price
  • EFCAB4A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521059
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Calcium release-activated channel regulator 2B; Calcium release-activated calcium channel regulator 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521059 349.00
Check your price
  • EFCAB4B Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521060
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Calcium release-activated channel regulator 2A; Calcium release-activated calcium channel regulator 2A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521060 349.00
Check your price
  • EFHD1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520918
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (5-20 µg/ml), Immunohistochemistry (20 ug/ml), Western Blot (2-4 ug/ml)
Alternate names:
Swiprosin-2; EF-hand domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520918 349.00
Check your price
  • EFHD2 Polyclonal Antibody for Western Blot, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518321
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.3-1 µg/ml)
Alternate names:
Swiprosin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518321 349.00
Check your price
  • EFHD2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520919
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Swiprosin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520919 349.00
Check your price
  • eGFP Tag Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # CAB4211
  • Size 100 ug
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Western Blot (1 ug/ml)
Alternate names:
Enhanced Green Fluorescence Protein, Venus, enhanced GFP
Catalog # Size Price (AUD) Your Price (AUD) Qty
CAB4211 149.00
Check your price
  • EGR2 Polyclonal Antibody for Western Blot, IHC and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518750
  • Size 100 ug
Applications
Immunohistochemistry (3-5µg/ml), Western Blot (0.03-0.1 ug/ml)
Alternate names:
Zinc finger protein Krox-20; Early growth response protein 2; AT591
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518750 349.00
Check your price
  • EGR4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518814
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
AT133
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518814 349.00
Check your price
  • EHD2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518303
  • Size 100 ug
Applications
Western Blot (0.003-0.01 ug/ml)
Alternate names:
PAST homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518303 349.00
Check your price
  • EHMT1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515277
  • Size 100 ug
Applications
Western Blot (1/500-1/2000)
Alternate names:
Lysine N-methyltransferase 1D; Histone H3-K9 methyltransferase 5; G9a-like protein 1; Euchromatic histone-lysine N-methyltransferase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515277 349.00
Check your price
  • eIF2b delta Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519345
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
eIF-2B GDP-GTP exchange factor subunit delta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519345 349.00
Check your price
  • eIF3a Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523285
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.25-1 µg/ml)
Alternate names:
eIF3 p185; eIF3 p180; eIF3 p167; eIF-3-theta; Eukaryotic translation initiation factor 3 subunit 10; p162; Centrosomin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523285 349.00
Check your price
  • eIF3b Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523278
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.05-0.25 µg/ml)
Alternate names:
eIF3 p116; eIF3 p110; eIF-3-eta; Prt1 homolog; Eukaryotic translation initiation factor 3 subunit 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523278 349.00
Check your price
  • eIF3d Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA523293
  • Size 100 ug
Applications
Western Blot (0.005-0.01 µg/ml)
Alternate names:
eIF3 p66; eIF-3-zeta; Eukaryotic translation initiation factor 3 subunit 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523293 349.00
Check your price
  • eIF3e Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519411
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
eIF-3 p48; Viral integration site protein INT-6 homolog; Eukaryotic translation initiation factor 3 subunit 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519411 349.00
Check your price
  • eIF3g Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523319
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.025-0.1 µg/ml)
Alternate names:
eIF3 p44; eIF3 p42; eIF-3-delta; Eukaryotic translation initiation factor 3 subunit 4; Eukaryotic translation initiation factor 3 RNA-binding subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523319 349.00
Check your price
  • eIF3g Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523287
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (4.0-5 µg/ml)
Alternate names:
eIF3 p44; eIF3 p42; eIF-3-delta; Eukaryotic translation initiation factor 3 subunit 4; Eukaryotic translation initiation factor 3 RNA-binding subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523287 349.00
Check your price
  • eIF3j Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Horse, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523312
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
eIF3 p35; eIF-3-alpha; Eukaryotic translation initiation factor 3 subunit 1; eIF-3-alpha-A; Eukaryotic translation initiation factor 3 subunit 1-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523312 349.00
Check your price
  • eIF4A1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523273
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.25-1 µg/ml)
Alternate names:
ATP-dependent RNA helicase eIF4A-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523273 349.00
Check your price
  • eIF4A2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Chicken, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523264
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.025-0.05 µg/ml)
Alternate names:
ATP-dependent RNA helicase eIF4A-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523264 349.00
Check your price
  • eIF4A2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523260
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
ATP-dependent RNA helicase eIF4A-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523260 349.00
Check your price
  • eIF4B Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517071
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517071 309.00
Check your price
  • eIF4B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523261
  • Size 100 ug
Applications
Western Blot (0.25-0.50 µg/ml)
Alternate names:
EIF-4B, PRO1843
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523261 349.00
Check your price
  • eIF4B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523286
  • Size 100 ug
Applications
Western Blot (0.005-1 µg/ml)
Alternate names:
EIF-4B, PRO1843
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523286 349.00
Check your price
  • eIF5A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519191
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
eIF-4D; Rev-binding factor; Eukaryotic initiation factor 5A isoform 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519191 349.00
Check your price
  • ELF1 Polyclonal Antibody for Western Blot and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518072
  • Size 100 ug
Applications
Western Blot (0.3-1.0 ug/ml)
Alternate names:
E74-like factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518072 349.00
Check your price