Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  40  50  60  70  80  90  100  110  200  300  400  500  600  700  800  900  1000  1605 
481 - 510 of 48136 products displayed
  • F(ab')2-Donkey anti-Sheep IgG (H+L), HRP
  • Host Donkey
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # A24561
  • Size 1 mg
Applications
ELISA (1:500-1:20,000), Immunohistochemistry (1:500-1:20,000), Western Blot (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24561 149.00
Check your price
  • F(ab')2-Donkey anti-Sheep IgG (H+L), TRITC
  • Host Donkey
  • Target Species Sheep
  • Conjugate TRITC
  • Catalog # A24566
  • Size 1 mg
Applications
Immunofluorescence (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24566 104.00
Check your price
  • F(ab')2-Goat anti-Human IgA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24460
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24460 139.00
Check your price
  • F(ab')2-Goat anti-Human IgG (H+L)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24466
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24466 85.00
Check your price
  • F(ab')2-Goat anti-Human IgG (H+L)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 31165
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31165 104.00
Check your price
  • F(ab')2-Goat anti-Human IgG (H+L) Cross-Adsorbed
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24472
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24472 104.00
Check your price
  • F(ab')2-Goat anti-Human IgG / IgA / IgM
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24496
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24496 85.00
Check your price
  • F(ab')2-Goat anti-Human IgG Fc Cross-Adsorbed
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24478
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24478 129.00
Check your price
  • F(ab')2-Goat anti-Human IgG Fc gamma
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 31163
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31163 149.00
Check your price
  • F(ab')2-Goat anti-Human IgG, IgM, IgA (H+L), FITC
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # 31539
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31539 158.00
Check your price
  • F(ab')2-Goat anti-Human IgM Heavy Chain
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24484
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24484 125.00
Check your price
  • F(ab')2-Goat anti-Human IgM Heavy Chain Cross-Adsorbed
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A24490
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24490 136.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # A24514
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24514 83.25
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # A24526
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24526 158.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 31185
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31185 147.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Cross-Adsorbed, FITC
  • Host Goat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # 31565
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31565 174.00
Check your price
  • F(ab')2-Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # A24520
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24520 204.00
Check your price
  • F(ab')2-Goat anti-Mouse IgM (Mu)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 31178
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31178 186.00
Check your price
  • F(ab')2-Goat anti-Mouse IgM Cross-Adsorbed
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 31186
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31186 186.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L)
  • Host Goat
  • Target Species Rabbit
  • Conjugate Unconjugated
  • Catalog # A24533
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24533 73.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed
  • Host Goat
  • Target Species Rabbit
  • Conjugate Unconjugated
  • Catalog # A24539
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24539 94.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed
  • Host Goat
  • Target Species Rabbit
  • Conjugate Unconjugated
  • Catalog # 31239
  • Size 1 mg
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31239 158.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L) Cross-Adsorbed, FITC
  • Host Goat
  • Target Species Rabbit
  • Conjugate FITC
  • Catalog # 31636
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31636 173.00
Check your price
  • F(ab')2-Goat anti-Rabbit IgG (H+L), FITC
  • Host Goat
  • Target Species Rabbit
  • Conjugate FITC
  • Catalog # 31579
  • Size 1 mg
Applications
Flow Cytometry (1:50-1:200), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (1:50-1:200), Immunoprecipitation (1:50-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31579 147.00
Check your price
  • F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # A24551
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24551 169.00
Check your price
  • F(ab')2-Goat anti-Rat IgG (H+L) Highly Cross-Adsorbed
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # A24557
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24557 301.00
Check your price
  • F(ab')2-Rabbit anti-Goat IgG (H+L)
  • Host Rabbit
  • Target Species Goat
  • Conjugate Unconjugated
  • Catalog # A24448
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24448 115.00
Check your price
  • F(ab')2-Rabbit anti-Goat IgG (H+L) Cross-Adsorbed
  • Host Rabbit
  • Target Species Goat
  • Conjugate Unconjugated
  • Catalog # A24454
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A24454 139.00
Check your price
  • F(ab')2-Rabbit anti-Mouse IgG Cross-Adsorbed
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA533287
  • Size 1 mg
Applications
ELISA (1:500-1:1000), Immunohistochemistry (Frozen) (1:100 -1:500), Immunohistochemistry (Paraffin) (1:100 -1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533287 225.00
Check your price
  • F(ab')2-Rabbit anti-Mouse IgG, FITC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # PA184348
  • Size 1 mg
Applications
Flow Cytometry (1:25-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184348 370.00
Check your price