Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  158  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  190  200  210  220  230  240  250  260  270  300  400  500  600  700  800  900  1000  1100  1630 
5161 - 5190 of 48877 products displayed
  • SRPX2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520844
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Sushi-repeat protein upregulated in leukemia
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520844 349.00
Check your price
  • SRXN1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519037
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
SRXN1; sulfiredoxin 1 homolog (S. cerevisiae); C20orf139; Npn3; SRX1; YKL086W; dJ850E9.2; sulfiredoxin 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519037 349.00
Check your price
  • SRY Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517181
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Testis-determining factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517181 309.00
Check your price
  • SSX2IP Polyclonal Antibody for IF and ICC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518258
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent)
Alternate names:
SSX2-interacting protein; Afadin DIL domain-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518258 349.00
Check your price
  • ST2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523316
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (4.0-5 µg/ml)
Alternate names:
Protein ST2; Protein T1; Lymphocyte antigen 84; Fos-responsive gene 1 protein; Fit-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523316 349.00
Check your price
  • ST2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523206
  • Size 100ug
Applications
Western Blot (3-5 µg/ml)
Alternate names:
Protein ST2; Protein T1; Lymphocyte antigen 84
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523206 349.00
Check your price
  • ST2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520077
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Protein ST2; Protein T1; Lymphocyte antigen 84
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520077 349.00
Check your price
  • ST3GAL6 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520748
  • Size 100ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Sialyltransferase 10; ST3Gal VI; CMP-NeuAc:beta-galactoside alpha-2,3-sialyltransferase VI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520748 349.00
Check your price
  • STAG2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519217
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Stromal antigen 2; SCC3 homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519217 349.00
Check your price
  • STAG2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518722
  • Size 100ug
Applications
Western Blot (0.01 ug/ml)
Alternate names:
Stromal antigen 2; SCC3 homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518722 349.00
Check your price
  • STAM2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518500
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Hrs-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518500 349.00
Check your price
  • STAP2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517952
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Breast tumor kinase substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517952 349.00
Check your price
  • STAP2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517958
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.05-0.3 ug/ml)
Alternate names:
Breast tumor kinase substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517958 349.00
Check your price
  • STAT1 Polyclonal Antibody for Western Blot, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519858
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunoprecipitation (2-4 µg), Western Blot (1:1000-1:2000)
Alternate names:
Transcription factor ISGF-3 components p91/p84
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519858 349.00
Check your price
  • STAT1 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534504
  • Size 100ug
Applications
Immunofluorescence (20 ug/ml), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Transcription factor ISGF-3 components p91/p84
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534504 349.00
Check your price
  • STAT3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518562
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Acute-phase response factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518562 349.00
Check your price
  • STAT3 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19105
  • Size 100ug
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (4-8 ug), Western Blot (Assay-dependent)
Alternate names:
Acute-phase response factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19105 309.00
Check your price
  • STAT4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519271
  • Size 100ug
Applications
Western Blot (0.2-0.5 ug/ml)
Alternate names:
STAT4; signal transducer and activator of transcription 4; SLEB11; OTTHUMP00000163553
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519271 349.00
Check your price
  • STAT5 alpha Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517183
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517183 309.00
Check your price
  • Staufen Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521038
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
RP3-470L14.2, STAU; double-stranded RNA-binding protein Staufen homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521038 349.00
Check your price
  • STEAP1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520403
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Six-transmembrane epithelial antigen of prostate 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520403 349.00
Check your price
  • STEAP1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520404
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Six-transmembrane epithelial antigen of prostate 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520404 349.00
Check your price
  • STEAP2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520405
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Six-transmembrane epithelial antigen of prostate 2; SixTransMembrane protein of prostate 1; Protein up-regulated in metastatic prostate cancer; Prostate cancer-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520405 349.00
Check your price
  • STEAP3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520406
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Tumor suppressor-activated pathway protein 6; Six-transmembrane epithelial antigen of prostate 3; Protein nm1054; Dudulin-2; pHyde
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520406 349.00
Check your price
  • STEAP4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518782
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Tumor necrosis factor, alpha-induced protein 9; SixTransMembrane protein of prostate 2; Six-transmembrane epithelial antigen of prostate 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518782 349.00
Check your price
  • STEAP4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520407
  • Size 100ug
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Six-transmembrane epithelial antigen of prostate 4; Tumor necrosis factor-alpha-induced adipose-related protein; Dudulin-4; Tumor necrosis factor, alpha-induced protein 9; SixTransMembrane protein of prostate 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520407 349.00
Check your price
  • STIM1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520371
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520371 349.00
Check your price
  • STIM2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520372
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520372 349.00
Check your price
  • STIM2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520373
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520373 349.00
Check your price
  • STING Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523337
  • Size 100ug
Applications
Western Blot (5-10 µg/ml)
Alternate names:
Transmembrane protein 173; Mediator of IRF3 activation; Endoplasmic reticulum interferon stimulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523337 349.00
Check your price