Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  159  160  161  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  190  200  210  220  230  240  250  260  270  300  400  500  600  700  800  900  1000  1100  1611 
5191 - 5220 of 48310 products displayed
  • FBXW2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518189
  • Size 100 ug
Applications
Immunohistochemistry (2-3µg/ml), Western Blot (0.3-1.5 ug/ml)
Alternate names:
Protein MD6; F-box and WD-40 domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518189 349.00
Check your price
  • FceR1 alpha Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517074
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
IgE Fc receptor subunit alpha; Fc-epsilon RI-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517074 309.00
Check your price
  • FCGR2B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518362
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (3-6 µg/ml), Western Blot (0.3-1 µg/ml)
Alternate names:
Fc-gamma RII-c; CDw32; Fc-gamma RII-b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518362 349.00
Check your price
  • FCHO2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534390
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (2.5 ug/mL), Immunohistochemistry (2.5 ug/mL), Western Blot (1-2 ug/mL)
Alternate names:
FCHO2, FCH domain only 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534390 349.00
Check your price
  • Fe65 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518560
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Protein Fe65
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518560 349.00
Check your price
  • Feline Leukemia Virus Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA514707
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
FLV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514707 349.00
Check your price
  • FEM1A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518060
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Prostaglandin E receptor 4-associated protein; FEM1-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518060 349.00
Check your price
  • FEM1C Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517940
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
FEM1-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517940 349.00
Check your price
  • Ferritin Heavy Chain Polyclonal Antibody for Western Blot, IHC, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519058
  • Size 100 ug
Applications
Immunohistochemistry (Assay Dependent), Western Blot (0.03-0.1 ug/ml)
Alternate names:
Cell proliferation-inducing gene 15 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519058 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514733
  • Size 100 ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514733 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514694
  • Size 100 ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514694 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514736
  • Size 100 ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514736 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514693
  • Size 100 ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 0
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514693 349.00
Check your price
  • Ferritin Monoclonal Antibody for IF, ICC, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514735
  • Size 100 ug
Applications
ELISA (1 µg/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514735 349.00
Check your price
  • Ferritin Polyclonal Antibody for Western Blot, IHC, ELISA and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519059
  • Size 100 ug
Applications
ELISA (1-2ug/ml), Immunohistochemistry (3-6ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
FTL; ferritin, light polypeptide; MGC71996; L apoferritin; ferritin L subunit; ferritin L-chain; ferritin light chain; ferritin light polypeptide-like 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519059 349.00
Check your price
  • FEZ1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518608
  • Size 100 ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Zygin-1; Zygin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518608 349.00
Check your price
  • FEZ1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520396
  • Size 100 ug
Applications
Western Blot (1 ug/ml)
Alternate names:
Zygin-1; Zygin I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520396 349.00
Check your price
  • FEZ2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520456
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Zygin-2; Zygin II; Zygin-related protein types I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520456 349.00
Check your price
  • FEZ2 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520459
  • Size 100 ug
Applications
Immunofluorescence (5 µg/ml), Immunohistochemistry (5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Zygin-2; Zygin II; Zygin-related protein types I/II
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520459 349.00
Check your price
  • FGF20 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523480
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
FGF-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523480 349.00
Check your price
  • FGF21 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518811
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
; fibroblast growth factor 21; FGF21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518811 349.00
Check your price
  • FGF23 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518636
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Tumor-derived hypophosphatemia-inducing factor; Phosphatonin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518636 349.00
Check your price
  • FGF4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520483
  • Size 100 ug
Applications
Immunocytochemistry (2.5 µg/ml), Immunofluorescence (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Transforming protein KS3; Heparin-binding growth factor 4; Heparin secretory-transforming protein 1; K-fibroblast growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520483 349.00
Check your price
  • FGFR2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518914
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Keratinocyte growth factor receptor; K-sam
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518914 349.00
Check your price
  • FGFR3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534574
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (25 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Fibroblast Growth Factor Receptor 3, ACH, CEK2, HSFGFR3EX, JTK4, FGFR-3, fibroblast growth factor receptor 3 variant 4, hydroxyaryl-protein kinase, tyrosine kinase JTK4, FGF Receptor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534574 349.00
Check your price
  • FGL1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519204
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Liver fibrinogen-related protein 1; Hepatocyte-derived fibrinogen-related protein 1; Hepassocin; HP-041
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519204 349.00
Check your price
  • FGR Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA517976
  • Size 100 ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
p55-Fgr; Proto-oncogene c-Fgr
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517976 349.00
Check your price
  • FHL1 Isoform B Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518306
  • Size 100 ug
Applications
Western Blot (0.01-0.1 ug/ml)
Alternate names:
Skeletal muscle LIM-protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518306 349.00
Check your price
  • FHL1 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518305
  • Size 100 ug
Applications
Immunohistochemistry (2-2µg/ml), Western Blot (0.05-0.3 ug/ml)
Alternate names:
Skeletal muscle LIM-protein 1; RBP-associated molecule 14-1; KyoT
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518305 349.00
Check your price
  • FHL2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518259
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Skeletal muscle LIM-protein 3; LIM domain protein DRAL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518259 349.00
Check your price