Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  70  80  90  100  110  120  130  140  150  162  163  164  165  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  200  210  220  230  240  250  260  270  300  400  500  600  700  800  900  1000  1100  1630 
5281 - 5310 of 48882 products displayed
  • TCCR Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519985
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
ZcytoR1; Type I T-cell cytokine receptor; Cytokine receptor-like 1; Cytokine receptor WSX-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519985 349.00
Check your price
  • TCF12 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521039
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Transcription factor HTF-4; Salivary-specific cAMP response element-binding protein alpha; E-box-binding protein; DNA-binding protein HTF4; Class B basic helix-loop-helix protein 20; Class A helix-loop-helix transcription factor ME1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521039 349.00
Check your price
  • TCF19 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523385
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Transcription factor SC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523385 349.00
Check your price
  • TCF19 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131982
  • Size 100ug
Applications
ELISA (1:32,000), Western Blot (1-2 µg/ml)
Alternate names:
Transcription factor SC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131982 399.00
Check your price
  • TCF19 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518190
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Transcription factor SC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518190 349.00
Check your price
  • TCF2 Polyclonal Antibody for Western Blot, ICC and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518642
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Variant hepatic nuclear factor 1; Transcription factor 2; Homeoprotein LFB3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518642 349.00
Check your price
  • TCF3 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518641
  • Size 100ug
Applications
Immunohistochemistry (3-5µg/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
Transcription factor ITF-1; Transcription factor 3; Kappa-E2-binding factor; Immunoglobulin transcription factor 1; Immunoglobulin enhancer-binding factor E12/E47; Class B basic helix-loop-helix protein 21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518641 349.00
Check your price
  • TCF3 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520900
  • Size 100ug
Applications
Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 ug/ml)
Alternate names:
Transcription factor ITF-1; Transcription factor 3; Kappa-E2-binding factor; Immunoglobulin transcription factor 1; Immunoglobulin enhancer-binding factor E12/E47; Class B basic helix-loop-helix protein 21; Transcription factor A1; Transcription regulator Pan; Pancreas specific transcription factor 1c
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520900 349.00
Check your price
  • TCF7L2 Polyclonal Antibody for Western Blot, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA110041
  • Size 100ug
Applications
Immunocytochemistry (1 ug/ml), Immunohistochemistry (Paraffin) (10 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
T-cell-specific transcription factor 4; HMG box transcription factor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110041 249.00
Check your price
  • TCFL5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518588
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
HPV-16 E2-binding protein 1; Cha transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518588 349.00
Check your price
  • TCHHL1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518970
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
S100 calcium-binding protein A17; Protein S100-A17; Basalin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518970 349.00
Check your price
  • TCIRG1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534419
  • Size 100ug
Applications
Western Blot (0.5 ug/mL)
Alternate names:
Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3; T-cell immune response cDNA7 protein; T-cell immune regulator 1; Osteoclastic proton pump 116 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534419 349.00
Check your price
  • TCL1A Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518689
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Protein p14 TCL1; Oncogene TCL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518689 349.00
Check your price
  • TCP-1 gamma Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519134
  • Size 100ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
mTRiC-P5; Matricin; CCT-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519134 349.00
Check your price
  • TCP1 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518251
  • Size 100ug
Applications
Immunohistochemistry (1-2µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
CCT-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518251 349.00
Check your price
  • TCR alpha/beta Monoclonal Antibody, Biotin conjugate for IHC (F) and Flow
  • Host Armenian Hamster
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517649
  • Size 100ug
Applications
Flow Cytometry (1.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Tcra, Tcrb, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517649 109.00
Check your price
  • TCR alpha/beta Monoclonal Antibody, FITC conjugate for Flow
  • Host Armenian Hamster
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517651
  • Size 100ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
Tcra, Tcrb, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517651 119.00
Check your price
  • TCR alpha/beta Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110451
  • Size 100ug
Applications
Flow Cytometry (2-4 ug/ml)
Alternate names:
Tcrb, Tcra, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110451 159.00
Check your price
  • TCR alpha/beta Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110453
  • Size 100ug
Applications
Flow Cytometry (1:300)
Alternate names:
Tcrb, Tcra, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110453 249.00
Check your price
  • TCR alpha/beta Monoclonal Antibody, Biotin conjugate for IHC (F) and Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517542
  • Size 100ug
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Tcrb, Tcra, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517542 159.00
Check your price
  • TCR alpha/beta Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517544
  • Size 100ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Tcrb, Tcra, FLJ22602, MGC117436, MGC22624, MGC23964, MGC71411, TCRA, TCRD, TRA, T-cell antigen receptor, alpha polypeptide, T-cell receptor, TCR beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517544 159.00
Check your price
  • TCR DO11.10 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA517660
  • Size 100ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517660 399.00
Check your price
  • TCR DO11.10 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MA517661
  • Size 100ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517661 159.00
Check your price
  • TCR DO11.10 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA517662
  • Size 100ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517662 139.00
Check your price
  • TCR DO11.10 Monoclonal Antibody, PE-Cyanine5 conjugate for Flow
  • Host Mouse
  • Target Species Mouse
  • Conjugate PE-Cyanine5
  • Catalog # MA517664
  • Size 100ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517664 425.00
Check your price
  • TCTEX1D1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534444
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/mL), Western Blot (1 ug/mL)
Alternate names:
TCTEX1D1, TCTEX1 domain containing 1, FLJ40873, MGC125768
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534444 349.00
Check your price
  • TCTEX1D2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534445
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/mL), Western Blot (1 ug/mL)
Alternate names:
TCTEX1D2, TCTEX1 domain containing 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534445 349.00
Check your price
  • TCTEX1D3 Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534446
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1 ug/mL)
Alternate names:
T-complex-associated testis-expressed protein 3; T-complex testis-specific protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534446 349.00
Check your price
  • TCTEX1D4 Polyclonal Antibody for Western Blot, IF and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534447
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Immunohistochemistry (20 ug/ml), Western Blot (1 ug/mL)
Alternate names:
Tctex-2-beta; Protein N22.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534447 349.00
Check your price
  • TCTN1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520983
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
UNQ9369/PRO34160, JBTS13, TECT1; tectonic-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520983 349.00
Check your price