Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  166  167  168  169  170  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1630 
5401 - 5430 of 48877 products displayed
  • TLR2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523114
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Toll/interleukin-1 receptor-like protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523114 349.00
Check your price
  • TLR3 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516273
  • Size 100ug
Applications
Flow Cytometry (1 µg/10^6 cells), Western Blot (2-4 µg/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516273 349.00
Check your price
  • TLR3 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516184
  • Size 100ug
Applications
Flow Cytometry (2-4 µg/1x10^6 cells), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (2 µg/1x10^6 cells), Western Blot (1-3 µg/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516184 349.00
Check your price
  • TLR3 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Bovine, Human, Marsupial, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523292
  • Size 100ug
Applications
Flow Cytometry (1 µg/10^6 cells), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523292 349.00
Check your price
  • TLR3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523349
  • Size 100ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523349 349.00
Check your price
  • TLR3 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523510
  • Size 100ug
Applications
Flow Cytometry (1 µg/10^6 cells), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (4-6 µg/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523510 349.00
Check your price
  • TLR3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520184
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (2 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520184 349.00
Check your price
  • TLR3 Polyclonal Antibody for Western Blot, IHC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA523105
  • Size 100ug
Applications
Flow Cytometry (1 µg/10^6 cells), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:50), Western Blot (1-3 µg/ml)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523105 349.00
Check your price
  • TLR3 Polyclonal Antibody, FITC conjugate for Flow
  • Host Rabbit
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # PA523108
  • Size 100ug
Applications
Flow Cytometry (1 µg/10^6 cells)
Alternate names:
Toll-like Receptor 3, IIAE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523108 349.00
Check your price
  • TLR4 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), Flow, ChIP and IV
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA516216
  • Size 100ug
Applications
Flow Cytometry (0.5-2 µg/10^6 cells), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:500), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516216 349.00
Check your price
  • TLR4 Monoclonal Antibody for Western Blot, Flow, IP, Neu, FN and IV
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # MA516205
  • Size 100ug
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2-3 µg/10^6 cells), Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunoprecipitation (1:10-1:500), Neutralization (Assay-Dependent)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516205 349.00
Check your price
  • TLR4 Monoclonal Antibody for Flow and IP
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516210
  • Size 100ug
Applications
Flow Cytometry (1-2 µg/10^6 cells), Immunoprecipitation (1:10-1:500)
Alternate names:
Toll-like Receptor 4, ARMD10, TLR-4, TOLL, hToll, homolog of Drosophila toll
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516210 349.00
Check your price
  • TLR4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523270
  • Size 100ug
Applications
Western Blot (2-5 µg/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523270 349.00
Check your price
  • TLR4 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523123
  • Size 100ug
Applications
Immunoprecipitation (1:10-1:500), Western Blot (2-5 µg/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523123 349.00
Check your price
  • TLR4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523124
  • Size 100ug
Applications
Immunocytochemistry (2-5 µg/ml), Immunofluorescence (2-5 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (2-5 µg/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523124 349.00
Check your price
  • TLR4 Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523125
  • Size 100ug
Applications
Flow Cytometry (1 µg/10^6 cells), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:50), Western Blot (1-3 µg/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523125 349.00
Check your price
  • TLR4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Horse, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523284
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.1-0.5 µg/ml)
Alternate names:
hToll
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523284 349.00
Check your price
  • TLR5 Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516237
  • Size 100ug
Applications
Flow Cytometry (0.5-1 µg/10^6 cells), Immunohistochemistry (Frozen) (10 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Toll/interleukin-1 receptor-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516237 349.00
Check your price
  • TLR5 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516233
  • Size 100ug
Applications
Flow Cytometry (1-3 µg/10^6 cells in 100 µl), Western Blot (1-3 µg/ml)
Alternate names:
Toll/interleukin-1 receptor-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516233 349.00
Check your price
  • TLR5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520185
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Toll/interleukin-1 receptor-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520185 349.00
Check your price
  • TLR5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520186
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Toll/interleukin-1 receptor-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520186 349.00
Check your price
  • TLR6 Monoclonal Antibody for IHC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516173
  • Size 100ug
Applications
Flow Cytometry (3 µg/10^6 cells), Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
Toll-like Receptor 6, CD antigen 286
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516173 349.00
Check your price
  • TLR6 Polyclonal Antibody for Western Blot, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520188
  • Size 100ug
Applications
Immunocytochemistry (2 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Toll-like Receptor 6, CD antigen 286
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520188 349.00
Check your price
  • TLR6 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520187
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Toll-like Receptor 6, CD antigen 286
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520187 349.00
Check your price
  • TLR7 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516247
  • Size 100ug
Applications
Flow Cytometry (1-2 µg/10^6 cells), Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Western Blot (10 µg/ml)
Alternate names:
UNQ248/PRO285
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516247 349.00
Check your price
  • TLR7 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523174
  • Size 100ug
Applications
Flow Cytometry (1-2 µg/10^6 cells), Western Blot (0.5-2 µg/ml)
Alternate names:
UNQ248/PRO285
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523174 349.00
Check your price
  • TLR7 Polyclonal Antibody for IHC, IHC (F) and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523488
  • Size 100ug
Applications
Flow Cytometry (0.5-2 µg/1x10^6 cells), Immunohistochemistry (Frozen) (1:20)
Alternate names:
UNQ248/PRO285
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523488 349.00
Check your price
  • TLR7 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520053
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
RP23-139P21.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520053 349.00
Check your price
  • TLR7 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523126
  • Size 100ug
Applications
Flow Cytometry (2-5 µg/1x10^6 cells), Immunocytochemistry (1:10-1:2000), Immunofluorescence (1:10-1:2000), Immunohistochemistry (Frozen) (1:10-1:2000), Western Blot (1-3 µg/ml)
Alternate names:
UNQ248/PRO285
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523126 349.00
Check your price
  • TLR7 Polyclonal Antibody, FITC conjugate for IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate FITC
  • Catalog # PA523127
  • Size 100ug
Applications
Flow Cytometry (2-5 µg/1x10^6 cells), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent)
Alternate names:
UNQ248/PRO285
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523127 349.00
Check your price