Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  171  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1622 
5551 - 5580 of 48655 products displayed
  • HHATL Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534355
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (2.5 ug/mL), Immunohistochemistry (2.5 ug/mL), Western Blot (1 ug/mL)
Alternate names:
Hedgehog acyltransferase-like protein; Glycerol uptake/transporter homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534355 349.00
Check your price
  • HINT1 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520613
  • Size 100 ug
Applications
Immunocytochemistry (2.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Protein kinase C-interacting protein 1; Protein kinase C inhibitor 1; Adenosine 5'-monophosphoramidase; 17 kDa inhibitor of protein kinase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520613 349.00
Check your price
  • HIP2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518121
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Ubiquitin-protein ligase; Ubiquitin-conjugating enzyme E2-25 kDa; Ubiquitin carrier protein; Huntingtin-interacting protein 2; E2 ubiquitin-conjugating enzyme K
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518121 349.00
Check your price
  • HIP55 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518046
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
SH3 domain-containing protein 7; HPK1-interacting protein of 55 kDa; Drebrin-F; Cervical mucin-associated protein; Cervical SH3P7; Actin-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518046 349.00
Check your price
  • HIPK3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523171
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Nuclear body-associated kinase 3; Fas-interacting serine/threonine-protein kinase; Androgen receptor-interacting nuclear protein kinase; Homolog of protein kinase YAK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523171 349.00
Check your price
  • HIPPI Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519110
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
HIP1-interacting protein; MHS4R2; Estrogen-related receptor beta-like protein 1; Dermal papilla-derived protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519110 349.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523450
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (4-7 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523450 349.00
Check your price
  • Histone H2A.X Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523465
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (2-4 µg/ml)
Alternate names:
Histone H2A.X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523465 349.00
Check your price
  • Histone H3.3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Chicken, Fruit fly, Horse, Human, Marsupial, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523288
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.25-0.5 µg/ml)
Alternate names:
Histone H3, H3.3A, H3.3B, H3F3, H3F3A, H3F3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523288 349.00
Check your price
  • HIV1 gp120 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520488
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 120, HIV-1 Glycoprotein 120
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520488 349.00
Check your price
  • HIV1 gp120 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520584
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 120, HIV-1 Glycoprotein 120
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520584 349.00
Check your price
  • HIV1 gp41 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17590
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 41, HIV-1 Glycoprotein 41
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17590 309.00
Check your price
  • HIV1 nef Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171502
  • Size 100 ug
Applications
ELISA (1:5000), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV-1 Nef region 150-158, BRU
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171502 325.00
Check your price
  • HIV1 nef Monoclonal Antibody for Western Blot, IHC and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171505
  • Size 100 ug
Applications
ELISA (1:10,000), Immunohistochemistry (Assay dependent), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 Nef region 171-190, BRU
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171505 325.00
Check your price
  • HIV1 nef Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171501
  • Size 100 ug
Applications
ELISA (1:5000), Immunocytochemistry (Assay Dependent), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV-1 Nef region 35-50, HXB2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171501 349.00
Check your price
  • HIV1 nef Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171503
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 Nef region 163-173, HIV1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171503 399.00
Check your price
  • HIV1 nef Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171500
  • Size 100 ug
Applications
ELISA (1:5000), Western Blot (1:500-1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 Nef region 31-35
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171500 325.00
Check your price
  • HIV1 nef Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171506
  • Size 100 ug
Applications
ELISA (1:25,000), Western Blot (1:5000)
Alternate names:
Human immunodeficiency virus, HIV-1 Nef region 195-205, HIV1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171506 325.00
Check your price
  • HIV1 nef Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171507
  • Size 100 ug
Applications
ELISA (1:5000), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV-1 Nef region 62-68
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171507 349.00
Check your price
  • HIV1 p17 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17588
  • Size 100 ug
Applications
ELISA (1:1000), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17588 275.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17586
  • Size 100 ug
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17586 309.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171521
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:500-1:000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 region 80-90
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171521 399.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171520
  • Size 100 ug
Applications
ELISA (1:25000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 region 74-84
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171520 399.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171515
  • Size 100 ug
Applications
ELISA (1:64,000), Immunofluorescence (1:1000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 region 48-62
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171515 399.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171518
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171518 349.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171516
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 p24 C-terminus 165-231
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171516 349.00
Check your price
  • HIV1 p24 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171517
  • Size 100 ug
Applications
ELISA (1:10,000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV1gp1, HIV p24
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171517 349.00
Check your price
  • HIV1 p55 Monoclonal Antibody for Western Blot, IHC, Flow and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17587
  • Size 100 ug
Applications
ELISA (1:1000), Flow Cytometry (1:1000), Immunohistochemistry (1:1000), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV p55, HIV p17
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17587 309.00
Check your price
  • HIV1 Rev Group M Monoclonal Antibody for Western Blot, IF, IHC, IP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171514
  • Size 100 ug
Applications
ELISA (1:10000), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV group M, HIV-1 Rev region 33-48, Regulator of Virion
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171514 399.00
Check your price
  • HIV1 Tat Monoclonal Antibody for Western Blot, IHC, ChIP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171508
  • Size 100 ug
Applications
ChIP assay (1-3 µl), ELISA (1:5000), Immunohistochemistry (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 Tat region 2-9, HBX2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171508 399.00
Check your price