Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  80  90  100  110  120  130  140  150  160  172  173  174  175  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  210  220  230  240  250  260  270  280  300  400  500  600  700  800  900  1000  1100  1622 
5581 - 5610 of 48655 products displayed
  • HIV1 Tat Monoclonal Antibody for Western Blot, ChIP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171510
  • Size 100 ug
Applications
ChIP assay (1-3 µl), ELISA (1:2000), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV-1 Tat region 2-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171510 399.00
Check your price
  • HIV1 Tat Monoclonal Antibody for Western Blot, ChIP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171509
  • Size 100 ug
Applications
ChIP assay (1-3 µl), ELISA (1:2000), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV-1 Tat region 6 -12, BH10
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171509 349.00
Check your price
  • HIV1 Tat Monoclonal Antibody for Western Blot, ChIP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171512
  • Size 100 ug
Applications
ChIP assay (1-3 µl), ELISA (1:5000), Western Blot (1:500-1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 Tat region 71-81
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171512 319.00
Check your price
  • HIV1 Tat Monoclonal Antibody for Western Blot, ChIP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171511
  • Size 100 ug
Applications
ChIP assay (1-3 µl), ELISA (Assay Dependent), Western Blot (1:500)
Alternate names:
Human immunodeficiency virus, HIV-1 Tat region 47-58, HBX2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171511 399.00
Check your price
  • HIV1 Tat Monoclonal Antibody for Western Blot, ChIP and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171513
  • Size 100 ug
Applications
ChIP assay (1-3 µl), ELISA (1:5000), Western Blot (1:500-1:1000)
Alternate names:
Human immunodeficiency virus, HIV-1 Tat region 59-67
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171513 325.00
Check your price
  • HIV2 gp36 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17589
  • Size 100 ug
Applications
ELISA (1:500), Western Blot (1:1000)
Alternate names:
Human immunodeficiency virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17589 309.00
Check your price
  • HLA-A2 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110344
  • Size 100 ug
Applications
Flow Cytometry (5-10 ug/ml), Immunoprecipitation (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110344 225.00
Check your price
  • HLA-A2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516587
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516587 425.00
Check your price
  • HLA-ABC Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17660
  • Size 100 ug
Applications
Flow Cytometry (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17660 275.00
Check your price
  • HLA-ABC Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate
  • Conjugate PE
  • Catalog # MA110346
  • Size 100 ug
Applications
Flow Cytometry (1-2 ug/ml)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110346 349.00
Check your price
  • HLA-DQA1/DQA2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519388
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
MHC class II DQA1; HLA-DCA; DC-alpha; DC-1 alpha chain; MHC class II DQA2; HLA-DQA1; HLA class II histocompatibility antigen, DQ(6) alpha chain; DX alpha chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519388 349.00
Check your price
  • HLA-DR Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Goat, Human, Sheep
  • Conjugate FITC
  • Catalog # MA516502
  • Size 100 ug
Applications
Flow Cytometry (1:5-1:10)
Alternate names:
MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516502 475.00
Check your price
  • HLA-DR Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17659
  • Size 100 ug
Applications
Flow Cytometry (1:1000)
Alternate names:
MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17659 275.00
Check your price
  • HLA-DR Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110354
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml)
Alternate names:
MHC class II antigen DRA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110354 349.00
Check your price
  • HLA-DR Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA518020
  • Size 100 ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
MHC class II antigen DRA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518020 159.00
Check your price
  • HLA-DR Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA518021
  • Size 100 ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
MHC class II antigen DRA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518021 159.00
Check your price
  • HLA-DR Monoclonal Antibody, Biotin conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA518017
  • Size 100 ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
MHC class II antigen DRA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518017 159.00
Check your price
  • HLA-DR Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA518018
  • Size 100 ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
MHC class II antigen DRA
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518018 159.00
Check your price
  • HLA-DR/DP Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110350
  • Size 100 ug
Applications
Flow Cytometry (1:400)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110350 249.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516598
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516598 325.00
Check your price
  • HLA-DR/DP/DQ Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA516983
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
MHC class II antigen DPB1; HLA class II histocompatibility antigen, DP(W4) beta chain; MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516983 319.00
Check your price
  • HLA-DR/DQ Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Goat, Human, Sheep
  • Conjugate FITC
  • Catalog # MA516518
  • Size 100 ug
Applications
Flow Cytometry (1:10)
Alternate names:
MHC class II antigen DRB1*15; DW2.2/DR2.2; MHC class II antigen DRB1*16; MHC class II antigen DQB1; MHC class II antigen DRB1*3; Clone P2-beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516518 499.00
Check your price
  • HLA-E Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110355
  • Size 100 ug
Applications
Flow Cytometry (2 ug/ml)
Alternate names:
MHC class I antigen E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110355 349.00
Check your price
  • HLA-G (soluble) Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110358
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (1-10 ug/ml), Immunohistochemistry (Paraffin, Frozen) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110358 199.00
Check your price
  • HLA-G (soluble) Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110364
  • Size 100 ug
Applications
Flow Cytometry (5 ug/ml)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110364 309.00
Check your price
  • HLA-G (soluble) Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110357
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/ml), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (10 ug/ml), Immunohistochemistry (Paraffin) (10 ug/ml), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110357 199.00
Check your price
  • HLA-G (soluble) Monoclonal Antibody, Biotin conjugate for Western Blot, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110360
  • Size 100 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (1 ug/ml), Immunocytochemistry (Assay Dependent), Western Blot (1:1000)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110360 275.00
Check your price
  • HLA-G (soluble) Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110368
  • Size 100 ug
Applications
Flow Cytometry (10 ug/ml)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110368 349.00
Check your price
  • HLA-G Monoclonal Antibody for ICC, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110359
  • Size 100 ug
Applications
ELISA (Assay-Dependent), Flow Cytometry (5-6 ug/ml), Immunocytochemistry (Assay-Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Immunoprecipitation (Assay-Dependent)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110359 159.00
Check your price
  • HLA-G Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110366
  • Size 100 ug
Applications
Flow Cytometry (1:200)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110366 275.00
Check your price