Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  90  100  110  120  130  140  150  160  170  176  177  178  179  180  181  182  183  184  185  186  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  210  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1630 
5701 - 5730 of 48881 products displayed
  • West Nile Virus Glycoprotein M Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA523102
  • Size 100ug
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (1 µg/ml), Immunoprecipitation (1:10-1:500), Western Blot (0.5-2 µg/ml)
Alternate names:
WNV envelope, West Nile envelope, WNV Glycoprotein M
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523102 349.00
Check your price
  • WFDC2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519293
  • Size 100ug
Applications
Western Blot (0.2-1 ug/ml)
Alternate names:
Putative protease inhibitor WAP5; Major epididymis-specific protein E4; Epididymal secretory protein E4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519293 349.00
Check your price
  • WHSC2 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA517199
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Wolf-Hirschhorn syndrome candidate 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517199 309.00
Check your price
  • WIPF1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518752
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Wiskott-Aldrich syndrome protein-interacting protein; Protein PRPL-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518752 349.00
Check your price
  • WIPI1 Polyclonal Antibody for Western Blot and IF
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534452
  • Size 100ug
Applications
Immunofluorescence (20 ug/mL), Western Blot (1-2 ug/mL)
Alternate names:
WD40 repeat protein interacting with phosphoinositides of 49 kDa; Atg18 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534452 349.00
Check your price
  • WIPI2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA534478
  • Size 100ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
WIPI49-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534478 349.00
Check your price
  • WISP1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518832
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Wnt-1-induced secreted protein; CCN family member 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518832 349.00
Check your price
  • WIZ Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519066
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Zinc finger protein 803; Widely-interspaced zinc finger-containing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519066 349.00
Check your price
  • WIZ Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521082
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Widely-interspaced zinc finger-containing protein; Zinc finger protein 803
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521082 349.00
Check your price
  • WNT10A Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520542
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
OODD, SSPS, STHAG4; protein Wnt-10a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520542 349.00
Check your price
  • WNT10B Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520530
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (2-4 ug/ml)
Alternate names:
Protein Wnt-12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520530 349.00
Check your price
  • WNT4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19118
  • Size 100ug
Applications
Western Blot (0.03-0.1 µg/ml)
Alternate names:
WNT4; wingless-type MMTV integration site family, member 4; WNT-4; WNT-4 protein precursor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19118 309.00
Check your price
  • Wnt5A Polyclonal Antibody for Western Blot, IHC (P)
  • Host Rabbit
  • Target Species Mouse, Human, Rat, Bovine, Non-human primate
  • Conjugate
  • Catalog # PA523178
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
hWNT5A, WNT-5A protein, protein Wnt-5a
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523178 Not Orderable Online
Check your price
  • WNT9B Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519149
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Protein Wnt-15; Protein Wnt-14b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519149 349.00
Check your price
  • WT1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518548
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
WT33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518548 349.00
Check your price
  • XAB2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517947
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (3-4 ug/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
XPA-binding protein 2; Protein HCNP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517947 349.00
Check your price
  • XAF1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520090
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-4 µg/ml)
Alternate names:
BIRC4-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520090 349.00
Check your price
  • XAF1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520038
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
BIRC4-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520038 349.00
Check your price
  • XBP1 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515767
  • Size 100ug
Applications
Immunocytochemistry (2 ug/ml), Immunofluorescence (4 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Hepatocarcinogenesis-related transcription factor; Tax-responsive element-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515767 349.00
Check your price
  • XBP1 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515768
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (4 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Hepatocarcinogenesis-related transcription factor; Tax-responsive element-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515768 349.00
Check your price
  • XBP1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518940
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Tax-responsive element-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518940 349.00
Check your price
  • XBP1 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520217
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Hepatocarcinogenesis-related transcription factor; Tax-responsive element-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520217 349.00
Check your price
  • XIAP Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520067
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (2 µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
X-linked inhibitor of apoptosis protein; Inhibitor of apoptosis protein 3; IAP-like protein; Baculoviral IAP repeat-containing protein 4; IAP homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520067 349.00
Check your price
  • XIAP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522812
  • Size 100ug
Applications
Western Blot (1:100-1:2000)
Alternate names:
X-linked inhibitor of apoptosis protein; Inhibitor of apoptosis protein 3; IAP-like protein; Baculoviral IAP repeat-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522812 349.00
Check your price
  • XLF Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518584
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
XRCC4-like factor; Protein cernunnos
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518584 349.00
Check your price
  • XPNPEP1 Polyclonal Antibody for Western Blot, ELISA and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA519139
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
X-prolyl aminopeptidase 1, soluble; X-Pro aminopeptidase 1; Soluble aminopeptidase P; Cytosolic aminopeptidase P; Aminoacylproline aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519139 349.00
Check your price
  • XRCC2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521279
  • Size 100ug
Applications
Western Blot (1:500-1:3000)
Alternate names:
X-ray repair cross-complementing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521279 349.00
Check your price
  • XRCC3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521280
  • Size 100ug
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
X-ray repair cross-complementing protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521280 349.00
Check your price
  • YAP1 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA517200
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Yes-associated protein YAP65 homolog; Protein yorkie homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517200 309.00
Check your price
  • YBX2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521014
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
MSY2 homolog; Germ cell-specific Y-box-binding protein; DNA-binding protein C; Contrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521014 349.00
Check your price