Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  100  110  120  130  140  150  160  170  180  187  188  189  190  191  192  193  194  195  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  220  230  240  250  260  270  280  290  300  400  500  600  700  800  900  1000  1100  1200  1630 
6031 - 6060 of 48882 products displayed
  • AGR2 Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141170
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Western Blot (1:100)
Alternate names:
Secreted cement gland protein XAG-2 homolog; HPC8; Protein Gob-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141170 349.00
Check your price
  • Agrin Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and RIA
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA125500
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:1000), Immunofluorescence (1:50-1:1000), Immunohistochemistry (Frozen) (Assay Dependent), Radioimmune Assays (1 µg/ml), Western Blot (2 µg/ml)
Alternate names:
RP23-139J18.2, Agrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125500 399.00
Check your price
  • Agrin Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA120380
  • Size 100µg
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (Assay dependent)
Alternate names:
RP23-139J18.2, Agrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120380 425.00
Check your price
  • Agrin Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA120378
  • Size 100µg
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (2 ug/ml), Immunohistochemistry (Assay-dependent), Western Blot (2 µg/ml)
Alternate names:
RP23-139J18.2, Agrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120378 399.00
Check your price
  • AgRP Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547895
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Agrt, Art
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547895 349.00
Check your price
  • AgRP Polyclonal Antibody for Western Blot and IHC (F)
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547831
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Agrt, Art
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547831 375.00
Check your price
  • AGXT Monoclonal Antibody for ICC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # A21964
  • Size 100µg
Applications
ELISA (5 ug/ml), Immunocytochemistry (5 ug/ml), Immunoprecipitation (Assay Dependent)
Alternate names:
Alanine--glyoxylate aminotransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21964 324.00
Check your price
  • AHA1 Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521096
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (1-5 µg/ml), Western Blot (Assay-Dependent)
Alternate names:
p38
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521096 309.00
Check your price
  • AHNAK Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA110050
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Desmoyokin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110050 249.00
Check your price
  • AID Monoclonal Antibody for Western Blot, IF, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 392500
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cytidine aminohydrolase; Activation-induced cytidine deaminase
Catalog # Size Price (AUD) Your Price (AUD) Qty
392500 309.00
Check your price
  • AIF Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bovine, C. elegans, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 456200
  • Size 100µg
Applications
Immunocytochemistry (5-10 ug/ml), Immunofluorescence (Assay Dependent), Western Blot (2-5 ug/ml)
Alternate names:
Programmed cell death protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
456200 330.00
Check your price
  • AIF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112503
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Programmed cell death protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112503 325.00
Check your price
  • AIF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112501
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Programmed cell death protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112501 325.00
Check your price
  • AIF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112502
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Programmed cell death protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112502 325.00
Check your price
  • AIF Polyclonal Antibody for Western Blot and IHC (P)
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA548092
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Programmed cell death protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548092 309.00
Check your price
  • AIF Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA548108
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1-1 µg/ml)
Alternate names:
Programmed cell death protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548108 309.00
Check your price
  • AIF Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130677
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500-1:2000)
Alternate names:
Programmed cell death protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130677 449.00
Check your price
  • AIM Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112793
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
SP-alpha; IgM-associated peptide; CT-2; Apoptosis inhibitory 6; Apoptosis inhibitor expressed by macrophages
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112793 325.00
Check your price
  • AIM Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA112794
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
SP-alpha; IgM-associated peptide; CT-2; Apoptosis inhibitory 6; Apoptosis inhibitor expressed by macrophages
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112794 309.00
Check your price
  • AIP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518715
  • Size 100µg
Applications
Western Blot (0.02-0.06 ug/ml)
Alternate names:
Aryl-hydrocarbon receptor-interacting protein; Immunophilin homolog ARA9; HBV X-associated protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518715 349.00
Check your price
  • AIP Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1514
  • Size 100µg
Applications
Western Blot (1 ug/ml)
Alternate names:
Aryl-hydrocarbon receptor-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1514 349.00
Check your price
  • AIRE Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524276
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524276 275.00
Check your price
  • AJAP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538044
  • Size 100µg
Applications
Western Blot (1-2 µg/ml)
Alternate names:
Membrane protein shrew-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538044 309.00
Check your price
  • AKAP10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538529
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein kinase A-anchoring protein 10; Dual specificity A kinase-anchoring protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538529 309.00
Check your price
  • AKAP11 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539868
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Protein kinase A-anchoring protein 11; A-kinase anchor protein 220 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539868 309.00
Check your price
  • AKAP13 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538670
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
p47; Protein kinase A-anchoring protein 13; Non-oncogenic Rho GTPase-specific GTP exchange factor; Lymphoid blast crisis oncogene; Human thyroid-anchoring protein 31; Guanine nucleotide exchange factor Lbc; Breast cancer nuclear receptor-binding auxiliary protein; AKAP-Lbc
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538670 309.00
Check your price
  • AKAP14 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538721
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Protein kinase A-anchoring protein 14; A-kinase anchor protein 28 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538721 309.00
Check your price
  • AKAP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538832
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein kinase A-anchoring protein 2; AKAP-KL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538832 309.00
Check your price
  • AKAP3 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131348
  • Size 100µg
Applications
ELISA (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
Sperm oocyte-binding protein; Protein kinase A-anchoring protein 3; Fibrousheathin-1; Fibrousheathin I; Fibrous sheath protein of 95 kDa; Cancer/testis antigen 82; A-kinase anchor protein 110 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131348 475.00
Check your price
  • AKAP4 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538015
  • Size 100µg
Applications
Immunofluorescence (20 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Protein kinase A-anchoring protein 4; Major sperm fibrous sheath protein; A-kinase anchor protein 82 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538015 309.00
Check your price