Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  196  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6301 - 6330 of 48881 products displayed
  • ARMX1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA538812
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
ARM protein lost in epithelial cancers on chromosome X 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538812 309.00
Check your price
  • ARMX2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538716
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
ARM protein lost in epithelial cancers on chromosome X 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538716 309.00
Check your price
  • ARMX3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538819
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
ARM protein lost in epithelial cancers on chromosome X 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538819 309.00
Check your price
  • Aromatase Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IHC (PFA)
  • Host Rabbit
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # PA519633
  • Size 100µg
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Immunohistochemistry (Frozen) (1:50), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunohistochemistry (PFA fixed) (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Estrogen synthase; Cytochrome P450 19A1; Cytochrome P-450AROM; CYPXIX
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519633 349.00
Check your price
  • Arp2 Polyclonal Antibody for Western Blot, IF and IP
  • Host Rabbit
  • Target Species Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA519760
  • Size 100µg
Applications
Immunofluorescence (1 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Actin-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519760 349.00
Check your price
  • ARPP21 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539069
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Thymocyte cAMP-regulated phosphoprotein; Regulator of calmodulin signaling
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539069 309.00
Check your price
  • ARRDC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538848
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538848 309.00
Check your price
  • ARRDC2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538850
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538850 309.00
Check your price
  • ARRDC3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538851
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Liver regeneration-related protein LRRG00048; TBP-2-like inducible membrane protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538851 309.00
Check your price
  • ARRDC4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538852
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Liver regeneration-related protein LRRG041/LRRGT00117
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538852 309.00
Check your price
  • ARSK Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539032
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Telethon sulfatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539032 309.00
Check your price
  • Artemin Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547350
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Neublastin; Enovin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547350 349.00
Check your price
  • Artemin Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547063
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (0.1-0.3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ENOVIN, EVN, NBN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547063 349.00
Check your price
  • Artemin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116738
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Neublastin; Enovin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116738 309.00
Check your price
  • Artemin Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123372
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
Neublastin; Enovin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123372 349.00
Check your price
  • Artemin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14142
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Neublastin; Enovin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14142 309.00
Check your price
  • Artemis Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131797
  • Size 100µg
Applications
ELISA (1:10,000-1:20,000), Immunohistochemistry (Paraffin) (1:1000), Western Blot (1:500-1:2000)
Alternate names:
SNM1-like protein; SNM1 homolog C; Protein A-SCID; DNA cross-link repair 1C protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131797 499.00
Check your price
  • Artemis Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA123464
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
SNM1-like protein; SNM1 homolog C; Protein A-SCID; DNA cross-link repair 1C protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123464 349.00
Check your price
  • Artemis Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14241
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
SNM1-like protein; SNM1 homolog C; Protein A-SCID; DNA cross-link repair 1C protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14241 349.00
Check your price
  • ARTS Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA130832
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Peanut-like protein 2; Cerebral protein 7; Cell division control-related protein 2; CE5B3 beta; Brain protein H5; Bradeion beta; Apoptosis-related protein in the TGF-beta signaling pathway
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130832 449.00
Check your price
  • ARX Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547896
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Aristaless-related homeobox
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547896 349.00
Check your price
  • Arylsulfatase A Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547330
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cerebroside-sulfatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547330 349.00
Check your price
  • Arylsulfatase B Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524327
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
N-acetylgalactosamine-4-sulfatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524327 275.00
Check your price
  • Arylsulfatase F Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA539030
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Arylsulfatase F, ASF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539030 309.00
Check your price
  • Arylsulfatase G Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547640
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ASG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547640 349.00
Check your price
  • Arylsulfatase G Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539031
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
ASG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539031 309.00
Check your price
  • AS160 Polyclonal Antibody for Western Blot, IHC (P), IHC (PFA) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519532
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:135), Immunohistochemistry (PFA fixed) (1:135), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Akt substrate of 160 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519532 349.00
Check your price
  • ASAH2 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547509
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Non-lysosomal ceramidase; N-acylsphingosine amidohydrolase 2; LCDase; BCDase; Acylsphingosine deacylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547509 375.00
Check your price
  • ASAH2 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA548033
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
N-acylsphingosine amidohydrolase 2; Acylsphingosine deacylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548033 309.00
Check your price
  • ASAHL Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524224
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
N-acylsphingosine amidohydrolase-like; Acid ceramidase-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524224 275.00
Check your price