Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  197  198  199  200  201  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  230  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1605 
6331 - 6360 of 48138 products displayed
  • NDFIP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534421
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/mL)
Alternate names:
NEDD4 WW domain-binding protein 5; Putative NFKB and MAPK-activating protein; Putative NF-kappa-B-activating protein 164; Putative MAPK-activating protein PM13; Breast cancer-associated protein SGA-1M
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534421 349.00
Check your price
  • NDP Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520722
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
X-linked exudative vitreoretinopathy 2 protein; Norrie disease protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520722 349.00
Check your price
  • NDRG1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523230
  • Size 100 ug
Applications
Western Blot (4.0-6 µg/ml)
Alternate names:
Rit42; Reducing agents and tunicamycin-responsive protein; Nickel-specific induction protein Cap43; N-myc downstream-regulated gene 1 protein; Differentiation-related gene 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523230 349.00
Check your price
  • NDRG1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518109
  • Size 100 ug
Applications
Immunohistochemistry (2-5µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Rit42; Reducing agents and tunicamycin-responsive protein; Nickel-specific induction protein Cap43; N-myc downstream-regulated gene 1 protein; Differentiation-related gene 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518109 349.00
Check your price
  • NDUFAF2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520831
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
NDUFA12-like protein; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2; Myc-induced mitochondrial protein; B17.2-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520831 349.00
Check your price
  • NDUFB9 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520922
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase B22 subunit; Complex I-B22; LYR motif-containing protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520922 349.00
Check your price
  • NDUFS1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519341
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Complex I-75kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519341 349.00
Check your price
  • NDUFS2 Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519342
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 49 kDa subunit; Complex I-49kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519342 349.00
Check your price
  • NDUFS3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519118
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 30 kDa subunit; Complex I-30kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519118 349.00
Check your price
  • NDUFS6 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519238
  • Size 100 ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 13 kDa-A subunit; Complex I-13kD-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519238 349.00
Check your price
  • NDUFS7 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519343
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
PSST subunit; NADH-ubiquinone oxidoreductase 20 kDa subunit; Complex I-20kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519343 349.00
Check your price
  • NDUFS8 Antibody
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519344
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
NADH-ubiquinone oxidoreductase 23 kDa subunit; Complex I-23kD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519344 349.00
Check your price
  • NEDD1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518626
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Neural precursor cell expressed developmentally down-regulated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518626 349.00
Check your price
  • NEDD4 Antibody
  • Host Rabbit
  • Target Species Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523492
  • Size 100 ug
Applications
Western Blot (4-6 µg/ml)
Alternate names:
Neural precursor cell expressed developmentally down-regulated protein 4; Cell proliferation-inducing gene 53 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523492 349.00
Check your price
  • NEDD4 Antibody
  • Host Rabbit
  • Target Species Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523462
  • Size 100 ug
Applications
Western Blot (4-6 µg/ml)
Alternate names:
Neural precursor cell expressed developmentally down-regulated protein 4; Cell proliferation-inducing gene 53 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523462 349.00
Check your price
  • NEDD8 Antibody (Monoclonal, 1A7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517133
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Ubiquitin-like protein Nedd8; Neural precursor cell expressed developmentally down-regulated protein 8; Neddylin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517133 309.00
Check your price
  • NEFH Antibody (Monoclonal, 8H10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517134
  • Size 100 ug
Applications
Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517134 309.00
Check your price
  • NEFH Antibody (Monoclonal, NF-05)
  • Host Mouse
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA110441
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (1 ug/ml)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110441 225.00
Check your price
  • NEFL Antibody (Monoclonal, 1H3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517135
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Neurofilament triplet L protein; 68 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517135 309.00
Check your price
  • NEFM Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519200
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Neurofilament triplet M protein; Neurofilament 3; 160 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519200 349.00
Check your price
  • NEIL1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518134
  • Size 100 ug
Applications
Immunohistochemistry (5-10µg/ml)
Alternate names:
Nei-like protein 1; Nei homolog 1; FPG1; Endonuclease VIII-like 1; DNA-(apurinic or apyrimidinic site) lyase Neil1; DNA glycosylase/AP lyase Neil1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518134 349.00
Check your price
  • NEK7 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519410
  • Size 100 ug
Applications
Western Blot (0.05-0.2 ug/ml)
Alternate names:
Never in mitosis A-related kinase 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519410 349.00
Check your price
  • NELF Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534508
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Nasal embryonic luteinizing hormone-releasing hormone factor; Juxtasynaptic attractor of caldendrin on dendritic boutons protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534508 349.00
Check your price
  • NENF Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520577
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Secreted protein of unknown function; Neuron-derived neurotrophic factor; Spinal cord injury-related protein 10; SCIRP10-related protein; Protein GIG47; Cell immortalization-related protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520577 349.00
Check your price
  • Nestin Antibody (Monoclonal, 1A9G12)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517136
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517136 309.00
Check your price
  • Nestin Antibody
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523386
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (3-5 µg/ml)
Alternate names:
NES, FLJ21841, Nbla00170
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523386 349.00
Check your price
  • NET1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518526
  • Size 100 ug
Applications
Immunohistochemistry (2.5µg/ml)
Alternate names:
Rho guanine nucleotide exchange factor 8; Proto-oncogene p65 Net1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518526 349.00
Check your price
  • NET1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518217
  • Size 100 ug
Applications
Immunohistochemistry (2.5µg/ml)
Alternate names:
Rho guanine nucleotide exchange factor 8; Proto-oncogene p65 Net1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518217 349.00
Check your price
  • Neuritin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520368
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (5-10 ug/ml)
Alternate names:
Candidate plasticity gene 15 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520368 349.00
Check your price
  • NeuroD1 Antibody (Monoclonal, 6E9G12)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517137
  • Size 100 ug
Applications
Western Blot (1:500-1:2000)
Alternate names:
Beta-cell E-box transcriptional activator 2; Class A basic helix-loop-helix protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517137 309.00
Check your price