Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  202  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1605 
6481 - 6510 of 48135 products displayed
  • NLRX1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521018
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Nucleotide-binding oligomerization domain protein 9; Nucleotide-binding oligomerization domain protein 5; Nucleotide-binding oligomerization domain protein 26; Caterpiller protein 11.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521018 349.00
Check your price
  • NMDAR1 Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518551
  • Size 100 ug
Applications
Western Blot (2-4 ug/ml)
Alternate names:
N-methyl-D-aspartate receptor subunit NR1; Glutamate [NMDA] receptor subunit zeta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518551 349.00
Check your price
  • NMDAR2B Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518536
  • Size 100 ug
Applications
Western Blot (2-4 ug/ml)
Alternate names:
N-methyl D-aspartate receptor subtype 2B; Glutamate [NMDA] receptor subunit epsilon-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518536 349.00
Check your price
  • NMDAR3A/NMDAR3B Antibody (Monoclonal, K35/40.1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA110881
  • Size 100 ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (1-10µg/mL)
Alternate names:
NMDA receptor 4; N-methyl-D-aspartate receptor subunit NR3B; NMDAR-L; N-methyl-D-aspartate receptor subtype 3A; NMDAR-L1; N-methyl-D-aspartate receptor; Glutamate receptor chi-1; N-methyl-D-aspartate receptor subtype 3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110881 375.00
Check your price
  • NMDAR3B Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519347
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
N-methyl-D-aspartate receptor subtype 3B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519347 349.00
Check your price
  • NO66 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518655
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Ribosomal oxygenase NO66; Nucleolar protein 66; Myc-associated protein with JmjC domain; Histone lysine demethylase NO66; 60S ribosomal protein L8 histidine hydroxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518655 349.00
Check your price
  • NOD1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518027
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Caspase recruitment domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518027 349.00
Check your price
  • NOD1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521015
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
Caspase recruitment domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521015 349.00
Check your price
  • NOD2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518572
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Inflammatory bowel disease protein 1; Caspase recruitment domain-containing protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518572 349.00
Check your price
  • NOD2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519991
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
Inflammatory bowel disease protein 1; Caspase recruitment domain-containing protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519991 349.00
Check your price
  • NOD2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519992
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (2-4 ug/ml)
Alternate names:
Inflammatory bowel disease protein 1; Caspase recruitment domain-containing protein 15
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519992 349.00
Check your price
  • NOD3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521016
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Nucleotide-binding oligomerization domain protein 3; Caterpiller protein 16.2; CARD15-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521016 349.00
Check your price
  • NOD4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521017
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Nucleotide-binding oligomerization domain protein 4; Nucleotide-binding oligomerization domain protein 27; Caterpiller protein 16.1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521017 349.00
Check your price
  • Noggin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522795
  • Size 100 ug
Applications
Western Blot (1:1000-1:2000)
Alternate names:
SYM1, SYNS1, symphalangism 1 (proximal)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522795 349.00
Check your price
  • Noggin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522829
  • Size 100 ug
Applications
Western Blot (1:1000-1:2000)
Alternate names:
SYM1, SYNS1, symphalangism 1 (proximal)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522829 349.00
Check your price
  • Nogo Receptor Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518380
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Nogo-66 receptor; Nogo receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518380 349.00
Check your price
  • Nogo-A Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520367
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Reticulon-5; RTN-x; Neuroendocrine-specific protein C homolog; Neuroendocrine-specific protein; Neurite outgrowth inhibitor; Foocen; Glut4 vesicle 20 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520367 349.00
Check your price
  • NONO Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518514
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
p54(nrb); NMT55; DNA-binding p52/p100 complex, 52 kDa subunit; 55 kDa nuclear protein; 54 kDa nuclear RNA- and DNA-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518514 349.00
Check your price
  • NOS1AP Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518084
  • Size 100 ug
Applications
Immunohistochemistry (5µg/ml)
Alternate names:
Nitric oxide synthase 1 adaptor protein; C-terminal PDZ ligand of neuronal nitric oxide synthase protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518084 349.00
Check your price
  • NOTCH1 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523181
  • Size 100 ug
Applications
Western Blot (3-5 µg/ml)
Alternate names:
Translocation-associated notch protein TAN-1; p300; mT14; Motch A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523181 349.00
Check your price
  • NOTCH1 Antibody, FITC (Monoclonal, mN1A)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate FITC
  • Catalog # MA516862
  • Size 100 ug
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Translocation-associated notch protein TAN-1; p300; mT14; Motch A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516862 349.00
Check your price
  • NOTCH2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523455
  • Size 100 ug
Applications
Western Blot (2-4 µg/ml)
Alternate names:
AGS2, HJCYS, hN2, Notch homolog 2, neurogenic locus notch homolog protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523455 349.00
Check your price
  • NOTCH2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522796
  • Size 100 ug
Applications
ELISA (1:100-1:2000), Immunohistochemistry (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
AGS2, HJCYS, hN2, Notch homolog 2, neurogenic locus notch homolog protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522796 349.00
Check your price
  • NOTCH3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522797
  • Size 100 ug
Applications
ELISA (1:100-1:2000), Immunohistochemistry (Frozen) (1:10-1:500), Western Blot (1:100-1:2000)
Alternate names:
CADASIL, CASIL, Notch homolog 3, neurogenic locus notch homolog protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522797 349.00
Check your price
  • NOTCH4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522798
  • Size 100 ug
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Immunohistochemistry (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
DAQB-143M3.4, INT3, NOTCH3, Notch homolog 4, neurogenic locus notch homolog protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522798 349.00
Check your price
  • NOTUM Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520788
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
hNOTUM
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520788 349.00
Check your price
  • NOVA1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518895
  • Size 100 ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
Ventral neuron-specific protein 1; Paraneoplastic Ri antigen; Onconeural ventral antigen 1; Neuro-oncological ventral antigen 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518895 349.00
Check your price
  • NOXA Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519068
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Protein Noxa; Immediate-early-response protein APR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519068 349.00
Check your price
  • NOXA Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519977
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (10 ug/ml), Immunohistochemistry (1 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Protein Noxa; Immediate-early-response protein APR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519977 349.00
Check your price
  • NOXO1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518256
  • Size 100 ug
Applications
Immunohistochemistry (10-20µg/ml)
Alternate names:
SH3 and PX domain-containing protein 5; Nox-organizing protein 1; Nox organizer 1; NADPH oxidase regulatory protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518256 349.00
Check your price