Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  110  120  130  140  150  160  170  180  190  203  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1622 
6511 - 6540 of 48655 products displayed
  • OGFOD2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521102
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521102 349.00
Check your price
  • OGG1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518747
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Ogg1; 8-oxoguanine DNA glycosylase; Mmh;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518747 349.00
Check your price
  • OGT Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518658
  • Size 100 ug
Applications
Western Blot (0.05-0.2 ug/ml)
Alternate names:
O-linked N-acetylglucosamine transferase 110 kDa subunit; O-GlcNAc transferase subunit p110
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518658 349.00
Check your price
  • OLIG2 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523456
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (1:200-1:400)
Alternate names:
Protein kinase C-binding protein RACK17; Protein kinase C-binding protein 2; Class E basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523456 349.00
Check your price
  • ORAI1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519045
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Transmembrane protein 142A; Protein orai-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519045 349.00
Check your price
  • ORAI2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520369
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1-4 µg/ml)
Alternate names:
Transmembrane protein 142B; CAP-binding protein complex-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520369 349.00
Check your price
  • ORAI3 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515779
  • Size 100 ug
Applications
Western Blot (2 µg/ml)
Alternate names:
Transmembrane protein 142C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515779 349.00
Check your price
  • ORAI3 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515778
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/ml), Immunofluorescence (5 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Transmembrane protein 142C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515778 349.00
Check your price
  • ORAI3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520375
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (2-4 ug/ml)
Alternate names:
Transmembrane protein 142C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520375 349.00
Check your price
  • ORAI3 Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520370
  • Size 100 ug
Applications
Immunocytochemistry (10 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
Transmembrane protein 142C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520370 349.00
Check your price
  • ORC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523390
  • Size 100 ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Replication control protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523390 349.00
Check your price
  • ORC4 Polyclonal Antibody for Western Blot, ChIP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA517950
  • Size 100 ug
Applications
ChIP assay (Assay Dependent), Western Blot (0.5-2 ug/ml)
Alternate names:
ORC4L; origin recognition complex, subunit 4-like (yeast); Orc4p; HSORC4; HsORC4; origin recognition complex, subunit 4 (yeast homolog)-like; Origin recognition complex, subunit 4, S. cerevisiae, homolog-like; ORC4; ORC4P; origin recognition complex subunit 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517950 349.00
Check your price
  • ORC6 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517948
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
ORC6L; origin recognition complex, subunit 6 homolog-like (yeast); ORC6; origin recognition complex subunit 6; origin recognition complex, subunit 6-like (yeast); origin recognition complex, subunit 6 (yeast homolog)-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517948 349.00
Check your price
  • ORMDL1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521146
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Adoplin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521146 349.00
Check your price
  • ORP1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518219
  • Size 100 ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
OSBPL1A; ORP1; OSBPL1B; FLJ10217; oxysterol binding protein-like 1A; OSBP-related protein 1; oxysterol-binding protein-like 1B; oxysterol-binding protein-related protein 1; ORP-1; oxysterol-binding protein-like 1A;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518219 349.00
Check your price
  • ORP2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518220
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
OSBPL2; ORP2; ORP-2; MGC4307; MGC8342; FLJ20223; KIAA0772; oxysterol binding protein-like 2; OSBP-related protein 2; oxysterol-binding protein-related protein 2; oxysterol-binding protein-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518220 349.00
Check your price
  • ORP2 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132067
  • Size 100 ug
Applications
ELISA (Assay-dependent), Western Blot (1-2 µg/ml)
Alternate names:
FLJ20223, KIAA0772, MGC4307, MGC8342, ORP-2, ORP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132067 349.00
Check your price
  • ORP5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518221
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Oxysterol-binding protein homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518221 349.00
Check your price
  • ORP9 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518222
  • Size 100 ug
Applications
Immunohistochemistry (2-4µg/ml)
Alternate names:
OSBPL9; ORP9; FLJ12492; FLJ14629; FLJ14801; FLJ32055; FLJ34384; MGC15035; oxysterol binding protein-like 9; OSBP-related protein 9; oxysterol-binding protein-related protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518222 349.00
Check your price
  • OSBP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518364
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
OSBP; oxysterol binding protein ; HGNC:8503; OSBP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518364 349.00
Check your price
  • OSBP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518218
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
OSBP; OSBP1; oxysterol binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518218 349.00
Check your price
  • OSBPL10 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518437
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
OSBPL10; oxysterol binding protein-like 10; HGNC:16395; FLJ20363; ORP10; OSBP9; OSBP-related protein 10; oxysterol-binding protein-like protein 10; oxysterol-binding protein-related protein 10; OTTHUMP00000161257
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518437 349.00
Check your price
  • OSBPL11 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518223
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
OSBPL11; ORP11; ORP-11; OSBP12; FLJ13012; FLJ13164; TCCCIA00292; oxysterol binding protein-like 11; OSBP-related protein 11; oxysterol-binding protein-related protein 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518223 349.00
Check your price
  • OSBPL11 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human
  • Conjugate Unconjugated
  • Catalog # PA132069
  • Size 100 ug
Applications
ELISA (Assay-dependent), Western Blot (1-2 µg/ml)
Alternate names:
FLJ13012, FLJ13164, ORP-11, ORP11, OSBP12, TCCCIA00292
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132069 399.00
Check your price
  • Osteocalcin Monoclonal Antibody for Western Blot, IHC, ELISA and RIA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # MA143028
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Gamma-carboxyglutamic acid-containing protein; Bone Gla protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA143028 199.00
Check your price
  • Osteopontin Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517180
  • Size 100 ug
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Uropontin; Urinary stone protein; Secreted phosphoprotein 1; Nephropontin; Bone sialoprotein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517180 309.00
Check your price
  • Osteopontin Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19076
  • Size 100 ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Uropontin; Urinary stone protein; Secreted phosphoprotein 1; Nephropontin; Bone sialoprotein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19076 349.00
Check your price
  • Osteopontin Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534579
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Secreted phosphoprotein 1; Minopontin; Early T-lymphocyte activation 1 protein; Calcium oxalate crystal growth inhibitor protein; Bone sialoprotein 1; 2AR; Uropontin; Urinary stone protein; Nephropontin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534579 349.00
Check your price
  • OTUB1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518960
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Ubiquitin-specific-processing protease OTUB1; Otubain-1; OTU domain-containing ubiquitin aldehyde-binding protein 1; Deubiquitinating enzyme OTUB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518960 349.00
Check your price
  • OTUB2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518961
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Ubiquitin-specific-processing protease OTUB2; Otubain-2; OTU domain-containing ubiquitin aldehyde-binding protein 2; Deubiquitinating enzyme OTUB2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518961 349.00
Check your price