Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  120  130  140  150  160  170  180  190  200  204  205  206  207  208  209  210  211  212  213  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  240  250  260  270  280  290  300  310  400  500  600  700  800  900  1000  1100  1200  1630 
6541 - 6570 of 48882 products displayed
  • beta Catenin Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P), IP and GS
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1301
  • Size 100µg
Applications
Gel Shift (Assay Dependent), Immunocytochemistry (1:50-1:1000), Immunofluorescence (1:50-1:1000), Immunohistochemistry (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:1000)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1301 275.00
Check your price
  • beta Catenin Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1300
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:50-1:1000), Immunohistochemistry (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:1000)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1300 275.00
Check your price
  • beta Catenin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA12001
  • Size 100µg
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Immunoprecipitation (1 ug/ml), Western Blot (1-3 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA12001 275.00
Check your price
  • beta Catenin Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 138400
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
138400 308.00
Check your price
  • beta Catenin Polyclonal Antibody for IF, ICC, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 712700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
712700 305.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IF, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rodent, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519469
  • Size 100µg
Applications
ELISA (0.1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (0.25 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519469 349.00
Check your price
  • beta COP Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA1068
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
Beta-coat protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1068 349.00
Check your price
  • beta Dystroglycan Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519731
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Dystrophin-associated glycoprotein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519731 349.00
Check your price
  • Beta Endorphin Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701295
  • Size 100µg
Applications
Western Blot (2-3 ug/ml)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
701295 309.00
Check your price
  • Beta Endorphin Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710319
  • Size 100µg
Applications
Western Blot (2-3 ug/ml)
Alternate names:
Corticotropin-lipotropin
Catalog # Size Price (AUD) Your Price (AUD) Qty
710319 309.00
Check your price
  • beta Galactosidase Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA1152
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:50-1:100), Immunofluorescence (1:50-1:100), Western Blot (1:500)
Alternate names:
Beta-Gal, RP3-337O18.1, GLB2, GSL, NGBE, PPCA, PPGB
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1152 349.00
Check your price
  • beta Galactosidase Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539059
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Elastin receptor 1; Acid beta-galactosidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539059 309.00
Check your price
  • beta Synuclein Polyclonal Antibody for IHC
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA118261
  • Size 100µg
Applications
Immunohistochemistry (1 µg/ml)
Alternate names:
Phosphoneuroprotein 14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118261 349.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA516308
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:2000-1:5000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308 225.00
Check your price
  • beta Tubulin Monoclonal Antibody for Western Blot, ICC and IHC (P)
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA119164
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Tubulin beta-5 chain; Tubulin beta-4 chain; Tubulin 5 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119164 309.00
Check your price
  • beta Tubulin Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Cat, Human, Mouse, Non-human primate, Rat, Xenopus, Bovine, Dog, Chicken
  • Conjugate Unconjugated
  • Catalog # PA141331
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.1-1 µg/ml)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141331 349.00
Check your price
  • beta-2 Adaptin Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA11066
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Plasma membrane adaptor HA2/AP2 adaptin beta subunit; Clathrin assembly protein complex 2 beta large chain; Beta-adaptin; Beta-2-adaptin; Adaptor-related protein complex 2 subunit beta; Adaptor protein complex AP-2 subunit beta; AP105B; Golgi adaptor HA1/AP1 adaptin beta subunit; Clathrin assembly protein complex 1 beta large chain; Beta-adaptin 1; Beta-1-adaptin; Adaptor-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11066 349.00
Check your price
  • beta-2 Defensin Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547388
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
DEFB-2, DEFB102, DEFB2, DEFB4, HBD-2, SAP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547388 349.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 701250
  • Size 100µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
701250 309.00
Check your price
  • beta-2 Microglobulin Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710274
  • Size 100µg
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
710274 309.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody for Western Blot, ICC, IHC (P), Flow, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119141
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1 µg/ml), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin) (10µg/ml), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent), Western Blot (2-4 µg/ml)
Alternate names:
Orla-B2m, B2M, Beta 2 microglobulin precursor, Beta chain of mhc class 1 proteins, Hdcma22p
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119141 309.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, Alexa Fluor® 647 conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 647
  • Catalog # MA518119
  • Size 100µg
Applications
Flow Cytometry (4ulx100ul whole blood or 10^6 cells), Immunocytochemistry (2ug/ml), Immunofluorescence (2ug/ml)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518119 309.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119506
  • Size 100µg
Applications
Flow Cytometry (1:200)
Alternate names:
Orla-B2m, B2M, Beta 2 microglobulin precursor, Beta chain of mhc class 1 proteins, Hdcma22p
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119506 309.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA119579
  • Size 100µg
Applications
Flow Cytometry (1:200)
Alternate names:
Orla-B2m, B2M, Beta 2 microglobulin precursor, Beta chain of mhc class 1 proteins, Hdcma22p
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119579 309.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A15737
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15737 133.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA119641
  • Size 100µg
Applications
Flow Cytometry (1 µg/ml)
Alternate names:
Orla-B2m, B2M, Beta 2 microglobulin precursor, Beta chain of mhc class 1 proteins, Hdcma22p
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119641 399.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A15770
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B2-microglobulin, B2M, CDABP0092, HDCMA22P, beta chain MHC class I molecules, MHC Class 1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15770 184.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody, PE conjugate for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA119679
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Orla-B2m, B2M, Beta 2 microglobulin precursor, Beta chain of mhc class 1 proteins, Hdcma22p
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119679 349.00
Check your price
  • beta-2 Microglobulin Monoclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA119413
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1 µg/ml), Immunohistochemistry (Paraffin) (5 ug/ml), Western Blot (2-4 ug/ml)
Alternate names:
Orla-B2m, B2M, Beta 2 microglobulin precursor, Beta chain of mhc class 1 proteins, Hdcma22p
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119413 309.00
Check your price
  • beta-2 Microglobulin Polyclonal Antibody for Western Blot, ELISA, IDF and IE
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # R02021D
  • Size 100µg
Applications
ELISA (Assay dependent), Immunodiffusion (Assay dependent), Immunoelectrophoresis (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Orla-B2m, B2M, Beta 2 microglobulin precursor, Beta chain of mhc class 1 proteins, Hdcma22p
Catalog # Size Price (AUD) Your Price (AUD) Qty
R02021D 309.00
Check your price