Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  214  215  216  217  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  250  260  270  280  290  300  310  320  400  500  600  700  800  900  1000  1100  1200  1622 
6841 - 6870 of 48655 products displayed
  • PRR5L Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521129
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Protein observed with Rictor-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521129 349.00
Check your price
  • PRR7 Monoclonal Antibody for Western Blot and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA110448
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Synaptic proline-rich membrane protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110448 309.00
Check your price
  • PRRT2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534537
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (25 ug/ml), Western Blot (1 ug/mL)
Alternate names:
Dispanin subfamily B member 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534537 349.00
Check your price
  • PRRX1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518831
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Paired-related homeobox protein 1; Homeobox protein PHOX1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518831 349.00
Check your price
  • PRTFDC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA520593
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
HHGP; phosphoribosyltransferase domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520593 349.00
Check your price
  • PRUNE2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518725
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
BNIP2 motif-containing molecule at the C-terminal region 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518725 349.00
Check your price
  • PSAP Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517159
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-500), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Proactivator polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517159 309.00
Check your price
  • PSD95 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA519114
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Synapse-associated protein 90; Postsynaptic density protein 95
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519114 349.00
Check your price
  • PSEN1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520376
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Protein S182
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520376 349.00
Check your price
  • PSMA Monoclonal Antibody for ICC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110335
  • Size 100 ug
Applications
Flow Cytometry (6 ug/ml), Immunocytochemistry (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
Pteroylpoly-gamma-glutamate carboxypeptidase; Prostate-specific membrane antigen; N-acetylated-alpha-linked acidic dipeptidase I; Membrane glutamate carboxypeptidase; Glutamate carboxypeptidase II; Folylpoly-gamma-glutamate carboxypeptidase; Folate hydrolase 1; Cell growth-inhibiting gene 27 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110335 249.00
Check your price
  • PSMA Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110336
  • Size 100 ug
Applications
Flow Cytometry (1:200)
Alternate names:
Pteroylpoly-gamma-glutamate carboxypeptidase; Prostate-specific membrane antigen; N-acetylated-alpha-linked acidic dipeptidase I; Membrane glutamate carboxypeptidase; Glutamate carboxypeptidase II; Folylpoly-gamma-glutamate carboxypeptidase; Folate hydrolase 1; Cell growth-inhibiting gene 27 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110336 399.00
Check your price
  • PSMA4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519368
  • Size 100 ug
Applications
Western Blot (0.3-1.0 ug/ml)
Alternate names:
Proteasome subunit L; Proteasome component C9; Multicatalytic endopeptidase complex subunit C9; Macropain subunit C9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519368 349.00
Check your price
  • PSMB10 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519146
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5-10 ug/ml), Western Blot (0.3-1 ug/ml)
Alternate names:
Proteasome subunit beta-2i; Proteasome MECl-1; Multicatalytic endopeptidase complex subunit MECl-1; Macropain subunit MECl-1; Low molecular mass protein 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519146 349.00
Check your price
  • PSMB3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519373
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Proteasome theta chain; Proteasome component C10-II; Proteasome chain 13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519373 349.00
Check your price
  • PSMB4 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519374
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Proteasome chain 3; Proteasome beta chain; Multicatalytic endopeptidase complex beta chain; Macropain beta chain; 26 kDa prosomal protein; Low molecular mass protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519374 349.00
Check your price
  • PSMB7 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519375
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Proteasome subunit Z; Multicatalytic endopeptidase complex chain Z; Macropain chain Z
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519375 349.00
Check your price
  • PSMB8 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519145
  • Size 100 ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Really interesting new gene 10 protein; Proteasome subunit beta-5i; Proteasome component C13; Multicatalytic endopeptidase complex subunit C13; Macropain subunit C13; Low molecular mass protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519145 349.00
Check your price
  • PSMB9 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519384
  • Size 100 ug
Applications
Western Blot (0.3-1.0 ug/ml)
Alternate names:
Really interesting new gene 12 protein; Proteasome subunit beta-1i; Proteasome chain 7; Multicatalytic endopeptidase complex chain 7; Macropain chain 7; Low molecular mass protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519384 349.00
Check your price
  • PSMD2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518324
  • Size 100 ug
Applications
Western Blot (0.03-0.3 ug/ml)
Alternate names:
Tumor necrosis factor type 1 receptor-associated protein 2; Protein 55.11; 26S proteasome subunit p97; 26S proteasome regulatory subunit S2; 26S proteasome regulatory subunit RPN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518324 349.00
Check your price
  • PSME1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518707
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Proteasome activator 28 subunit alpha; Interferon gamma up-regulated I-5111 protein; Activator of multicatalytic protease subunit 1; 11S regulator complex subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518707 349.00
Check your price
  • PSME2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518708
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
Proteasome activator 28 subunit beta; Activator of multicatalytic protease subunit 2; 11S regulator complex subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518708 349.00
Check your price
  • PSMF1 Polyclonal Antibody for Western Blot and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518133
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
PSMF1; PI31; proteasome (prosome, macropain) inhibitor subunit 1 (PI31); RP4-545L17.1; proteasome inhibitor hP131 subunit; proteasome inhibitor subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518133 349.00
Check your price
  • PSMG1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519076
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Down syndrome critical region protein 2; Chromosome 21 leucine-rich protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519076 349.00
Check your price
  • PSPH Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519113
  • Size 100 ug
Applications
Western Blot (0.05-1 ug/ml)
Alternate names:
O-phosphoserine phosphohydrolase; L-3-phosphoserine phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519113 349.00
Check your price
  • PST1 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Yeast
  • Conjugate Unconjugated
  • Catalog # PA534414
  • Size 100 ug
Applications
ELISA (1 ug/mL), Western Blot (1 ug/mL)
Alternate names:
Protoplast secreted protein 1; Haze protective factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534414 349.00
Check your price
  • PTBP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518197
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Heterogeneous nuclear ribonucleoprotein I; 57 kDa RNA-binding protein PPTB-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518197 349.00
Check your price
  • PTCH Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518454
  • Size 100 ug
Applications
Immunohistochemistry (3-6µg/ml)
Alternate names:
PTCH1; PTCH; patched homolog (Drosophila); HGNC:9585; BCNS; HPE7; NBCCS; PTC; PTC1; patched; patched (Drosophila) homolog; patched homolog 1 (Drosophila); RP11-435O5.3; FLJ26746; FLJ42602; PTCH11; OTTHUMP00000021709; OTTHUMP00000021710; PTCH protein +12b; PTCH protein +4'; PTCH protein -10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518454 349.00
Check your price
  • PTCH Polyclonal Antibody for Western Blot, IF, ICC and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518544
  • Size 100 ug
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1-3 ug/ml)
Alternate names:
PTCH; patched homolog (Drosophila); RP11-435O5.3; BCNS; FLJ42602; HPE7; NBCCS; PTC; PTC1; PTCH1 ; PTCH protein +12b; PTCH protein +4'; PTCH protein -10; patched; patched (Drosophila) homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518544 349.00
Check your price
  • PTCH Polyclonal Antibody for IF
  • Host Goat
  • Target Species Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA518776
  • Size 100 ug
Applications
Immunofluorescence (1-3µg/ml.)
Alternate names:
ptc1; patched 1 ; ptc-1; etID309849.2; patched1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518776 349.00
Check your price
  • PTCHD1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519402
  • Size 100 ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
PTCHD1; patched domain containing 1; FLJ30296; MGC149798
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519402 349.00
Check your price