Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  218  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  250  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1630 
6961 - 6990 of 48877 products displayed
  • CBP Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1847
  • Size 100µg
Applications
ChIP assay (1-3 ul), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:1000-1:5000), Western Blot (1-2 µg/ml)
Alternate names:
CREB Binding Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1847 375.00
Check your price
  • CBR3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539079
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
NADPH-dependent carbonyl reductase 3; Short chain dehydrogenase/reductase family 21C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539079 309.00
Check your price
  • CBX2 Monoclonal Antibody for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 730039
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
CDCA6, M33, SRXY5, Pc class homolog, cell division cycle associated 6, chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, modifier 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
730039 309.00
Check your price
  • CBX4 Monoclonal Antibody for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 730111
  • Size 100µg
Applications
Immunoprecipitation (0.5 mg/ml)
Alternate names:
Polycomb 2 homolog; Chromobox protein homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
730111 309.00
Check your price
  • CBX4 Monoclonal Antibody, Biotin conjugate for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 730099
  • Size 100µg
Applications
Immunoprecipitation (0.5 mg/ml)
Alternate names:
Polycomb 2 homolog; Chromobox protein homolog 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
730099 309.00
Check your price
  • CBX8 Monoclonal Antibody, Biotin conjugate for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # 730098
  • Size 100µg
Applications
Immunoprecipitation (0.5 mg/ml)
Alternate names:
Rectachrome 1; Polycomb 3 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
730098 309.00
Check your price
  • CBY1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521555
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
PKD2 interactor, Golgi and endoplasmic reticulum-associated 1; PIGEA-14; Cytosolic leucine-rich protein; ARPP-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521555 309.00
Check your price
  • CCAAT Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA537924
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
CEBP zeta, C/EBP zeta, Cebpz, AI848081, CBF2, Cbf, Cebpa-rs1, CCAAT-binding factor, CCAAT-box-binding transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537924 309.00
Check your price
  • CCDC102A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539112
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539112 309.00
Check your price
  • CCDC3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537980
  • Size 100µg
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
Fat/vessel-derived secretory protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537980 309.00
Check your price
  • CCDC83 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523184
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
HSD9, coiled-coil domain-containing protein 83
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523184 309.00
Check your price
  • CCL1 Monoclonal Antibody for Western Blot and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523873
  • Size 100µg
Applications
Neutralization (2-10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
T-cell activation protein 3; Small-inducible cytokine A1; SIS-epsilon; P500
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523873 275.00
Check your price
  • CCL1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523821
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (0.1-0.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
T lymphocyte-secreted protein I-309; Small-inducible cytokine A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523821 275.00
Check your price
  • CCL1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547952
  • Size 100µg
Applications
Neutralization (0.3-1.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell activation protein 3; Small-inducible cytokine A1; SIS-epsilon; P500
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547952 349.00
Check your price
  • CCL16 Polyclonal Antibody for Western Blot, IHC and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547948
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Neutralization (4-16 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A16; NCC-4; Monotactin-1; Lymphocyte and monocyte chemoattractant; Liver-expressed chemokine; LCC-1; IL-10-inducible chemokine; Chemokine LEC; Chemokine CC-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547948 349.00
Check your price
  • CCL2 (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP75663
  • Size 100µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Monocyte Chemotactic Protein-1, MCP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75663 550.00
Check your price
  • CCL21 Monoclonal Antibody for Western Blot, IHC, Flow and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523837
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
6Ckine, CKb9, ECL, SCYA21, SLC, TCA4, C-C motif chemokine 21, Efficient Chemoattractant for Lymphocytes, beta chemokine exodus-2, beta-chemokine exodus-2, exodus-2, secondary lymphoid tissue chemokine, secondary lymphoid-tissue chemokine, small inducible cytokine subfamily A (Cys-Cys), member 21, sma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523837 275.00
Check your price
  • CCL21 Polyclonal Antibody for Western Blot, IHC (F), Flow and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547016
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (0.5-2.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
6Ckine, CKb9, ECL, SCYA21, SLC, TCA4, C-C motif chemokine 21, Efficient Chemoattractant for Lymphocytes, beta chemokine exodus-2, beta-chemokine exodus-2, exodus-2, secondary lymphoid tissue chemokine, secondary lymphoid-tissue chemokine, small inducible cytokine subfamily A (Cys-Cys), member 21, sma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547016 375.00
Check your price
  • CCL21 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA129031
  • Size 100µg
Applications
Western Blot (1:5000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129031 349.00
Check your price
  • CCL23 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546958
  • Size 100µg
Applications
Neutralization (0.15-0.75 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A23; Myeloid progenitor inhibitory factor 1; Macrophage inflammatory protein 3; CK-beta-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546958 375.00
Check your price
  • CCL23 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547528
  • Size 100µg
Applications
Neutralization (0.5-2.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A23; Myeloid progenitor inhibitory factor 1; Macrophage inflammatory protein 3; CK-beta-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547528 349.00
Check your price
  • CCL27 Monoclonal Antibody for Western Blot, IHC and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523862
  • Size 100µg
Applications
Immunohistochemistry (8-25 µg/ml), Neutralization (0.5-3.0 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A27; Skinkine; IL-11 R-alpha-locus chemokine; ESkine; Cutaneous T-cell-attracting chemokine; CC chemokine ILC
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523862 275.00
Check your price
  • CCL27 Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546942
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A27; Skinkine; IL-11 R-alpha-locus chemokine; ESkine; Cutaneous T-cell-attracting chemokine; CC chemokine ILC
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546942 375.00
Check your price
  • CCL27 Polyclonal Antibody for Western Blot, IHC (F) and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547908
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Neutralization (10-20 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
ALP, CTACK, CTAK, ESKINE, ILC, PESKY, SCYA27
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547908 349.00
Check your price
  • CCL28 Monoclonal Antibody for Western Blot and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523865
  • Size 100µg
Applications
Neutralization (2.5-5.0 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A28
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523865 275.00
Check your price
  • CCL28 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547722
  • Size 100µg
Applications
Neutralization (5-25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547722 349.00
Check your price
  • CCL3 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701097
  • Size 100µg
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701097 309.00
Check your price
  • CCL3 Oligoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710127
  • Size 100µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710127 309.00
Check your price
  • CCL3 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524364
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524364 275.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547000
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (2-8 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547000 375.00
Check your price