Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  219  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  250  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1605 
6991 - 7020 of 48136 products displayed
  • RNF2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518130
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
RING finger protein BAP-1; RING finger protein 2; RING finger protein 1B; Protein DinG; Huntingtin-interacting protein 2-interacting protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518130 349.00
Check your price
  • RNF20 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534552
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (25 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
RING finger protein 20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534552 349.00
Check your price
  • RNF213 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518650
  • Size 100 ug
Applications
Immunohistochemistry (2.5µg/ml)
Alternate names:
RING finger protein 213; Mysterin; ALK lymphoma oligomerization partner on chromosome 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518650 349.00
Check your price
  • RNF216 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520079
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Zinc finger protein inhibiting NF-kappa-B; Ubiquitin-conjugating enzyme 7-interacting protein 1; Triad domain-containing protein 3; RING finger protein 216; UbcM4-interacting protein 83
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520079 349.00
Check your price
  • RNF3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518131
  • Size 100 ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
RING finger protein 3A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518131 349.00
Check your price
  • RNF31 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518085
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Zinc in-between-RING-finger ubiquitin-associated domain protein; RING finger protein 31; HOIL-1-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518085 349.00
Check your price
  • RNF39 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518128
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Protein HZFw
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518128 349.00
Check your price
  • RNF7 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519233
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Sensitive to apoptosis gene protein; Regulator of cullins 2; RING finger protein 7; CKII beta-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519233 349.00
Check your price
  • RNF8 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518132
  • Size 100 ug
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
RING finger protein 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518132 349.00
Check your price
  • RNF8 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520961
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
RING finger protein 8; LaXp180; ActA-interacting protein 37
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520961 349.00
Check your price
  • ROBO1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518460
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
H-Robo-1; Deleted in U twenty twenty
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518460 349.00
Check your price
  • ROBO3 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518714
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
Roundabout-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518714 349.00
Check your price
  • ROCK1 Antibody (Monoclonal, 1H4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517165
  • Size 100 ug
Applications
Western Blot (1:500-1:2000)
Alternate names:
p160 ROCK-1; Rho-associated, coiled-coil-containing protein kinase I; Rho-associated, coiled-coil-containing protein kinase 1; Renal carcinoma antigen NY-REN-35
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517165 309.00
Check your price
  • ROCK1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521130
  • Size 100 ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 ug/ml)
Alternate names:
p160 ROCK-1; Rho-associated, coiled-coil-containing protein kinase I; Rho-associated, coiled-coil-containing protein kinase 1; Renal carcinoma antigen NY-REN-35; p150 RhoA-binding kinase ROK beta; Liver regeneration-related protein LRRG199
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521130 349.00
Check your price
  • RON Antibody (Monoclonal, Zt/f2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518078
  • Size 100 ug
Applications
ELISA (Assay-dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1 ug/ml), Immunoprecipitation (Assay-dependent)
Alternate names:
p185-Ron; Protein-tyrosine kinase 8; CDw136
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518078 425.00
Check your price
  • RORA Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Horse, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523268
  • Size 100 ug
Applications
Western Blot (0.01-0.25 µg/ml)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523268 349.00
Check your price
  • RORA Antibody
  • Host Rabbit
  • Target Species Bovine, Chicken, Horse, Human, Marsupial, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523267
  • Size 100 ug
Applications
Western Blot (0.025-0.05 µg/ml)
Alternate names:
Retinoid-related orphan receptor-alpha; RAR-related orphan receptor A; Nuclear receptor subfamily 1 group F member 1; Nuclear receptor RZR-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523267 349.00
Check your price
  • RORC Antibody (Monoclonal, 4G419)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA516227
  • Size 100 ug
Applications
Flow Cytometry (0.25-0.5 µg/10^6 cells), Western Blot (5 µg/ml)
Alternate names:
Thymus orphan receptor; Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516227 349.00
Check your price
  • RORC Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523148
  • Size 100 ug
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Thymus orphan receptor; Retinoid-related orphan receptor-gamma; RAR-related orphan receptor C; Nuclear receptor subfamily 1 group F member 3; Nuclear receptor RZR-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523148 349.00
Check your price
  • RPA14 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521277
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Replication factor A protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521277 349.00
Check your price
  • RPA70 Antibody (Monoclonal, 4C4)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA517166
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Single-stranded DNA-binding protein; Replication factor A protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517166 309.00
Check your price
  • RPAIN Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520326
  • Size 100 ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
HRIP, RIP; RAP interaction protein; RPA-interacting protein; nuclear transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520326 349.00
Check your price
  • RPAIN Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520327
  • Size 100 ug
Applications
Western Blot (0.5-1 ug/ml)
Alternate names:
HRIP, RIP; RAP interaction protein; RPA-interacting protein; nuclear transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520327 349.00
Check your price
  • RPE65 Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518890
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Retinol isomerase; Retinal pigment epithelium-specific 65 kDa protein; All-trans-retinyl-palmitate hydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518890 349.00
Check your price
  • RPGRIP1L Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518785
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
RPGR-interacting protein 1-like protein; Nephrocystin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518785 349.00
Check your price
  • RPL17 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518799
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
PD-1; 60S ribosomal protein L23
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518799 349.00
Check your price
  • RPL18A Antibody (Monoclonal, 6G6G10)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517167
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517167 309.00
Check your price
  • RPL19 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518529
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
RPL19; ribosomal protein L19 ; HGNC:10312; DKFZp779D216; FLJ27452; MGC71997 ; 60S ribosomal protein L19; ribosomal protein L19, cytosolic, N-terminus truncated
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518529 349.00
Check your price
  • RPL8 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518743
  • Size 100 ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.01-0.03 ug/ml)
Alternate names:
ribosomal protein L8; 60S ribosomal protein L8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518743 349.00
Check your price
  • RPS19 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518452
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
RPS19; ribosomal protein S19; HGNC:10402; DBA; 40S ribosomal protein S19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518452 349.00
Check your price