Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  220  221  222  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1630 
7021 - 7050 of 48877 products displayed
  • CCR8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112539
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112539 325.00
Check your price
  • CCR8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519777
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519777 349.00
Check your price
  • CCR8 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121621
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (1-3 µl for 10^6 cells in 100 µl), Immunocytochemistry (1:800), Immunofluorescence (1:800), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
TER1; GPR-CY6; Chemokine receptor-like 1; CMKBRL2; CC chemokine receptor CHEMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121621 425.00
Check your price
  • CCR9 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA121618
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (1:200), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
Chemokine C-C receptor 10; GPR-9-6; G-protein coupled receptor 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121618 425.00
Check your price
  • CCRD6 Polyclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121614
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (1-3 µl for 10^6 cells in 100 µl), Immunohistochemistry (Paraffin) (1:250), Western Blot (1:1000)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; Chemokine receptor CCR-9; Chemokine receptor CCR-10; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121614 399.00
Check your price
  • CCRD6 Polyclonal Antibody for IF, ICC, IHC (P), Flow and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA121612
  • Size 100µg
Applications
ELISA (1:100,000), Flow Cytometry (1-5 µg/ml), Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (Paraffin) (1:250)
Alternate names:
Chemokine-binding protein D6; Chemokine-binding protein 2; C-C chemokine receptor D6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121612 399.00
Check your price
  • CCRL2 Monoclonal Antibody for IF, ICC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524209
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
Lipopolysaccharide-inducible C-C chemokine receptor; G-protein coupled beta chemokine receptor; Chemokine receptor CCR11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524209 275.00
Check your price
  • CCRN4L Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124489
  • Size 100µg
Applications
Immunohistochemistry (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Circadian deadenylase NOC; Carbon catabolite repression 4-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124489 375.00
Check your price
  • CD10 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119379
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Common acute lymphocytic leukemia antigen; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119379 199.00
Check your price
  • CD10 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119086
  • Size 100µg
Applications
Flow Cytometry (1 ug/ml), Immunoprecipitation (Assay Dependent)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Common acute lymphocytic leukemia antigen; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119086 199.00
Check your price
  • CD10 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112031
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Common acute lymphocytic leukemia antigen; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112031 225.00
Check your price
  • CD10 Polyclonal Antibody for Western Blot, IHC (F), IP, ELISA and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547075
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Neutralization (1.3 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Skin fibroblast elastase; Neutral endopeptidase 24.11; Enkephalinase; Atriopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547075 375.00
Check your price
  • CD100 Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524248
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Semaphorin-J; Semaphorin-C-like 2; M-Sema G
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524248 275.00
Check your price
  • CD100 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA134040
  • Size 100µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134040 309.00
Check your price
  • CD100 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547711
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Semaphorin-J; Semaphorin-C-like 2; M-Sema G
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547711 349.00
Check your price
  • CD100 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14202
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14202 309.00
Check your price
  • CD100 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14201
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
GR3; BB18; A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14201 399.00
Check your price
  • CD101 Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524021
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Glu-Trp-Ile EWI motif-containing protein 101
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524021 275.00
Check your price
  • CD103 Monoclonal Antibody, Pacific Blue™ conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate Pacific Blue™
  • Catalog # A15422
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15422 211.00
Check your price
  • CD103 Monoclonal Antibody, PE conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate PE
  • Catalog # HMCD10304
  • Size 100µg
Applications
Flow Cytometry (0.1-0.2 ug per 1x10^6 cells)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
HMCD10304 245.00
Check your price
  • CD103 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121434
  • Size 100µg
Applications
Flow Cytometry (2 µg for 5x10^5 cells), Immunohistochemistry (Frozen) (1:50-1:100), Immunoprecipitation (Assay dependent)
Alternate names:
Mucosal lymphocyte 1 antigen; Integrin alpha-IEL; HML-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121434 349.00
Check your price
  • CD103 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112189
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Mucosal lymphocyte 1 antigen; Integrin alpha-IEL; HML-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112189 249.00
Check your price
  • CD103 Monoclonal Antibody for IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA133553
  • Size 100µg
Applications
Flow Cytometry (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
Mucosal lymphocyte 1 antigen; Integrin alpha-IEL; HML-1 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133553 309.00
Check your price
  • CD103 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547236
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Integrin alpha M290
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547236 349.00
Check your price
  • CD104 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524024
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (8-25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
CD104 antigen, GP150, integrin beta-4, integrin beta-4 subunit, ITGB4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524024 275.00
Check your price
  • CD105 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523894
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunohistochemistry (Frozen) (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523894 275.00
Check your price
  • CD105 Monoclonal Antibody for Western Blot, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119231
  • Size 100µg
Applications
Flow Cytometry (2µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CD105, END, HHT1, ORW, ORW1, CD105 antigen, ENG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119231 199.00
Check your price
  • CD105 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA119408
  • Size 100µg
Applications
Flow Cytometry (10 µg/ml), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CD105, END, HHT1, ORW, ORW1, CD105 antigen, ENG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119408 349.00
Check your price
  • CD105 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Horse, Human, Non-human primate
  • Conjugate FITC
  • Catalog # MA180943
  • Size 100µg
Applications
Flow Cytometry (Neat)
Alternate names:
CD105, END, HHT1, ORW, ORW1, CD105 antigen, ENG
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180943 375.00
Check your price
  • CD105 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA546971
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.5-5 µg/ml)
Alternate names:
Cell surface MJ7/18 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546971 399.00
Check your price