Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  130  140  150  160  170  180  190  200  210  223  224  225  226  227  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  260  270  280  290  300  310  320  330  400  500  600  700  800  900  1000  1100  1200  1630 
7111 - 7140 of 48882 products displayed
  • CD146 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547470
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Melanoma-associated antigen MUC18; Melanoma cell adhesion molecule; Gicerin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547470 349.00
Check your price
  • CD146 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 346800
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (Assay Dependent)
Alternate names:
S-endo 1 endothelial-associated antigen; Melanoma-associated antigen MUC18; Melanoma-associated antigen A32; Melanoma cell adhesion molecule; Cell surface glycoprotein P1H12
Catalog # Size Price (AUD) Your Price (AUD) Qty
346800 309.00
Check your price
  • CD147 (soluble) Monoclonal Antibody for Western Blot, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119201
  • Size 100µg
Applications
Flow Cytometry (10µg/ml), Immunohistochemistry (Paraffin) (10µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1µg/ml)
Alternate names:
Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119201 199.00
Check your price
  • CD147 (soluble) Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119511
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119511 225.00
Check your price
  • CD147 (soluble) Monoclonal Antibody for IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA119410
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (10 ug/ml)
Alternate names:
Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119410 199.00
Check your price
  • CD147 (soluble) Monoclonal Antibody for Western Blot, Flow and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110103
  • Size 100µg
Applications
Flow Cytometry (4-10 µg/ml), Functional Assay (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110103 199.00
Check your price
  • CD147 Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # MA135092
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Membrane glycoprotein gp42; HT7 antigen; Basic immunoglobulin superfamily; Neurothelin; Blood-brain barrier HT7 antigen; 5A11 antigen; OX-47 antigen; Glycoprotein CE9; Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135092 309.00
Check your price
  • CD147 Polyclonal Antibody for Western Blot, IF, ICC, IHC, Flow and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 345600
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor cell-derived collagenase stimulatory factor; OK blood group antigen; Leukocyte activation antigen M6; Extracellular matrix metalloproteinase inducer; Collagenase stimulatory factor; 5F7
Catalog # Size Price (AUD) Your Price (AUD) Qty
345600 309.00
Check your price
  • CD148 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA547225
  • Size 100µg
Applications
Immunoprecipitation (2.5 µg/500 µg cell lysate), Western Blot (1 µg/ml)
Alternate names:
Protein-tyrosine phosphatase receptor type J; HPTP eta; Density-enhanced phosphatase 1; Susceptibility to colon cancer 1; HPTP beta-like tyrosine phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547225 375.00
Check your price
  • CD151 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710453
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Tetraspanin-24; Platelet-endothelial tetraspan antigen 3; Membrane glycoprotein SFA-1; GP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
710453 304.00
Check your price
  • CD151 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA181266
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Tetraspanin-24; Platelet-endothelial tetraspan antigen 3; Membrane glycoprotein SFA-1; GP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181266 425.00
Check your price
  • CD151 Monoclonal Antibody for IF, ICC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524175
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
Platelet-endothelial tetraspan antigen 3; Membrane glycoprotein SFA-1; GP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524175 275.00
Check your price
  • CD151 Monoclonal Antibody for Western Blot, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190964
  • Size 100µg
Applications
ELISA (Assay dependent), Flow Cytometry (1:50-1:100), Immunohistochemistry (Frozen) (1:50-1:150), Immunohistochemistry (Paraffin) (1:50-1:150), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Tetraspanin-24; Platelet-endothelial tetraspan antigen 3; Membrane glycoprotein SFA-1; GP27
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190964 309.00
Check your price
  • CD153 Monoclonal Antibody for Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523859
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1-4 µg/ml)
Alternate names:
CD30 ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523859 275.00
Check your price
  • CD153 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547045
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
CD30 ligand
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547045 349.00
Check your price
  • CD154 Monoclonal Antibody for Flow and IP
  • Host Hamster
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA133895
  • Size 100µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunoprecipitation (Assay dependent)
Alternate names:
Tumor necrosis factor ligand superfamily member 5; TNF-related activation protein; T-cell antigen Gp39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA133895 309.00
Check your price
  • CD154 Monoclonal Antibody for Western Blot, IHC, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523829
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (0.25 µg/10^6 cells), Immunohistochemistry (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Tumor necrosis factor ligand superfamily member 5; TNF-related activation protein; T-cell antigen Gp39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523829 275.00
Check your price
  • CD154 Monoclonal Antibody, PE conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate PE
  • Catalog # HMCD15404
  • Size 100µg
Applications
Flow Cytometry (0.1-0.2 ug per 1x10^6 cells)
Alternate names:
Tumor necrosis factor ligand superfamily member 5; TNF-related activation protein; T-cell antigen Gp39
Catalog # Size Price (AUD) Your Price (AUD) Qty
HMCD15404 216.00
Check your price
  • CD154 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546964
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Tumor necrosis factor ligand superfamily member 5; TNF-related activation protein; T-cell antigen Gp39
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546964 375.00
Check your price
  • CD155 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524315
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
nectin-like 5, nectin-like protein 5, PVR, PVS
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524315 275.00
Check your price
  • CD16 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112041
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112041 249.00
Check your price
  • CD16 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119364
  • Size 100µg
Applications
Flow Cytometry (2µg/ml), Immunoprecipitation (Assay Dependent)
Alternate names:
IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119364 309.00
Check your price
  • CD16 Monoclonal Antibody for Western Blot, Flow, IP and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119006
  • Size 100µg
Applications
Flow Cytometry (5-10 µg/ml), Functional Assay (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119006 199.00
Check your price
  • CD16 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119491
  • Size 100µg
Applications
Flow Cytometry (1:500)
Alternate names:
IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119491 225.00
Check your price
  • CD16 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # MA119340
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
IgG Fc receptor III-2; FcR-10; Fc-gamma RIII-alpha; CD16a antigen; IgG Fc receptor III-1; Fc-gamma RIII-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119340 199.00
Check your price
  • CD16 Polyclonal Antibody for Western Blot, IF, ICC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547230
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
FCGR3, FCGR3A, FCGR3B, CD16A, CD16B, Fc gamma Receptor III, Fc gamma Receptor 3, RP11-5K23.1, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, FCGR2, CD32, Fc gamma Receptor II, CD32 receptor 2, Fc gamma Receptor 2, FCGR2A, FCGR2B, FCGR2C, Low affinity immunoglobulin gamma Fc region receptor II-b, CDw32, Fc-gamma RII-b, Fc-gamma-RIIb, FcRII-b, FCG2, Low affinity immunoglobulin gamma Fc region receptor II-c, Fc-gamma RII-c, Fc-gamma-RIIc, FcRII-c
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547230 349.00
Check your price
  • CD163 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA180112
  • Size 100µg
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
CD163 antigen, hemoglobin scavenger receptor, macrophage-associated antigen, scavenger receptor cysteine-rich type 1 protein M130
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180112 499.00
Check your price
  • CD164 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524215
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells)
Alternate names:
Multi-glycosylated core protein 24; Endolyn
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524215 275.00
Check your price
  • CD166 Monoclonal Antibody for Western Blot and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523849
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (2 µg/ml)
Alternate names:
Activated leukocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523849 275.00
Check your price
  • CD166 Monoclonal Antibody for Western Blot, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523850
  • Size 100µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Activated leukocyte cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523850 275.00
Check your price