Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  140  150  160  170  180  190  200  210  220  228  229  230  231  232  233  234  235  236  237  238  239  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  260  270  280  290  300  310  320  330  340  400  500  600  700  800  900  1000  1100  1200  1630 
7261 - 7290 of 48881 products displayed
  • CD30 Monoclonal Antibody, PE conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate PE
  • Catalog # HMCD3004
  • Size 100µg
Applications
Flow Cytometry (0.1-0.2 ug per 1x10^6 cells)
Alternate names:
Lymphocyte activation antigen CD30; CD30L receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
HMCD3004 223.00
Check your price
  • CD30 Monoclonal Antibody for IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134431
  • Size 100µg
Applications
Flow Cytometry (2 µg for 10^5 cells or 100 µl whole blood), Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (Assay dependent)
Alternate names:
Lymphocyte activation antigen CD30; Ki-1 antigen; CD30L receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134431 399.00
Check your price
  • CD30 Monoclonal Antibody for IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119412
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (5 ug/ml)
Alternate names:
Lymphocyte activation antigen CD30; Ki-1 antigen; CD30L receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119412 199.00
Check your price
  • CD300a Monoclonal Antibody for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523927
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Myeloid-associated immunoglobulin-like receptor 1; Mast cell-derived paired immunoglobulin-like receptor 1; Leukocyte mono-Ig-like receptor 1; CD300 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523927 275.00
Check your price
  • CD300b Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524069
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Leukocyte mono-Ig-like receptor 5; Immune receptor expressed on myeloid cells 3; CD300 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524069 275.00
Check your price
  • CD300c Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521179
  • Size 100µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Immunoglobulin superfamily member 16; CMRF35-A1; CD300 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521179 325.00
Check your price
  • CD300e Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523940
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Polymeric immunoglobulin receptor 2; Immune receptor expressed on myeloid cells 2; CMRF35-A5; CD300 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523940 275.00
Check your price
  • CD300f Monoclonal Antibody for IF, ICC and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523943
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
NK inhibitory receptor; Immunoglobulin superfamily member 13; Immune receptor expressed on myeloid cells 1; CD300 antigen-like family member F
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523943 275.00
Check your price
  • CD300f Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524018
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Myeloid-associated immunoglobulin-like receptor 5; Leukocyte mono-Ig-like receptor 3; CD300 antigen-like family member F
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524018 275.00
Check your price
  • CD300f Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547399
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Myeloid-associated immunoglobulin-like receptor 5; Leukocyte mono-Ig-like receptor 3; CD300 antigen-like family member F
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547399 349.00
Check your price
  • CD300g Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547755
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Nepmucin; CD300 antigen-like family member G
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547755 349.00
Check your price
  • CD301 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547665
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Macrophage lectin 2; C-type lectin superfamily member 14; Macrophage galactose/N-acetylgalactosamine-specific lectin; Macrophage asialoglycoprotein-binding protein 1; MMGL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547665 375.00
Check your price
  • CD301a/b Polyclonal Antibody for Western Blot, Flow and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547609
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547609 349.00
Check your price
  • CD301b Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547407
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
CD301b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547407 349.00
Check your price
  • CD307 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524020
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
Immune receptor translocation-associated protein 2; Fc receptor homolog 5; BXMAS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524020 275.00
Check your price
  • CD307 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA548003
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Immune receptor translocation-associated protein 2; Fc receptor homolog 5; BXMAS1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548003 309.00
Check your price
  • CD31 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Bovine, Goat, Sheep
  • Conjugate Unconjugated
  • Catalog # MA180367
  • Size 100µg
Applications
Flow Cytometry (1/10-1/25), Immunoprecipitation (Assay Dependent)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180367 399.00
Check your price
  • CD31 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Goat, Sheep
  • Conjugate FITC
  • Catalog # MA180360
  • Size 100µg
Applications
Flow Cytometry (Neat)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180360 499.00
Check your price
  • CD31 Monoclonal Antibody for IHC (F), IHC (P), Flow, IP and ELISA
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA140074
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent.), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent.), Immunoprecipitation (Assay Dependent)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140074 399.00
Check your price
  • CD31 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 370700
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
370700 304.00
Check your price
  • CD31 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119199
  • Size 100µg
Applications
Flow Cytometry (1-10 ug/ml), Western Blot (Assay Dependent)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119199 225.00
Check your price
  • CD31 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119510
  • Size 100µg
Applications
Flow Cytometry (1:500)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119510 275.00
Check your price
  • CD31 Monoclonal Antibody for IF, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA180069
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1:10-1:100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:10-1:100)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180069 159.00
Check your price
  • CD31 Monoclonal Antibody for IF, ICC, IHC, Flow and IP
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA126196
  • Size 100µg
Applications
Flow Cytometry (1:50-1:100 (10 µl for 10^6 cells in 100 µl)), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Assay-dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126196 399.00
Check your price
  • CD31 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA182378
  • Size 100µg
Applications
Flow Cytometry (neat-1/10)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182378 349.00
Check your price
  • CD320 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547155
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Transcobalamin receptor; FDC-signaling molecule 8D6; 8D6 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547155 349.00
Check your price
  • CD33 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112099
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
gp67; Sialic acid-binding Ig-like lectin 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112099 225.00
Check your price
  • CD33 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA119522
  • Size 100µg
Applications
Flow Cytometry (1:1000)
Alternate names:
gp67; Sialic acid-binding Ig-like lectin 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119522 225.00
Check your price
  • CD33 Monoclonal Antibody, PE-Cyanine5.5 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine5.5
  • Catalog # A15454
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
gp67; Sialic acid-binding Ig-like lectin 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15454 249.00
Check your price
  • CD34 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Dog
  • Conjugate FITC
  • Catalog # MA181639
  • Size 100µg
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181639 499.00
Check your price