Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  150  160  170  180  190  200  210  220  230  240  241  242  243  244  245  246  247  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  280  290  300  310  320  330  340  350  400  500  600  700  800  900  1000  1100  1200  1630 
7621 - 7650 of 48882 products displayed
  • CEACAM1 Polyclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547842
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Murine hepatitis virus receptor; MHVR1; Biliary glycoprotein D; Biliary glycoprotein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547842 349.00
Check your price
  • CEACAM1/CEACAM3/CEACAM8 Monoclonal Antibody for IHC (F) and Flow
  • Host Rat
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA190031
  • Size 100µg
Applications
Flow Cytometry (10 µl for 10^6 cells in 100 µl), Immunohistochemistry (Frozen) (Assay dependent)
Alternate names:
Non-specific cross-reacting antigen NCA-95; Carcinoembryonic antigen CGM6; CD67 antigen; Biliary glycoprotein 1; Carcinoembryonic antigen CGM1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190031 309.00
Check your price
  • CEACAM1/CEACAM5 Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117767
  • Size 100µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunohistochemistry (Frozen) (1-2 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Biliary glycoprotein 1; Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117767 550.00
Check your price
  • CEACAM5/CEACAM6 Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117762
  • Size 100µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunohistochemistry (Frozen) (1-2 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen; Meconium antigen 100; Carcinoembryonic antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117762 550.00
Check your price
  • CEACAM6 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524164
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524164 275.00
Check your price
  • CEACAM6 Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117765
  • Size 100µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 ug/10^6 cells), Immunohistochemistry (Frozen) (1-2 ug/10^6 cells), Western Blot (4 ug/ml)
Alternate names:
Normal cross-reacting antigen; Non-specific crossreacting antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117765 550.00
Check your price
  • CEACAM7 Monoclonal Antibody for Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117764
  • Size 100µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 µg/10^6 cells)
Alternate names:
Carcinoembryonic antigen CGM2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117764 550.00
Check your price
  • CEACAM8 Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126144
  • Size 100µg
Applications
Flow Cytometry (10 µl for 5x10^5 cells), Immunohistochemistry (Frozen) (1:25-1:50)
Alternate names:
Non-specific cross-reacting antigen NCA-95; Carcinoembryonic antigen CGM6; CD67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126144 375.00
Check your price
  • CEACAM8 Monoclonal Antibody for Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA117768
  • Size 100µg
Applications
ELISA (1:200-1:400), Flow Cytometry (1.2 µg/ 10^6 cells)
Alternate names:
Non-specific cross-reacting antigen NCA-95; Carcinoembryonic antigen CGM6; CD67 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA117768 550.00
Check your price
  • CECR2 Monoclonal Antibody for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 730031
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
Cat eye syndrome critical region protein 2, CECR2, KIAA1740
Catalog # Size Price (AUD) Your Price (AUD) Qty
730031 309.00
Check your price
  • CELSR2 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547869
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Multiple epidermal growth factor-like domains protein 3; Flamingo homolog 3; Epidermal growth factor-like protein 2; Cadherin family member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547869 349.00
Check your price
  • CELSR3 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538869
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Multiple epidermal growth factor-like domains protein 2; Flamingo homolog 1; Epidermal growth factor-like protein 1; Cadherin family member 11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538869 309.00
Check your price
  • CEND Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538870
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
BM88 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538870 309.00
Check your price
  • CENPF Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123185
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Mitosin; Kinetochore protein CENPF; AH antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123185 309.00
Check your price
  • CENPH Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA522523
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Interphase centromere complex protein 35
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA522523 309.00
Check your price
  • CENPI Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA14315
  • Size 100µg
Applications
Western Blot (1:250)
Alternate names:
Follicle-stimulating hormone primary response protein; FSH primary response protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14315 309.00
Check your price
  • CENPP Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523311
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
CENP-P
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523311 309.00
Check your price
  • Centaurin alpha-1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # MA124606
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124606 309.00
Check your price
  • CEP131 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538978
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Pre-acrosome localization protein 1; 5-azacytidine-induced protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538978 309.00
Check your price
  • CEP135 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539089
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Centrosomal protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539089 309.00
Check your price
  • CEP170 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 413200
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
KARP-1-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
413200 305.00
Check your price
  • CEP290 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539088
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Tumor antigen se2-2; Nephrocystin-6; Cancer/testis antigen 87; Bardet-Biedl syndrome 14 protein; Bardet-Biedl syndrome 14 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539088 309.00
Check your price
  • CEP350 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539093
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Centrosome-associated protein of 350 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539093 309.00
Check your price
  • CEP41 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539090
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Testis-specific gene A14 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539090 309.00
Check your price
  • CEP76 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539091
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
C18orf9, HsT1705, centrosomal protein of 76 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539091 309.00
Check your price
  • CEP78 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539092
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
C9orf81, IP63, centrosomal protein of 78 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539092 309.00
Check your price
  • CEP95 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539114
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Coiled-coil domain-containing protein 45
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539114 309.00
Check your price
  • CEP97 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539223
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Leucine-rich repeat and IQ domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539223 309.00
Check your price
  • CER1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547233
  • Size 100µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Cerberus-related protein; Cerberus-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547233 349.00
Check your price
  • Cerebellin 2 Monoclonal Antibody for IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524324
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (8-25 µg/ml)
Alternate names:
CBLN2, UNQ1892/PRO4338, Cerebellin2, Cerebellin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524324 275.00
Check your price