Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  50  60  70  80  90  100  110  120  200  300  400  500  600  700  800  900  1000  1611 
751 - 780 of 48310 products displayed
  • hCG Monoclonal Antibody for Western Blot, IHC (P), IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9812
  • Size 1 mg
Applications
ELISA (2-10µg/ml), Immunohistochemistry (Paraffin) (1:20), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9812 600.00
Check your price
  • hCG Monoclonal Antibody for Western Blot, IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9813
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (5µg/ml), Radioimmune Assays (Assay Dependent), Western Blot (1:250-1:500)
Alternate names:
Chorionic gonadotrophin chain beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9813 600.00
Check your price
  • Hemoglobin Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9505
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Hemoglobin alpha chain; Alpha-globin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9505 600.00
Check your price
  • Hemoglobin Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH9506
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
Hemoglobin alpha chain; Alpha-globin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9506 600.00
Check your price
  • Hemoglobin Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129342
  • Size 1 mg
Applications
ELISA (5 µg/ml), Western Blot (1:1000-1:5000)
Alternate names:
Hemoglobin alpha chain; Alpha-globin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129342 309.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9701
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9701 600.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9704
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9704 600.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9705
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9705 600.00
Check your price
  • Hepatitis B Virus Surface Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MIH9706
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH9706 600.00
Check your price
  • Hepatitis B Virus Surface Polyclonal Antibody for Western Blot and ELISA
  • Host Horse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA173083
  • Size 1 mg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HepB, HepB surface antigen, Hepatitis B Surface Antigen, HBsAg
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173083 349.00
Check your price
  • Hexokinase Polyclonal Antibody, HRP conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Yeast
  • Conjugate HRP
  • Catalog # PA127232
  • Size 1 mg
Applications
ELISA (1:4000-1:20,000), Western Blot (1:500-1:2000)
Alternate names:
Hexokinase-A; Hexokinase PI
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127232 349.00
Check your price
  • HSV Type 1 VP21/VP22a Monoclonal Antibody for Western Blot, IF, IP and IM
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516798
  • Size 1 mg
Applications
Immunofluorescence (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516798 375.00
Check your price
  • HSV Type 2 gG Monoclonal Antibody for Western Blot, IF, IHC (F) and IP
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516575
  • Size 1 mg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:100), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, Glycoprotein G
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516575 375.00
Check your price
  • Human IgG Isotype Control, HRP conjugate for BLOCK and Ctrl
  • Host Human
  • Target Species Not Applicable
  • Conjugate HRP
  • Catalog # 31465
  • Size 1 mg
Applications
Blocking Assay (Assay Dependent), Control (Assay-Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Negative Control, Isotype Control, Immunoglobulin Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
31465 109.00
Check your price
  • Human Interleukin 4 (IL-4) for Western Blot, IP, ELISA and Ctrl
  • Host Yeast
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIL4MI
  • Size 1 mg
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Pitrakinra; Lymphocyte stimulatory factor 1; Binetrakin; B-cell stimulatory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL4MI 3,625.00
Check your price
  • Human Serum Albumin Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH3003
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
cell growth inhibiting protein 42, growth-inhibiting protein 20, serum albumin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH3003 600.00
Check your price
  • Human Serum Albumin Monoclonal Antibody for IP, ELISA, RIA and CA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH3005
  • Size 1 mg
Applications
Competition Assay (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
cell growth inhibiting protein 42, growth-inhibiting protein 20, serum albumin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH3005 600.00
Check your price
  • Human Serum Albumin Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH3001
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
cell growth inhibiting protein 42; growth-inhibiting protein 20; serum albumin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH3001 600.00
Check your price
  • Human Serum Albumin Monoclonal Antibody for IP, ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MIH3002
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
cell growth inhibiting protein 42, growth-inhibiting protein 20, serum albumin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MIH3002 600.00
Check your price
  • IFN gamma Monoclonal Antibody for Western Blot, ICC, IHC (F), ELISA and Neu
  • Host Mouse
  • Target Species Rat, Mouse
  • Conjugate Unconjugated
  • Catalog # ARC4033
  • Size 1 mg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
IFN-gamma, IFN-y, Interferon y, IFNG
Catalog # Size Price (AUD) Your Price (AUD) Qty
ARC4033 433.00
Check your price
  • IFN gamma Polyclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PBOIFNGI
  • Size 1 mg
Applications
ELISA (Assay Dependent)
Alternate names:
IFN-gamma; boIFNG; interferon gamma; interferon gamma type 2; interferon-gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PBOIFNGI 349.00
Check your price
  • IFN gamma Polyclonal Antibody for Western Blot, IHC, ELISA and Neu
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # ARC4833
  • Size 1 mg
Applications
Immunohistochemistry (Assay Dependent), Neutralization (Assay Dependent)
Alternate names:
IFN-gamma, IFN-y, Interferon y, IFNG
Catalog # Size Price (AUD) Your Price (AUD) Qty
ARC4833 330.00
Check your price
  • IFN gamma Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PP700
  • Size 1 mg
Applications
Western Blot (Assay Dependent)
Alternate names:
IFN-gamma; interferon gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PP700 349.00
Check your price
  • IFN gamma Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # P700
  • Size 1 mg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Immune interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
P700 349.00
Check your price
  • IL-1 alpha (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75583
  • Size 1 mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75583 2,600.00
Check your price
  • IL-1 alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # P420A
  • Size 1 mg
Applications
Western Blot (Assay Dependent)
Alternate names:
Hematopoietin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
P420A 309.00
Check your price
  • IL-1 beta (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75586
  • Size 1 mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75586 2,600.00
Check your price
  • IL-1 beta Polyclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PBOIL1BI
  • Size 1 mg
Applications
ELISA (Assay Dependent)
Alternate names:
IL-1 beta; interleukin-1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PBOIL1BI 275.00
Check your price
  • IL-1 beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PP425
  • Size 1 mg
Applications
Western Blot (Assay Dependent)
Alternate names:
IL-1 beta; interleukin-1 beta; prointerleukin-1 beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PP425 304.00
Check your price
  • IL-1 beta Polyclonal Antibody for Western Blot, IHC, ELISA and Neu
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # P420B
  • Size 1 mg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Neutralization (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catabolin
Catalog # Size Price (AUD) Your Price (AUD) Qty
P420B 349.00
Check your price