Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  248  249  250  251  252  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  280  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1605 
7861 - 7890 of 48148 products displayed
  • COL23A1 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538896
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538896 309.00
Check your price
  • COL4A2 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538875
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
canstatin, collagen alpha-2(IV) chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538875 309.00
Check your price
  • COL4A3 (Cleaved Leu1425) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539876
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Goodpasture antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539876 309.00
Check your price
  • COL4A3 (Cleaved Pro1426) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA539877
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Goodpasture antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539877 309.00
Check your price
  • COL4A3 Polyclonal Antibody for IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537854
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (4-6 µg/ml)
Alternate names:
Goodpasture antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537854 309.00
Check your price
  • COL4A3 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA538876
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Goodpasture antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538876 309.00
Check your price
  • COL4A3BP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112911
  • Size 100µg
Applications
Western Blot (Assay dependent)
Alternate names:
StAR-related lipid transfer protein 11; START domain-containing protein 11; Goodpasture antigen-binding protein; Ceramide transfer protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112911 325.00
Check your price
  • COL4A3BP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112935
  • Size 100µg
Applications
Western Blot (1-5 µg/ml)
Alternate names:
StAR-related lipid transfer protein 11; START domain-containing protein 11; Goodpasture antigen-binding protein; Ceramide transfer protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112935 325.00
Check your price
  • COL4A4 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA538877
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
COL1A1, COL1A2, COL2A1, COL3A1, COL4A1, COL4A2, COL4A3, COL4A4, COL4A5, COL4A6, COL5A1, COL5A2, COL5A3, Collagen 1, Collagen 2, Collagen 3, Collagen 4, Collagen 5, pan Collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538877 309.00
Check your price
  • COL4A5 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538878
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
COL1A1, COL1A2, COL2A1, COL3A1, COL4A1, COL4A2, COL4A3, COL4A4, COL4A5, COL4A6, COL5A1, COL5A2, COL5A3, Collagen 1, Collagen 2, Collagen 3, Collagen 4, Collagen 5, pan Collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538878 309.00
Check your price
  • COL4A6 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538879
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
COL1A1, COL1A2, COL2A1, COL3A1, COL4A1, COL4A2, COL4A3, COL4A4, COL4A5, COL4A6, COL5A1, COL5A2, COL5A3, Collagen 1, Collagen 2, Collagen 3, Collagen 4, Collagen 5, pan Collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538879 309.00
Check your price
  • COL5A2 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538880
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Collagen alpha-2(V) chain, COL5A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538880 309.00
Check your price
  • COL5A3 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538881
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
COL1A1, COL1A2, COL2A1, COL3A1, COL4A1, COL4A2, COL4A3, COL4A4, COL4A5, COL4A6, COL5A1, COL5A2, COL5A3, Collagen 1, Collagen 2, Collagen 3, Collagen 4, Collagen 5, pan Collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538881 309.00
Check your price
  • COL6A2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538882
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
PP3610, collagen VI, alpha-2 polypeptide, collagen alpha-2(VI) chain, human mRNA for collagen VI alpha-2 C-terminal globular domain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538882 309.00
Check your price
  • COL6A3 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538883
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538883 309.00
Check your price
  • COL9A2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538886
  • Size 100µg
Applications
Western Blot (1:500-1:3000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538886 309.00
Check your price
  • COL9A3 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538887
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538887 309.00
Check your price
  • COLEC12 Polyclonal Antibody for Western Blot, IF, ICC, Flow and BLOCK
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547369
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Scavenger receptor with C-type lectin; Scavenger receptor class A member 4; Nurse cell scavenger receptor 2; Collectin placenta protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547369 349.00
Check your price
  • COLEC12 Polyclonal Antibody for Western Blot, Flow and BLOCK
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547456
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
Scavenger receptor with C-type lectin; Collectin placenta protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547456 349.00
Check your price
  • Collagen I Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA1141
  • Size 100µg
Applications
ELISA (0.4-50 µg/ml), Immunohistochemistry (Frozen) (1:200-1:1000), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:500-1:2000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1141 349.00
Check your price
  • Collagen I Monoclonal Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183073
  • Size 100µg
Applications
ELISA (1:1000), Immunohistochemistry (Frozen) (1:20)
Alternate names:
Alpha-2 type I collagen; Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183073 349.00
Check your price
  • Collagen I Polyclonal Antibody for IHC (F) and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA135998
  • Size 100µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA135998 375.00
Check your price
  • Collagen I Polyclonal Antibody for IHC (F), ELISA and DB
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA136145
  • Size 100µg
Applications
Dot blot (Assay dependent), ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136145 349.00
Check your price
  • Collagen I Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA126204
  • Size 100µg
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:5000-1:10,000)
Alternate names:
Alpha-1 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126204 399.00
Check your price
  • Collagen I/III Polyclonal Antibody for IHC (F) and ELISA
  • Host Rabbit
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA135035
  • Size 100µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alpha-1 type I collagen; Alpha-2 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA135035 399.00
Check your price
  • Collagen I/III Polyclonal Antibody for IHC (F) and ELISA
  • Host Rabbit
  • Target Species Dog
  • Conjugate Unconjugated
  • Catalog # PA136069
  • Size 100µg
Applications
ELISA (Assay dependent.), Immunohistochemistry (Frozen) (Assay dependent.)
Alternate names:
Alpha-1 type I collagen; Alpha-2 type I collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136069 349.00
Check your price
  • Collagen I/III Polyclonal Antibody for IHC (F), ELISA and DB
  • Host Rabbit
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # PA136140
  • Size 100µg
Applications
Dot blot (Assay dependent), ELISA (Assay dependent.), Immunohistochemistry (Frozen) (1:20)
Alternate names:
COL1A1, COL1A2, COL3A1, Collagen 1, Collagen 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA136140 375.00
Check your price
  • Collagen II Monoclonal Antibody for IHC (F), IHC (P), ELISA and FN
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA140065
  • Size 100µg
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140065 550.00
Check your price
  • Collagen II Monoclonal Antibody for IHC (F), IHC (P), ELISA and FN
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA140066
  • Size 100µg
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent.), Immunohistochemistry (Paraffin) (Assay Dependent.)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140066 550.00
Check your price
  • Collagen II Polyclonal Antibody for Western Blot, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA126206
  • Size 100µg
Applications
ELISA (1:4000-1:8000), Immunohistochemistry (Frozen) (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:100), Western Blot (1:5000-1:10,000)
Alternate names:
Alpha-1 type II collagen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126206 349.00
Check your price