Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  160  170  180  190  200  210  220  230  240  253  254  255  256  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  290  300  310  320  330  340  350  360  400  500  600  700  800  900  1000  1100  1200  1611 
8011 - 8040 of 48315 products displayed
  • A26C2/A26C3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539360
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Prostate, ovary, testis-expressed protein on chromosome 22; ANKRD26-like family C member 3; Prostate, ovary, testis-expressed protein on chromosome 14; ANKRD26-like family C member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539360 309.00
Check your price
  • A33 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524139
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Glycoprotein A33
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524139 275.00
Check your price
  • A33 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547444
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Glycoprotein A33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547444 349.00
Check your price
  • A33 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547386
  • Size 100 µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Glycoprotein A33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547386 349.00
Check your price
  • AA5 Virus VP3 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123359
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
HADV-5, AdV5, AV5, Adeno-associated virus 5, AAV-5 VP3, Adeno-associated 5 Virus VP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123359 375.00
Check your price
  • AA5 Virus VP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA14108
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
HADV-5, AdV5, AV5, Adeno-associated virus 5, AAV-5 VP3, Adeno-associated 5 Virus VP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14108 309.00
Check your price
  • AARS2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537957
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Alanyl-tRNA synthetase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537957 309.00
Check your price
  • AASDHPPT Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539222
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
LYS5 ortholog; Alpha-aminoadipic semialdehyde dehydrogenase-phosphopantetheinyl transferase; 4'-phosphopantetheinyl transferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539222 309.00
Check your price
  • AATK Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524176
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
p35-binding protein; Lemur tyrosine kinase 1; CDK5-binding protein; Brain apoptosis-associated tyrosine kinase; Apoptosis-associated tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524176 275.00
Check your price
  • AAV Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA14106
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
HADV, AdV, AAV, Adeno-Associated Virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14106 375.00
Check your price
  • AAV4 Virus Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123358
  • Size 100 µg
Applications
ELISA (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
HADV-4, AdV4, AV4, AAV-4, Adeno-associated virus 4, Adeno-associated 4 Virus
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123358 349.00
Check your price
  • AAV4 Virus VP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA14107
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
HADV-4, AdV4, AV4, Adeno-associated virus 4, AAV-4 VP3, Adeno-associated 4 Virus VP3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14107 309.00
Check your price
  • ABCA1 Monoclonal Antibody for Western Blot, IHC (F), IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191401
  • Size 100 µg
Applications
ELISA (3-6 µg/ml), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:2000)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191401 399.00
Check your price
  • ABCA10 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521013
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
FKHL6, FREAC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521013 309.00
Check your price
  • ABCA8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539040
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
ATP-binding cassette sub-family A member 8, KIAA0822
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539040 309.00
Check your price
  • ABCB9 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521478
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
TAP-like protein; ATP-binding cassette transporter 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521478 309.00
Check your price
  • ABCF1 Monoclonal Antibody for IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518335
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml)
Alternate names:
TNF-alpha-stimulated ABC protein; ATP-binding cassette 50
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518335 309.00
Check your price
  • ABHD12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538994
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Abhydrolase domain-containing protein 12; 2-arachidonoylglycerol hydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538994 309.00
Check your price
  • ABHD12B Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538995
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Variant 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538995 309.00
Check your price
  • ABHD14A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538996
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538996 309.00
Check your price
  • ABHD2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538997
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Protein PHPS1-2; Lung alpha/beta hydrolase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538997 309.00
Check your price
  • ABHD4 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538998
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Lyso-N-acylphosphatidylethanolamine lipase; Alpha/beta-hydrolase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538998 309.00
Check your price
  • ABHD6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538999
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Western Blot (1:500-1:1000)
Alternate names:
Abhydrolase domain-containing protein 6; 2-arachidonoylglycerol hydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538999 309.00
Check your price
  • ABHD7 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539000
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Epoxide hydrolase-related protein; Abhydrolase domain-containing protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539000 309.00
Check your price
  • ABHD9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539001
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Abhydrolase domain-containing protein 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539001 309.00
Check your price
  • ACAA1 Monoclonal Antibody for Western Blot, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # A21984
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (1.0 ug/ml), Western Blot (5 ug/ml )
Alternate names:
Peroxisomal 3-oxoacyl-CoA thiolase; Beta-ketothiolase; Acetyl-CoA acyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21984 330.00
Check your price
  • ACAD11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112907
  • Size 100 µg
Applications
Western Blot (1-5 µg/ml)
Alternate names:
FLJ12592, MGC150619
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112907 309.00
Check your price
  • ACAT1 Monoclonal Antibody for ICC, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A21985
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (5 ug/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
T2; Acetoacetyl-CoA thiolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21985 309.00
Check your price
  • ACAT2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518338
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Cytosolic acetoacetyl-CoA thiolase; Acetyl-CoA transferase-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518338 309.00
Check your price
  • ACAT2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124413
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (4-8 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Cytosolic acetoacetyl-CoA thiolase; Acetyl-CoA transferase-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124413 399.00
Check your price