Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  257  258  259  260  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  290  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1630 
8131 - 8160 of 48882 products displayed
  • CST1 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539123
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Salivary cystatin-SA-1; Cystatin-1; Cystain-SA-I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539123 309.00
Check your price
  • CST11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539119
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
CST8L, SC13, dJ322G13.6, SC13delta, cystatin-11, cystatin-related epididymal-specific protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539119 309.00
Check your price
  • CST9L Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539120
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Testatin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539120 309.00
Check your price
  • CSTF1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518666
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Cleavage stimulation factor 50 kDa subunit; CF-1 50 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518666 309.00
Check your price
  • CSTF2 Monoclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518668
  • Size 100µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (10 µg/ml), Immunofluorescence (10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Cleavage stimulation factor 64 kDa subunit; CF-1 64 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518668 309.00
Check your price
  • CSTF2 Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA537985
  • Size 100µg
Applications
Immunoprecipitation (2µg/500µg lysate), Western Blot (2µg)
Alternate names:
Cleavage stimulation factor 64 kDa subunit; CF-1 64 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537985 309.00
Check your price
  • CSTF2T Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539109
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
TauCstF-64; Cleavage stimulation factor 64 kDa subunit tau variant; CF-1 64 kDa subunit tau variant
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539109 309.00
Check your price
  • CSTF3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537856
  • Size 100µg
Applications
Western Blot (0.03-0.1 µg/ml)
Alternate names:
Cleavage stimulation factor 77 kDa subunit; CF-1 77 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537856 309.00
Check your price
  • CSTL1 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539121
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
RCET11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539121 309.00
Check your price
  • CTBS Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524198
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
CTBS, chitobiase, di-N-acetyl-, CTBdi-N-acetylchitobiase, EC 3.2.1.-
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524198 275.00
Check your price
  • CTCF Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141663
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
CTCFL paralog; CCCTC-binding factor; 11-zinc finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141663 309.00
Check your price
  • CTDSP1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA537961
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.1-0.3 µg/ml)
Alternate names:
Small C-terminal domain phosphatase 1; Nuclear LIM interactor-interacting factor 3; Golli-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537961 309.00
Check your price
  • CTDSP2 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520929
  • Size 100µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Small CTD phosphatase 2; Small C-terminal domain phosphatase 2; Protein OS-4; Nuclear LIM interactor-interacting factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520929 309.00
Check your price
  • CTHRC1 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538054
  • Size 100µg
Applications
Immunofluorescence (20 µg/ml), Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Protein NMTC1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538054 309.00
Check your price
  • CtIP Monoclonal Antibody for Western Blot, IF, ICC, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123304
  • Size 100µg
Applications
ChIP assay (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
Sporulation in the absence of SPO11 protein 2 homolog; Retinoblastoma-interacting protein and myosin-like; Retinoblastoma-binding protein 8; CtBP-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123304 349.00
Check your price
  • CTLA-4 Monoclonal Antibody, PE conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate PE
  • Catalog # HMCD15204
  • Size 100µg
Applications
Flow Cytometry (0.1-0.2 ug per 1x10^6 cells)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
HMCD15204 267.00
Check your price
  • CTLA-4 Monoclonal Antibody for Western Blot, Flow and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523842
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (2.5-10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523842 275.00
Check your price
  • CTLA-4 Monoclonal Antibody for Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112205
  • Size 100µg
Applications
Neutralization (Assay Dependent)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112205 425.00
Check your price
  • CTLA-4 Monoclonal Antibody, PE conjugate for Flow
  • Host Armenian Hamster
  • Target Species Mouse
  • Conjugate PE
  • Catalog # A27090
  • Size 100µg
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
A27090 162.00
Check your price
  • CTLA-4 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547547
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Cytotoxic T-lymphocyte-associated antigen 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547547 375.00
Check your price
  • CTNNA3 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA106311
  • Size 100µg
Applications
Immunocytochemistry (Assay dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:25-1:50), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Cadherin-associated protein; Alpha T-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106311 375.00
Check your price
  • CUBN Polyclonal Antibody for Western Blot and IHC
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547527
  • Size 100µg
Applications
Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Intrinsic factor-vitamin B12 receptor; Intrinsic factor-cobalamin receptor; Intestinal intrinsic factor receptor; 460 kDa receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547527 349.00
Check your price
  • CUG-BP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA14130
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
RNA-binding protein BRUNOL-3; Neuroblastoma apoptosis-related RNA-binding protein; ELAV-type RNA-binding protein 3; CUG-BP- and ETR-3-like factor 2; CUG triplet repeat RNA-binding protein 2; Bruno-like protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14130 309.00
Check your price
  • Cullin 1 Monoclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 322400
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Immunoprecipitation (5-10 ug), Western Blot (1-3 ug/ml)
Alternate names:
CUL-1, cullin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
322400 305.00
Check your price
  • Cullin 1 Polyclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 718700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
CUL-1, cullin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
718700 304.00
Check your price
  • Cullin 1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124260
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
CUL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124260 399.00
Check your price
  • Cullin 2 Polyclonal Antibody for Western Blot, IHC (P), Flow, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 511800
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:50), Immunoprecipitation (2-3 µg/ml), Western Blot (1:250-1:1000)
Alternate names:
CUL-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
511800 307.00
Check your price
  • Cullin 3 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 342200
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CUL-3, cullin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
342200 304.00
Check your price
  • Cullin 5 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 399000
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Vasopressin-activated calcium-mobilizing receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
399000 309.00
Check your price
  • CX3CL1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA1031
  • Size 100µg
Applications
ELISA (Assay dependent), Western Blot (1:1000)
Alternate names:
Small-inducible cytokine D1; Neurotactin; CX3C membrane-anchored chemokine; C-X3-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1031 349.00
Check your price