Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  170  180  190  200  210  220  230  240  250  261  262  263  264  265  266  267  268  269  270  271  272  273  274  275  276  277  278  279  280  281  282  283  284  285  286  287  288  289  290  300  310  320  330  340  350  360  370  400  500  600  700  800  900  1000  1100  1200  1630 
8251 - 8280 of 48882 products displayed
  • Cyclin B2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Mouse, Non-human primate, Xenopus, Human
  • Conjugate Unconjugated
  • Catalog # MA1156
  • Size 100µg
Applications
Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:500), Immunoprecipitation (1:1000), Western Blot (1:1000)
Alternate names:
CCNB2, Cyclin B2, HsT17299, G2/Mitotic-Specific Cyclin-B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1156 349.00
Check your price
  • Cyclin D1 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701421
  • Size 100µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
701421 309.00
Check your price
  • Cyclin D1 Oligoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710428
  • Size 100µg
Applications
Flow Cytometry (1-3 ug x 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Western Blot (2-3 ug/ml)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
710428 309.00
Check your price
  • Cyclin D1 Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 333500
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Immunohistochemistry (2-10 ug/ml), Immunoprecipitation (5 ug), Western Blot (0.3-1.0 ug/ml)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
333500 304.00
Check your price
  • Cyclin D1 Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA110324
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (2-8 ug/ml), Immunocytochemistry (1 ug/ml), Immunohistochemistry (Frozen) (2 ug/ml), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (1 ug/ml), Western Blot (1 ug/ml)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110324 249.00
Check your price
  • Cyclin D1 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA112296
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112296 309.00
Check your price
  • Cyclin D1 Monoclonal Antibody for Western Blot, IF, IHC, Flow, IP and IA
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # AHF0082
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay-Dependent), Inhibition Assays (Assay Dependent), Western Blot (Assay-Dependent)
Alternate names:
PRAD1 oncogene; BCL-1 oncogene; B-cell lymphoma 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHF0082 304.00
Check your price
  • Cyclin D2 Monoclonal Antibody for Western Blot, IF, IHC, Flow, IP and IA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # AHF0112
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay-Dependent), Inhibition Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Vin-1 proto-oncogene
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHF0112 309.00
Check your price
  • Cyclin D2 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112297
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Vin-1 proto-oncogene
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112297 349.00
Check your price
  • Cyclin D3 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112298
  • Size 100µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
RP5-973N23.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112298 309.00
Check your price
  • Cyclin E Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human
  • Conjugate
  • Catalog # MA112295
  • Size 100µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CCNE
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112295 Not Orderable Online
Check your price
  • Cyclin E Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14123
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
CCNE
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14123 349.00
Check your price
  • Cyclin E2 Monoclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701814
  • Size 100µg
Applications
Flow Cytometry (2-4 µg x 10^6 cells), Western Blot (1- 2 µg/ml)
Alternate names:
G1/S-Specific Cyclin-E2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701814 309.00
Check your price
  • Cyclin E2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14122
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
CYCE2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14122 309.00
Check your price
  • Cyclin G1 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518550
  • Size 100µg
Applications
ELISA (Assay-Dependent)
Alternate names:
Cyclin-G1, Cyclin-G, CCNG1, CCNG, CYCG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518550 309.00
Check your price
  • Cyclin G2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA124255
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124255 399.00
Check your price
  • Cyclophilin 40 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1028
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Rotamase D; Cyclophilin-related protein; Cyclophilin-40; 40 kDa peptidyl-prolyl cis-trans isomerase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1028 375.00
Check your price
  • Cyclophilin A for Ctrl
  • Host E. Coli
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # RP020
  • Size 100µg
Applications
Control (Assay Dependent)
Alternate names:
CyP A, Peptidyl-prolyl cis-trans isomerase A, EC 5.2.1.8, PPIase, Rotamase, Cyclosporin A-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP020 419.00
Check your price
  • Cyclophilin B Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 370600
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
S-cyclophilin; Rotamase B; Cyclophilin B; CYP-S1
Catalog # Size Price (AUD) Your Price (AUD) Qty
370600 304.00
Check your price
  • Cyclophilin B Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1027A
  • Size 100µg
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (0.5 µg/ml)
Alternate names:
S-cyclophilin; Rotamase B; Cyclophilin B; CYP-S1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1027A 225.00
Check your price
  • Cyclophilin D Monoclonal Antibody for Western Blot and ICC
  • Host Mouse
  • Target Species Bovine, C. elegans, Human, Rat
  • Conjugate Unconjugated
  • Catalog # 455900
  • Size 100µg
Applications
Immunocytochemistry (10 ug/ml), Western Blot (1.0 ug/ml)
Alternate names:
Rotamase F; Cyclophilin F; Cyclophilin D; Mitochondrial cyclophilin
Catalog # Size Price (AUD) Your Price (AUD) Qty
455900 361.00
Check your price
  • Cyclosporin A Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA191110
  • Size 100µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191110 275.00
Check your price
  • CYLD Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Bovine, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # 437700
  • Size 100µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Ubiquitin-specific-processing protease CYLD; Ubiquitin thioesterase CYLD; Deubiquitinating enzyme CYLD
Catalog # Size Price (AUD) Your Price (AUD) Qty
437700 306.00
Check your price
  • CYLD Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141097
  • Size 100µg
Applications
Immunocytochemistry (5-10 µg/ml), Immunofluorescence (5-10 µg/ml), Immunohistochemistry (Assay-Dependent), Western Blot (2-4 µg/ml)
Alternate names:
Ubiquitin-specific-processing protease CYLD; Ubiquitin thioesterase CYLD; Deubiquitinating enzyme CYLD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141097 375.00
Check your price
  • CYP11B1/CYP11B2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539581
  • Size 100µg
Applications
Western Blot (1:500-1:3000)
Alternate names:
Steroid 18-hydroxylase; Cytochrome P-450C18; Cytochrome P-450Aldo; CYPXIB2; Aldosterone-synthesizing enzyme; Aldosterone synthase; Steroid 11-beta-hydroxylase; Cytochrome P-450c11; CYPXIB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539581 309.00
Check your price
  • CYP1B1 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519619
  • Size 100µg
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:1500), Western Blot (1 µg/ml)
Alternate names:
CYPIB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519619 349.00
Check your price
  • CYP2A13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538906
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
CYPIIA13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538906 309.00
Check your price
  • CYP2D10 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA537998
  • Size 100µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Debrisoquine 4-hydroxylase; Cytochrome P450-DB5; Cytochrome P450-CMF1B; CYPIID10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537998 309.00
Check your price
  • CYP2D6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539598
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Debrisoquine 4-hydroxylase; Cytochrome P450-DB1; Cholesterol 25-hydroxylase; CYPIID6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539598 309.00
Check your price
  • CYP2D6 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539599
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Debrisoquine 4-hydroxylase; Cytochrome P450-DB1; Cholesterol 25-hydroxylase; CYPIID6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539599 309.00
Check your price