Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  43  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  72  80  90  100  110  120  130  140  150  200  300  400  500  600  700  800  900  1000  1605 
1711 - 1740 of 48136 products displayed
  • Influenza A H1N1 Antibody, FITC
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA127201
  • Size 1 mL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
Influenza H1N1, H1N1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127201 399.00
Check your price
  • Influenza A H1N1 Antibody, HRP
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA533140
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:20-1:200)
Alternate names:
Influenza H1N1, H1N1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533140 550.00
Check your price
  • Influenza A H1N1 Virions Antibody, Biotin
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173046
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza H1N1, H1N1, H1N1 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173046 349.00
Check your price
  • Influenza A H1N1 Virions Antibody, FITC
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173044
  • Size 1 mL
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
Influenza H1N1, H1N1, H1N1 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173044 309.00
Check your price
  • Influenza A H1N1 Virions Antibody, HRP
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173045
  • Size 1 mL
Applications
ELISA (1:200-1:1000)
Alternate names:
Influenza H1N1, H1N1, H1N1 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173045 309.00
Check your price
  • Influenza A H3N2 Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA127282
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Assay-dependent), Western Blot (Assay-dependent)
Alternate names:
Influenza H3N2, H3N2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127282 375.00
Check your price
  • Influenza A H3N2 Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17222
  • Size 1 mL
Applications
Immunofluorescence (1:1000)
Alternate names:
Influenza H3N2, H3N2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17222 375.00
Check your price
  • Influenza A H3N2 Antibody, Biotin
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA126179
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1000)
Alternate names:
Influenza H3N2, H3N2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126179 399.00
Check your price
  • Influenza A H3N2 M1 Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17223
  • Size 1 mL
Applications
Immunofluorescence (1:1000)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17223 309.00
Check your price
  • Influenza A H3N2 M1 Antibody, Biotin
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173052
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173052 309.00
Check your price
  • Influenza A H3N2 M1 Antibody, FITC
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173050
  • Size 1 mL
Applications
Immunofluorescence (1:10)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173050 349.00
Check your price
  • Influenza A H3N2 M1 Antibody, HRP
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173051
  • Size 1 mL
Applications
ELISA (1:200)
Alternate names:
Influenza H3N2, H3N2, H3N2 Matrix protein, H3N2 M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173051 375.00
Check your price
  • Influenza A H3N2 Virions Antibody, Biotin
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173049
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza H3N2, H3N2, H3N2 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173049 309.00
Check your price
  • Influenza A H3N2 Virions Antibody, FITC
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173047
  • Size 1 mL
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
Influenza H3N2, H3N2, H3N2 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173047 375.00
Check your price
  • Influenza A H3N2 Virions Antibody, HRP
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173048
  • Size 1 mL
Applications
ELISA (1:200)
Alternate names:
Influenza H3N2, H3N2, H3N2 virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173048 309.00
Check your price
  • Influenza A M1 Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA185626
  • Size 1 mL
Applications
Immunofluorescence (1:50-1:160)
Alternate names:
Influenza Matrix protein M1, Influenza M1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185626 399.00
Check your price
  • Influenza A NP Antibody (Monoclonal, D67J)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17057
  • Size 1 mL
Applications
ELISA (1:20), Immunocytochemistry (1:20), Immunofluorescence (1:10), Immunohistochemistry (Frozen) (1:10)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17057 349.00
Check your price
  • Influenza A NP Antibody (Monoclonal, I97D)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17056
  • Size 1 mL
Applications
ELISA (1:20 -1:200), Immunofluorescence (1:10 -1:50), Immunohistochemistry (1:10-1:50)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17056 349.00
Check your price
  • Influenza B NP Antibody (Monoclonal, H89B)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17058
  • Size 1 mL
Applications
Immunofluorescence (1:10)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17058 309.00
Check your price
  • Influenza B Virions Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA130853
  • Size 1 mL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Influenza Virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130853 425.00
Check your price
  • Influenza B Virions Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17225
  • Size 1 mL
Applications
Immunofluorescence (1:1000)
Alternate names:
Influenza Virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17225 309.00
Check your price
  • Influenza B Virions Antibody, Biotin
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173038
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Influenza Virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173038 309.00
Check your price
  • Influenza B Virions Antibody, FITC
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173036
  • Size 1 mL
Applications
Immunofluorescence (1:10)
Alternate names:
Influenza Virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173036 309.00
Check your price
  • Influenza B Virions Antibody, HRP
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173037
  • Size 1 mL
Applications
ELISA (1:200)
Alternate names:
Influenza Virions
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173037 309.00
Check your price
  • Inner Mitochondrial Membrane Antibody (Monoclonal, H6/C12)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA180361
  • Size 1 mL
Applications
ELISA (1/50), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1/10)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180361 349.00
Check your price
  • iNOS Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA137925
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase NOS2; NOS type II; Macrophage NOS; Inducible NO synthase; Hepatocyte NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137925 389.00
Check your price
  • ITGB5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137932
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
FLJ26658
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137932 449.00
Check your price
  • Japanese Encephalitis Virus gp E Antibody (Monoclonal, JEI)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171256
  • Size 1 mL
Applications
ELISA (5-10X), Immunofluorescence (5-10X)
Alternate names:
JEV, Flavivirus, Japanese B Encephalitis Virus E-glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171256 375.00
Check your price
  • JM4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137961
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
PRAF2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137961 449.00
Check your price
  • Ketoprofen Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175228
  • Size 1 mL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175228 700.00
Check your price