Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  44  45  46  47  48  49  50  51  52  53  54  55  56  57  58  59  60  61  62  63  64  65  66  67  68  69  70  71  72  73  80  90  100  110  120  130  140  150  200  300  400  500  600  700  800  900  1000  1630 
1741 - 1770 of 48881 products displayed
  • ATG4B Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523188
  • Size 100ug
Applications
Western Blot (2-4 µg/ml)
Alternate names:
Autophagy-related protein 4 homolog B; Autophagy-related cysteine endopeptidase 1; Autophagin-1; AUT-like 1 cysteine endopeptidase; bAut2B; Autophagy-related cysteine endopeptidase 2B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523188 349.00
Check your price
  • ATG5 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA523186
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523186 349.00
Check your price
  • ATG5 Polyclonal Antibody for Western Blot and IHC
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520688
  • Size 100ug
Applications
Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Apoptosis-specific protein; APG5-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520688 349.00
Check your price
  • ATG7 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523360
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Ubiquitin-activating enzyme E1-like protein; Autophagy-related protein 7; ATG12-activating enzyme E1 ATG7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523360 349.00
Check your price
  • ATG7 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520173
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Ubiquitin-activating enzyme E1-like protein; Autophagy-related protein 7; ATG12-activating enzyme E1 ATG7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520173 349.00
Check your price
  • ATG7 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520174
  • Size 100ug
Applications
Immunocytochemistry (10 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (1-4 µg/ml)
Alternate names:
Ubiquitin-activating enzyme E1-like protein; Autophagy-related protein 7; ATG12-activating enzyme E1 ATG7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520174 349.00
Check your price
  • ATG9A Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521043
  • Size 100ug
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
APG9-like 1; mATG9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521043 349.00
Check your price
  • ATG9B Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520998
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Nitric oxide synthase 3-overlapping antisense gene protein; APG9-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520998 349.00
Check your price
  • ATGL Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518902
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Transport-secretion protein 2; TTS2.2; Pigment epithelium-derived factor; IPLA2-zeta; Desnutrin; Calcium-independent phospholipase A2; Adipose triglyceride lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518902 349.00
Check your price
  • ATGL Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518661
  • Size 100ug
Applications
Western Blot (0.2-0.6 ug/ml)
Alternate names:
Transport-secretion protein 2; TTS2.2; Pigment epithelium-derived factor; IPLA2-zeta; Desnutrin; Calcium-independent phospholipase A2; Adipose triglyceride lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518661 349.00
Check your price
  • ATGL Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518672
  • Size 100ug
Applications
Immunohistochemistry (2-3µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
Transport-secretion protein 2; TTS2.2; Pigment epithelium-derived factor; IPLA2-zeta; Desnutrin; Calcium-independent phospholipase A2; Adipose triglyceride lipase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518672 349.00
Check your price
  • ATOH8 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520710
  • Size 100ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Okadin; Helix-loop-helix protein mATH-6; Helix-loop-helix protein hATH-6; Class A basic helix-loop-helix protein 21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520710 349.00
Check your price
  • ATP Citrate Lyase Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA517027
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Citrate cleavage enzyme; ATP-citrate (pro-S-)-lyase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517027 309.00
Check your price
  • ATP11A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520995
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 ug/ml)
Alternate names:
P4-ATPase flippase complex alpha subunit ATP11A; ATPase class VI type 11A; ATPase IS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520995 349.00
Check your price
  • ATP11B Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520996
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 ug/ml)
Alternate names:
P4-ATPase flippase complex alpha subunit ATP11B; ATPase class VI type 11B; ATPase IR
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520996 349.00
Check your price
  • ATP13A1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518860
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
ATP13A1; ATPase type 13A1; ATP13A; CGI-152; DKFZp761L1623; FLJ31858; FLJ41786; FLJ43873; FLJ90317; KIAA1825; cation transporting ATPase; cation-transporting ATPase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518860 349.00
Check your price
  • ATP1B1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519199
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Sodium/potassium-dependent ATPase subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519199 349.00
Check your price
  • ATP2C1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521126
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
ATP-dependent Ca(2+) pump PMR1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521126 349.00
Check your price
  • ATP2C2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521127
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Secretory pathway Ca(2+)-ATPase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521127 349.00
Check your price
  • ATP6IP2 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518205
  • Size 100ug
Applications
Immunohistochemistry (0.1-1.0ug/ml), Western Blot (0.25-1 ug/ml)
Alternate names:
Renin/prorenin receptor; ATPase H(+)-transporting lysosomal-interacting protein 2; ATPase H(+)-transporting lysosomal accessory protein 2; Vacuolar ATP synthase membrane sector-associated protein M8-9; N14F; Embryonic liver differentiation factor 10; ER-localized type I transmembrane adaptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518205 349.00
Check your price
  • ATR Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520017
  • Size 100ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (10 ug/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
FRAP-related protein 1; Ataxia telangiectasia and Rad3-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520017 349.00
Check your price
  • Aurora A Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132191
  • Size 100ug
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase 15; Breast tumor-amplified kinase; Aurora/IPL1-related kinase 1; Aurora 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132191 399.00
Check your price
  • Aurora A Polyclonal Antibody for Western Blot, IP and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19013
  • Size 100ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
Serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase 15; Breast tumor-amplified kinase; Aurora/IPL1-related kinase 1; Aurora 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19013 309.00
Check your price
  • Aurora C Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519256
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Serine/threonine-protein kinase aurora-C; Serine/threonine-protein kinase 13; Aurora/IPL1/Eg2 protein 2; Aurora/IPL1-related kinase 3; Aurora 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519256 349.00
Check your price
  • AVEN Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520287
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
PDCD12; cell death regulator Aven; programmed cell death 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520287 349.00
Check your price
  • AVEN Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520288
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
PDCD12; cell death regulator Aven; programmed cell death 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520288 349.00
Check your price
  • Avian Influenza HA Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520099
  • Size 100ug
Applications
ELISA (1 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Influenza A Hemagglutinin, Avian Hemagglutinin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520099 349.00
Check your price
  • Avian Influenza HA Type 2 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520293
  • Size 100ug
Applications
ELISA (1 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Influenza A Hemagglutinin, Avian Hemagglutinin 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520293 349.00
Check your price
  • Avian Influenza HA Type 3 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA520294
  • Size 100ug
Applications
ELISA (1 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Influenza A Hemagglutinin, Avian Hemagglutinin 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520294 349.00
Check your price
  • AXIN1 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518024
  • Size 100ug
Applications
Immunohistochemistry (3-5µg/ml)
Alternate names:
Axis inhibition protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518024 349.00
Check your price