Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  61  62  63  64  65  66  67  68  69  70  71  72  73  74  75  76  77  78  79  80  81  82  83  84  85  86  87  88  89  90  100  110  120  130  140  150  160  170  200  300  400  500  600  700  800  900  1000  1605 
2251 - 2280 of 48136 products displayed
  • Phospho-eIF4E (Ser209) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # 44528G
  • Size 10 blots
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (2-3 ug/ml), Immunofluorescence (2-3 ug/ml), Western Blot (Assay Dependent)
Alternate names:
mRNA cap-binding protein; eIF-4F 25 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
44528G 309.00
Check your price
  • Phospho-ErbB2 (Tyr1248) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44904G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (1:500-1:2000)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44904G 304.00
Check your price
  • Phospho-ERK1/ERK2 (Thr185, Tyr187) Antibody
  • Host Rabbit
  • Target Species Bovine, C. elegans, Chicken, Fruit fly, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 44680G
  • Size 10 blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Myelin xP42 protein kinase; Myelin basic protein kinase; M phase MAP kinase; p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44680G 306.00
Check your price
  • Phospho-ERK5 (Tyr218, Tyr220) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44612G
  • Size 10 blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Extracellular signal-regulated kinase 5; Big MAP kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44612G 304.00
Check your price
  • Phospho-ETS1 (Ser251) Antibody
  • Host Rabbit
  • Target Species Chicken, Rat
  • Conjugate Unconjugated
  • Catalog # 441107G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p54
Catalog # Size Price (AUD) Your Price (AUD) Qty
441107G 309.00
Check your price
  • Phospho-ETS1 (Ser282, Ser285) Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 441111G
  • Size 10 blots
Applications
ChIP assay (5 µg), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
p54
Catalog # Size Price (AUD) Your Price (AUD) Qty
441111G 304.00
Check your price
  • Phospho-ETS1 (Ser282) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 441109G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p54
Catalog # Size Price (AUD) Your Price (AUD) Qty
441109G 309.00
Check your price
  • Phospho-ETS1 (Thr38) Antibody
  • Host Rabbit
  • Target Species Chicken, Mouse, Rabbit, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 441104G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p54
Catalog # Size Price (AUD) Your Price (AUD) Qty
441104G 304.00
Check your price
  • Phospho-ETS2 (Thr72) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441105G
  • Size 10 blots
Applications
ChIP assay (1-5 µg), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:500-1:2000 )
Alternate names:
ETS2IT1, oncogene ETS-2, protein C-ets-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
441105G 305.00
Check your price
  • Phospho-FAK (Ser722) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44588G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Protein phosphatase 1 regulatory subunit 71; Focal adhesion kinase-related nonkinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44588G 309.00
Check your price
  • Phospho-FAK (Ser732) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44590G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44590G 304.00
Check your price
  • Phospho-FAK (Ser910) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44596G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Protein phosphatase 1 regulatory subunit 71; Focal adhesion kinase-related nonkinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44596G 304.00
Check your price
  • Phospho-FAK (Tyr397) Antibody
  • Host Rabbit
  • Target Species Chicken, Fruit fly, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # 44624G
  • Size 10 blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44624G 309.00
Check your price
  • Phospho-FAK (Tyr397) Antibody (Monoclonal, 141-9)
  • Host Rabbit
  • Target Species Chicken, Fruit fly, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 44625G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44625G 305.00
Check your price
  • Phospho-FAK (Tyr407) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44650G
  • Size 10 blots
Applications
Flow Cytometry (1:100-1:500), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44650G 309.00
Check your price
  • Phospho-FAK (Tyr576) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 44652G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44652G 304.00
Check your price
  • Phospho-FAK (Tyr577) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # 44614G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44614G 309.00
Check your price
  • Phospho-FAK (Tyr861) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44626G
  • Size 10 blots
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
pp125FAK; p125FAK; Protein-tyrosine kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
44626G 306.00
Check your price
  • Phospho-FAK2 (Tyr402) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44618G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44618G 306.00
Check your price
  • Phospho-FAK2 (Tyr402) Antibody, Alexa Fluor® 488
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Alexa Fluor® 488
  • Catalog # 44618A1
  • Size 10 blots
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44618A1 309.00
Check your price
  • Phospho-FAK2 (Tyr579, Tyr580) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44636G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44636G 309.00
Check your price
  • Phospho-FAK2 (Tyr579) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44632G
  • Size 10 blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44632G 304.00
Check your price
  • Phospho-FAK2 (Tyr580) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44634G
  • Size 10 blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44634G 303.00
Check your price
  • Phospho-FAK2 (Tyr881) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44620G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (Assay Dependent)
Alternate names:
Related adhesion focal tyrosine kinase; Proline-rich tyrosine kinase 2; Focal adhesion kinase 2; Cell adhesion kinase beta; Calcium-regulated non-receptor proline-rich tyrosine kinase; Calcium-dependent tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
44620G 309.00
Check your price
  • Phospho-FGFR1 (Tyr653, Tyr654) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441140G
  • Size 10 blots
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Proto-oncogene c-Fgr; N-sam; Fms-like tyrosine kinase 2; Basic fibroblast growth factor receptor 1; MFR
Catalog # Size Price (AUD) Your Price (AUD) Qty
441140G 305.00
Check your price
  • Phospho-Filamin 2 (Ser2113) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441120G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Gamma-filamin; Filamin-2; Actin-binding-like protein; ABP-L; ABP-280-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
441120G 309.00
Check your price
  • Phospho-FLT3 (Tyr955) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441130G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Tyrosine-protein kinase receptor flk-2; Fms-like tyrosine kinase 3; Fetal liver kinase 2; FL cytokine receptor; Stem cell tyrosine kinase 1; Fetal liver kinase-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
441130G 309.00
Check your price
  • Phospho-GAB1 (Tyr627) Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 44568G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Growth factor receptor bound protein 2-associated protein 1; GRB2-associated binder 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44568G 309.00
Check your price
  • Phospho-GATA4 (Ser105) Antibody
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 44948
  • Size 10 blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-1.0 ug/ml)
Alternate names:
GATA-binding factor 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
44948 309.00
Check your price
  • Phospho-GSK3 beta (Tyr216, Tyr279) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44604G
  • Size 10 blots
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Serine/threonine-protein kinase GSK3B; Factor A
Catalog # Size Price (AUD) Your Price (AUD) Qty
44604G 304.00
Check your price