Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  64  65  66  67  68  69  70  71  72  73  74  75  76  77  78  79  80  81  82  83  84  85  86  87  88  89  90  91  92  93  100  110  120  130  140  150  160  170  200  300  400  500  600  700  800  900  1000  1605 
2341 - 2370 of 48123 products displayed
  • Phospho-PKC delta (Ser664) Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 44976G
  • Size 10 blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
nPKC-delta; Tyrosine-protein kinase PRKCD
Catalog # Size Price (AUD) Your Price (AUD) Qty
44976G 303.00
Check your price
  • Phospho-PKC epsilon (Ser729) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44977G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
nPKC-epsilon
Catalog # Size Price (AUD) Your Price (AUD) Qty
44977G 306.00
Check your price
  • Phospho-PKC eta (Thr655) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44969
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (0.1-0.75 ug/ml)
Alternate names:
nPKC-eta; PKC-L
Catalog # Size Price (AUD) Your Price (AUD) Qty
44969 309.00
Check your price
  • Phospho-PKC gamma (Thr514) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44956
  • Size 10 blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (0.1-1 µg/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44956 309.00
Check your price
  • Phospho-PKC gamma (Thr655) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44965
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (0.1-1.0 ug/ml)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44965 309.00
Check your price
  • Phospho-PKC gamma (Thr674) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44975G
  • Size 10 blots
Applications
Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta, nPKC-delta, PRKCD, Sphingosine-dependent protein kinase-1, SDK-1, Tyrosine-protein kinase PRKCD, Protein kinase C delta type, Protein kinase C epsilon type, nPKC-epsilon, PRKCE, PKCE, Protein kinase C gamma type, PKC-gamma, PRKCG, PKCG, Protein kinase C eta type, PKC-L, nPKC-eta, PRKCH, PKCL, PRKCL, Protein kinase C iota type, PRKCI, DXS1179E, Protein kinase C theta type, nPKC-theta, PRKCQ, PRKCT, Protein kinase C zeta type, nPKC-zeta, PRKCZ, PKC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44975G 309.00
Check your price
  • Phospho-PKC lambda/iota (Thr555, Thr563) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44968G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (Assay Dependent)
Alternate names:
nPKC-iota; Atypical protein kinase C-lambda/iota
Catalog # Size Price (AUD) Your Price (AUD) Qty
44968G 304.00
Check your price
  • Phospho-PKC mu (Ser742) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44961G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
nPKC-mu; nPKC-D1; Protein kinase D; Protein kinase C mu type
Catalog # Size Price (AUD) Your Price (AUD) Qty
44961G 309.00
Check your price
  • Phospho-PKR (Thr451) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44668G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
p68 kinase; Tyrosine-protein kinase EIF2AK2; Serine/threonine-protein kinase TIK; Protein kinase RNA-activated; P1/eIF-2A protein kinase; Interferon-inducible RNA-dependent protein kinase; Eukaryotic translation initiation factor 2-alpha kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
44668G 305.00
Check your price
  • Phospho-PLCG1 (Tyr783) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44696G
  • Size 10 blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Phospholipase C-gamma-1; Phosphoinositide phospholipase C-gamma-1; Phospholipase C-II; PLC-148
Catalog # Size Price (AUD) Your Price (AUD) Qty
44696G 304.00
Check your price
  • Phospho-PPP1R2 (Thr72) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 441160G
  • Size 10 blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein phosphatase inhibitor 2, IPP-2, PPP1R2, IPP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
441160G 309.00
Check your price
  • Phospho-PRAS40 (Thr246) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441100G
  • Size 10 blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
40 kDa proline-rich AKT substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
441100G 304.00
Check your price
  • Phospho-PTEN (Ser370) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 441060G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:1000)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441060G 309.00
Check your price
  • Phospho-PTEN (Ser380, Thr382, Ser385) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441066G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441066G 309.00
Check your price
  • Phospho-PTEN (Ser385) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 441064G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441064G 309.00
Check your price
  • Phospho-PTEN (Thr383) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 441062G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay Dependent)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441062G 309.00
Check your price
  • Phospho-RAC1/CDC42 (Ser71) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44214G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
p21-Rac1; Ras-like protein TC25; Cell migration-inducing gene 5 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44214G 303.00
Check your price
  • Phospho-RANBP3 (Ser58) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 441180G
  • Size 10 blots
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ran Binding Protein 3, RANB3
Catalog # Size Price (AUD) Your Price (AUD) Qty
441180G 309.00
Check your price
  • Phospho-Rb (Ser249, Thr252) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44584G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Western Blot (1:500-1:2000)
Alternate names:
pp110; pRb; p105-Rb
Catalog # Size Price (AUD) Your Price (AUD) Qty
44584G 309.00
Check your price
  • Phospho-Rb (Thr356) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44578
  • Size 10 blots
Applications
Western Blot (0.1-1.0 ug/ml)
Alternate names:
pp110; pRb; p105-Rb
Catalog # Size Price (AUD) Your Price (AUD) Qty
44578 309.00
Check your price
  • Phospho-Rb (Thr821) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44582G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
pp110; pRb; p105-Rb
Catalog # Size Price (AUD) Your Price (AUD) Qty
44582G 309.00
Check your price
  • Phospho-Rb (Thr826) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44576G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
pp110; pRb; p105-Rb
Catalog # Size Price (AUD) Your Price (AUD) Qty
44576G 309.00
Check your price
  • Phospho-RET (Tyr1016) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44277G
  • Size 10 blots
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Proto-oncogene c-Ret; Cadherin family member 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
44277G 309.00
Check your price
  • Phospho-RSK1 (Ser221) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44924G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44924G 309.00
Check your price
  • Phospho-RSK1 (Ser380) Antibody
  • Host Rabbit
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 44928G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Ribosomal S6 kinase 1; MAP kinase-activated protein kinase 1a; 90 kDa ribosomal protein S6 kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44928G 304.00
Check your price
  • Phospho-S6 (Ser235, Ser236) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44922G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Western Blot (0.5-2 µg/ml)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
44922G 309.00
Check your price
  • Phospho-S6 (Ser236) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44921G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
44921G 309.00
Check your price
  • Phospho-S6 (Ser244, Ser247) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44923G
  • Size 10 blots
Applications
Western Blot (Assay Dependent)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
44923G 309.00
Check your price
  • Phospho-SGK1 (Ser422) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 441264G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (Assay Dependent)
Alternate names:
Serum/glucocorticoid-regulated kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441264G 309.00
Check your price
  • Phospho-SGK1 (Thr256) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 441260G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:500-1:2000 )
Alternate names:
Serum/glucocorticoid-regulated kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
441260G 305.00
Check your price