Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  73  74  75  76  77  78  79  80  81  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  110  120  130  140  150  160  170  180  200  300  400  500  600  700  800  900  1000  1630 
2611 - 2640 of 48877 products displayed
  • CYP17A1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518504
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (3-5 ug/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Steroid 17-alpha-monooxygenase; Cytochrome P450-C17; Cytochrome P450 17A1; CYPXVII; 17-alpha-hydroxyprogesterone aldolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518504 349.00
Check your price
  • CYP1A1 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517063
  • Size 100ug
Applications
Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Cytochrome P450-P1; Cytochrome P450-C; Cytochrome P450 form 6; CYPIA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517063 309.00
Check your price
  • CYP1B1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523139
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
CYPIB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523139 349.00
Check your price
  • CYP1B1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523167
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
CYPIB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523167 349.00
Check your price
  • CYP24A1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519406
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Cytochrome P450-CC24; Cytochrome P450 24A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519406 349.00
Check your price
  • CYP2E1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519381
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (3-5 ug/ml), Western Blot (0.05-0.2 ug/ml)
Alternate names:
Cytochrome P450-J; CYPIIE1; 4-nitrophenol 2-hydroxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519381 349.00
Check your price
  • CYP3A4 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517064
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Taurochenodeoxycholate 6-alpha-hydroxylase; Quinine 3-monooxygenase; Nifedipine oxidase; Cytochrome P450-PCN1; Cytochrome P450 NF-25; Cytochrome P450 HLp; Cytochrome P450 3A3; Cholesterol 25-hydroxylase; CYPIIIA4; CYPIIIA3; Albendazole sulfoxidase; Albendazole monooxygenase; 1,8-cineole 2-exo-monooxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517064 309.00
Check your price
  • CYP7B1 Polyclonal Antibody for IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518224
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (3-6 µg/ml)
Alternate names:
Oxysterol 7-alpha-hydroxylase; Cytochrome P450 7B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518224 349.00
Check your price
  • CYPOR Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519377
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
POR; P450 (cytochrome) oxidoreductase; CPR; CYPOR; DKFZp686G04235; FLJ26468; P450R; NADPH-dependent cytochrome P450 reductase; cytochrome P450 reductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519377 349.00
Check your price
  • Cypridina luciferase Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # PA1182
  • Size 100ug
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Cypridina luciferase, Cypridina noctiluca luciferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1182 309.00
Check your price
  • Cystatin 8 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519253
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Cystatin-related epididymal spermatogenic protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519253 349.00
Check your price
  • Cystatin 8 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519252
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Cystatin-related epididymal spermatogenic protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519252 349.00
Check your price
  • Cystatin C Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519108
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Post-gamma-globulin; Neuroendocrine basic polypeptide; Gamma-trace; Cystatin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519108 349.00
Check your price
  • Cystatin C Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518043
  • Size 100ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Cystatin-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518043 349.00
Check your price
  • Cystatin F Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518044
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Leukocystatin; Cystatin-like metastasis-associated protein; Cystatin-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518044 349.00
Check your price
  • CYTIP Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518019
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Pleckstrin homology Sec7 and coiled-coil domains-binding protein; Cytohesin-binding protein HE; Cytohesin-associated scaffolding protein; Cytohesin binder and regulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518019 349.00
Check your price
  • Cytochrome C Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19586
  • Size 100ug
Applications
ELISA (1:2000-1:10,000), Western Blot (1:2000-1:10,000)
Alternate names:
cyt c
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19586 375.00
Check your price
  • Cytochrome C Polyclonal Antibody for Western Blot and ELISA
  • Host Chicken
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19585
  • Size 100ug
Applications
ELISA (1:2,000-1:10,000), Western Blot (1:2,000-1:10,000)
Alternate names:
cyt c
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19585 309.00
Check your price
  • Cytohesin 2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA518833
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
SEC7 homolog B; PH, SEC7 and coiled-coil domain-containing protein 2; ARF nucleotide-binding site opener; ARF exchange factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518833 349.00
Check your price
  • Cytokeratin 13 Monoclonal Antibody for Western Blot and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517109
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Western Blot (1:500-1:2000)
Alternate names:
Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517109 309.00
Check your price
  • Cytokeratin 13 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519049
  • Size 100ug
Applications
Immunohistochemistry (2-4µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519049 349.00
Check your price
  • Cytokeratin 18 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate FITC
  • Catalog # MA110326
  • Size 100ug
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Cytokeratin endo B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110326 275.00
Check your price
  • Cytokeratin 18 Monoclonal Antibody, FITC conjugate for IF and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110327
  • Size 100ug
Applications
Flow Cytometry (1-5 ug/ml), Immunofluorescence (Assay dependent)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110327 275.00
Check your price
  • Cytokeratin 5 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517058
  • Size 100ug
Applications
Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517058 309.00
Check your price
  • Cytokeratin 5 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA517057
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517057 309.00
Check your price
  • Cytokeratin Pan Monoclonal Antibody, APC conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Mammal
  • Conjugate APC
  • Catalog # MA110325
  • Size 100ug
Applications
Flow Cytometry (5 ug/ml), Immunocytochemistry (1:100-1:400), Immunofluorescence (1:100-1:400)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Keratin-10; Cytokeratin-10; Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A; Keratin-13; Cytokeratin-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110325 349.00
Check your price
  • Cytomegalovirus gB Late Antigen Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17598
  • Size 100ug
Applications
ELISA (1:1000), Immunofluorescence (Assay Dependent), Western Blot (1:1000)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17598 309.00
Check your price
  • Cytomegalovirus gB Late Antigen Monoclonal Antibody, FITC conjugate for IF
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA516701
  • Size 100ug
Applications
Immunofluorescence (Assay Dependent)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516701 399.00
Check your price
  • Cytomegalovirus IE 72 kDa Antigen Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17596
  • Size 100ug
Applications
ELISA (Assay dependent), Immunofluorescence (Assay dependent), Western Blot (Assay dependent)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17596 275.00
Check your price
  • Cytomegalovirus pp65 Monoclonal Antibody for Western Blot, IF and ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17597
  • Size 100ug
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CMV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17597 309.00
Check your price