Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  79  80  81  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  110  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1630 
2791 - 2820 of 48882 products displayed
  • DSCAM Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518559
  • Size 100ug
Applications
Western Blot (2-6 ug/ml)
Alternate names:
CHD2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518559 349.00
Check your price
  • DTX3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA519212
  • Size 100ug
Applications
Western Blot (0.2-0.5 ug/ml)
Alternate names:
Protein deltex-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519212 349.00
Check your price
  • DUSP10 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518008
  • Size 100ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
Mitogen-activated protein kinase phosphatase 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518008 349.00
Check your price
  • DUSP16 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Non-human primate, Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523140
  • Size 100ug
Applications
ChIP assay (1-3 ul), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Mitogen-activated protein kinase phosphatase 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523140 349.00
Check your price
  • DUSP16 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517984
  • Size 100ug
Applications
Immunohistochemistry (2-4µg/ml)
Alternate names:
Mitogen-activated protein kinase phosphatase 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517984 349.00
Check your price
  • DUSP18 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523128
  • Size 100ug
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Low molecular weight dual specificity phosphatase 20
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523128 349.00
Check your price
  • DUSP8 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518007
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Dual specificity protein phosphatase hVH-5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518007 349.00
Check your price
  • DVL1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523409
  • Size 100ug
Applications
Western Blot (5-7 µg/ml)
Alternate names:
DSH homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523409 349.00
Check your price
  • DVL3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Dog, Horse, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523401
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
DSH homolog 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523401 349.00
Check your price
  • DYDC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519251
  • Size 100ug
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
DYDC1; DPY30 domain containing 1; DPY30D1; FLJ43920; bA36D19.5; BC019250 protein; OTTHUMP00000019963
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519251 349.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1142
  • Size 100ug
Applications
ELISA (100ng/ml), Western Blot (1ug/ml)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1142 159.00
Check your price
  • Dynactin 1 Polyclonal Antibody for Western Blot, ICC, IHC, IHC (P) and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518095
  • Size 100ug
Applications
Immunocytochemistry (Assay-dependent), Immunohistochemistry (2-4µg/ml), Western Blot (0.5-2 ug/ml)
Alternate names:
p150-glued; p135; DAP-150; 150 kDa dynein-associated polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518095 349.00
Check your price
  • Dynactin 3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519399
  • Size 100ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Dynactin light chain p24; Dynactin complex subunit 22 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519399 349.00
Check your price
  • Dynactin 4 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517065
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Dynactin subunit p62
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517065 309.00
Check your price
  • DYRK1A Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521124
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
RP86; Protein kinase minibrain homolog; Dual specificity YAK1-related kinase; MP86; HP86
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521124 349.00
Check your price
  • DYRK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521125
  • Size 100ug
Applications
Immunocytochemistry (10 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
dual specificity tyrosine-phosphorylation-regulated kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521125 349.00
Check your price
  • DYX1C1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518347
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5-10 ug/ml), Western Blot (0.1 ug/ml)
Alternate names:
DYX1C1; DYX1; EKN1; DYXC1; FLJ37882; dyslexia susceptibility 1 candidate 1; MGC70618; RD
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518347 349.00
Check your price
  • DYX1C1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518517
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
DYX1C1; EKN1; dyslexia susceptibility 1 candidate 1; DYX1; DYXC1; FLJ37882; MGC70618
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518517 349.00
Check your price
  • E-cadherin Monoclonal Antibody for Western Blot, ICC, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110192
  • Size 100ug
Applications
Flow Cytometry (5-10 ug/ml), Immunocytochemistry (4-8 ug/ml), Immunohistochemistry (Frozen) (4-8 ug/ml), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Uvomorulin; Epithelial cadherin; CAM 120/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110192 225.00
Check your price
  • E-Selectin/P-Selectin Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate FITC
  • Catalog # MA516428
  • Size 100ug
Applications
Flow Cytometry (Neat)
Alternate names:
Leukocyte-endothelial cell adhesion molecule 2; Endothelial leukocyte adhesion molecule 1; CD62 antigen-like family member E; Platelet activation dependent granule-external membrane protein; Leukocyte-endothelial cell adhesion molecule 3; Granule membrane protein 140; CD62 antigen-like family member P
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516428 325.00
Check your price
  • E2F1 Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA517070
  • Size 100ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
pRB-binding protein E2F-1; Retinoblastoma-binding protein 3; Retinoblastoma-associated protein 1; PBR3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517070 309.00
Check your price
  • E2F6 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519143
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
E2F6; E2F transcription factor 6; E2F-6; MGC111545; E2F transcription factor 6, isoform 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519143 349.00
Check your price
  • E2F7 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519104
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
E2F7; E2F transcription factor 7; FLJ12981
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519104 349.00
Check your price
  • EAR2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523321
  • Size 100ug
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
V-erbA-related protein 2; COUP transcription factor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523321 349.00
Check your price
  • EBF1 Polyclonal Antibody for Western Blot, IP and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518720
  • Size 100ug
Applications
Immunoprecipitation (Assay Dependent), Western Blot (0.3-0.1 ug/ml)
Alternate names:
Early B-cell factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518720 349.00
Check your price
  • EBI3 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516255
  • Size 100ug
Applications
Flow Cytometry (1-3 µg/10^6 cells), Western Blot (0.1-3 µg/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516255 349.00
Check your price
  • EBI3 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523297
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523297 349.00
Check your price
  • EBI3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523228
  • Size 100ug
Applications
Western Blot (2-6 µg/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523228 349.00
Check your price
  • EBI3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520663
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520663 349.00
Check your price
  • EBI3 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520662
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1 ug/ml)
Alternate names:
Epstein-Barr virus-induced gene 3 protein homolog; Epstein-Barr virus-induced gene 3 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520662 349.00
Check your price