Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  20  30  40  50  60  70  82  83  84  85  86  87  88  89  90  91  92  93  94  95  96  97  98  99  100  101  102  103  104  105  106  107  108  109  110  111  120  130  140  150  160  170  180  190  200  300  400  500  600  700  800  900  1000  1630 
2881 - 2910 of 48882 products displayed
  • EpCAM Monoclonal Antibody for Western Blot, ICC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110195
  • Size 100ug
Applications
Flow Cytometry (1-3 ug/ml), Immunocytochemistry (4 ug/ml), Immunohistochemistry (Frozen) (5-10 ug/ml), Immunohistochemistry (Paraffin) (5-10 ug/ml), Immunoprecipitation (1-2 ug/ml), Western Blot (1-4 ug/ml)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110195 159.00
Check your price
  • EphA1 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515271
  • Size 100ug
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Tyrosine-protein kinase receptor EPH; Erythropoietin-producing hepatoma receptor; EPH tyrosine kinase 1; EPH tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515271 349.00
Check your price
  • EphA10 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520775
  • Size 100ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
EphA10s protein; ephrin type-A receptor 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520775 349.00
Check your price
  • EphA2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523144
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Tyrosine-protein kinase receptor ECK; Epithelial cell kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523144 349.00
Check your price
  • EphB2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518586
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Tyrosine-protein kinase receptor EPH-3; Tyrosine-protein kinase TYRO5; Renal carcinoma antigen NY-REN-47; EPH-like kinase 5; EPH tyrosine kinase 3; ELK-related tyrosine kinase; Developmentally-regulated Eph-related tyrosine kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518586 349.00
Check your price
  • EphB6 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515280
  • Size 100ug
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Tyrosine-protein kinase-defective receptor EPH-6; MEP; HEP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515280 349.00
Check your price
  • Ephrin A1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519397
  • Size 100ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Immediate early response protein B61; EPH-related receptor tyrosine kinase ligand 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519397 349.00
Check your price
  • Ephrin B2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519361
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
HTK ligand; EPH-related receptor tyrosine kinase ligand 5; ELF-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519361 349.00
Check your price
  • EPM2A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA523252
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (0.25-0.50 µg/ml)
Alternate names:
Lafora PTPase; Glucan phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523252 349.00
Check your price
  • EPM2A Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518944
  • Size 100ug
Applications
Immunohistochemistry (2.5-3.8µg/ml), Western Blot (0.1-0.3 ug/ml)
Alternate names:
Lafora PTPase; Glucan phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518944 349.00
Check your price
  • EPM2A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534436
  • Size 100ug
Applications
Immunocytochemistry (25 ug/ml), Immunofluorescence (25 ug/ml), Western Blot (1-2 ug/mL)
Alternate names:
Lafora PTPase; Glucan phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534436 349.00
Check your price
  • EPS8 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518196
  • Size 100ug
Applications
Immunohistochemistry (5-10µg/ml)
Alternate names:
EPS8; epidermal growth factor receptor pathway substrate 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518196 349.00
Check your price
  • ERAB Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19002
  • Size 100ug
Applications
Western Blot (0.3-2 µg/ml)
Alternate names:
Type II HADH; Short-chain type dehydrogenase/reductase XH98G2; Short chain dehydrogenase/reductase family 5C member 1; Mitochondrial ribonuclease P protein 2; Endoplasmic reticulum-associated amyloid beta-peptide-binding protein; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 17-beta-hydroxysteroid dehydrogenase 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19002 309.00
Check your price
  • ERAP2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518948
  • Size 100ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Leukocyte-derived arginine aminopeptidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518948 349.00
Check your price
  • ErbB2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and IP
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520740
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1-2 ug/ml)
Alternate names:
p185erbB2; Tyrosine kinase-type cell surface receptor HER2; Proto-oncogene c-ErbB-2; Proto-oncogene Neu; Metastatic lymph node gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520740 349.00
Check your price
  • ErbB3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19040
  • Size 100ug
Applications
Western Blot (0.1-0.3 µg/ml)
Alternate names:
Tyrosine kinase-type cell surface receptor HER3; Proto-oncogene-like protein c-ErbB-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19040 309.00
Check your price
  • ERC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518105
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Rab6-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518105 349.00
Check your price
  • ERCC1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518837
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
ERCC1; excision repair cross-complementing rodent repair deficiency, complementation group 1 (includes overlapping antisense sequence); COFS4; UV20; excision repair cross-complementing 1; excision repair protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518837 349.00
Check your price
  • ERG Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518465
  • Size 100ug
Applications
Immunohistochemistry (10-20µg/ml)
Alternate names:
Transforming protein ERG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518465 349.00
Check your price
  • ERK1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA19041
  • Size 100ug
Applications
Western Blot (0.1-0.3 µg/ml)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19041 309.00
Check your price
  • ERK2 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA515304
  • Size 100ug
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515304 349.00
Check your price
  • ERK2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518329
  • Size 100ug
Applications
Western Blot (0.5-2 ug/ml)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518329 349.00
Check your price
  • ERLIN1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519152
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1; Protein KE04; Endoplasmic reticulum lipid raft-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519152 349.00
Check your price
  • ERLIN2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518413
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2; Endoplasmic reticulum lipid raft-associated protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518413 349.00
Check your price
  • ERP29 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519034
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Endoplasmic reticulum resident protein 31; Endoplasmic reticulum resident protein 28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519034 349.00
Check your price
  • ERP44 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518936
  • Size 100ug
Applications
Western Blot (0.2-0.6 ug/ml)
Alternate names:
Thioredoxin domain-containing protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518936 349.00
Check your price
  • ERp57 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA19078
  • Size 100ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Endoplasmic reticulum resident protein 60; Endoplasmic reticulum resident protein 57; Disulfide isomerase ER-60; 58 kDa microsomal protein; 58 kDa glucose-regulated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19078 309.00
Check your price
  • ERRF Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA534534
  • Size 100ug
Applications
Immunocytochemistry (25 ug/ml), Immunofluorescence (25 ug/ml), Immunohistochemistry (Assay Dependent), Western Blot (1 ug/mL)
Alternate names:
ERRF, Estrogen receptor-related factor, ER-related factor, RP11-5P18.4, C1orf64
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534534 349.00
Check your price
  • Erythrocytes Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517578
  • Size 100ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517578 159.00
Check your price
  • Erythrocytes Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517580
  • Size 100ug
Applications
Flow Cytometry (0.2 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517580 159.00
Check your price