Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
331 - 360 of 2000 products displayed
  • beta Tubulin Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Mouse
  • Target Species Amphibian, Bovine, Chicken, Human, Insect, Mouse, Non-human primate, Plant, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA511732
  • Size 500 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain; Tubulin beta class IIa
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511732 309.00
Check your price
  • Goat anti-Mouse IgM Heavy Chain Cross-Adsorbed Secondary Antibody, Alexa Fluor® 488 conjugate for IF, ICC and IHC
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 488
  • Catalog # A21042
  • Size 250 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21042 149.00
Check your price
  • HA Epitope Tag Monoclonal Antibody for IP
  • Host Mouse
  • Target Species Tag
  • Conjugate
  • Catalog # 26181
  • Size 2mL
Applications
Immunoprecipitation (20-100µl)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
26181 498.00
Check your price
  • Folate Receptor alpha Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523917
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Ovarian tumor-associated antigen MOv18; KB cells FBP; Folate receptor, adult; Folate receptor 1; Adult folate-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523917 275.00
Check your price
  • LST1 Monoclonal Antibody for Western Blot, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110400
  • Size 100 ug
Applications
Immunocytochemistry (Assay dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Protein B144
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110400 249.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC, IP and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 31430
  • Size 2 mL
Applications
Flow Cytometry (1:500-1:5,000), Immunocytochemistry (1:5,000-1:100,000), Immunofluorescence (1:5,000-1:100,000), Immunohistochemistry (1:5,000-1:100,000), Immunoprecipitation (1:500-1:5,000), Western Blot (1:5,000-1:200,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
31430 179.00
Check your price
  • Rabbit anti-Sheep IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Rabbit
  • Target Species Sheep
  • Conjugate HRP
  • Catalog # 818620
  • Size 1.5 mg
Applications
ELISA (1:200-1:20,000), Immunohistochemistry (1:200-1:20,000), Western Blot (1:200-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
818620 149.00
Check your price
  • Angiotensin II Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533340
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml)
Alternate names:
Serpin A8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533340 349.00
Check your price
  • GFP Tag Polyclonal Antibody for Western Blot, IF and ICC
  • Host Chicken
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # A10262
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Green Fluorescence Protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10262 332.00
Check your price
  • Tau Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # MN1000
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Immunohistochemistry (Paraffin) (1:500-1:2000), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1000 309.00
Check your price
  • Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, APC conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # A865
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A865 309.00
Check your price
  • PD-L1 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520343
  • Size 100 µg
Applications
Flow Cytometry (0.5 ug/ml), Immunocytochemistry (20 ug/ml), Immunofluorescence (20µg/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
B7 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520343 349.00
Check your price
  • PCNA Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human, Insect, Mouse, Non-human primate, Yeast
  • Conjugate Unconjugated
  • Catalog # 133900
  • Size 200 µg
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Immunohistochemistry (Paraffin) (1:50-1:500), Immunoprecipitation (2-5 ug), Western Blot (0.1-1 ug/ml)
Alternate names:
Cyclin
Catalog # Size Price (AUD) Your Price (AUD) Qty
133900 308.00
Check your price
  • Goat anti-Mouse IgG2c Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # PA129288
  • Size 500 µg
Applications
ELISA (1:10,000-1:30,000), Immunohistochemistry (Assay-Dependent), Western Blot (1:1000-1:10,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129288 232.00
Check your price
  • Phalloidin, DyLight® 350 conjugate for IF, ICC and IHC
  • Host
  • Target Species Many
  • Conjugate DyLight® 350
  • Catalog # 21830
  • Size 300 units
Applications
Immunocytochemistry (1-5 units/ml), Immunofluorescence (1-5 units/ml), Immunohistochemistry (Assay dependent)
Alternate names:
DY350-Phalloidin, F-actin toxin, Amanita phalloides
Catalog # Size Price (AUD) Your Price (AUD) Qty
21830 373.00
Check your price
  • CD32 Monoclonal Antibody for IHC (F), Flow, IP and FN
  • Host Mouse
  • Target Species Dog, Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # MA181191
  • Size 0.5 mg
Applications
Flow Cytometry (20 µg/ml), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (1:500-1:1000), Immunoprecipitation (20 µg/ml)
Alternate names:
Fc-gamma RII-c; CDw32; Fc-gamma RII-a; Fc-gamma RII-b
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181191 700.00
Check your price
  • Dengue Virus Type 1-4 Monoclonal Antibody for Western Blot, IF, ICC, ELISA and DB
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA127093
  • Size 250 µg
Applications
Dot blot (Assay Dependent), ELISA (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1 µg/ml)
Alternate names:
Dengue pan
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127093 399.00
Check your price
  • Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, Alexa Fluor® 594 conjugate for IF, ICC, IHC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 594
  • Catalog # A21135
  • Size 250 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21135 149.00
Check your price
  • SERCA1 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Mouse
  • Target Species Amphibian, Dog, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3912
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Immunofluorescence (1:500), Immunohistochemistry (1:20), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:500-1:5000)
Alternate names:
Endoplasmic reticulum class 1/2 Ca(2+) ATPase; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; Calcium pump 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3912 375.00
Check your price
  • NDRG2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA543119
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
NDRG1-related protein; N-myc downstream-regulated gene 2 protein; Antidepressant-related protein ADRG123; Protein Syld709613; Protein Ndr2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543119 309.00
Check your price
  • Laminin alpha-5 Monoclonal Antibody for IF, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183286
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Laminin-15 subunit alpha; Laminin-11 subunit alpha; Laminin-10 subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183286 650.00
Check your price
  • SLC47A2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537990
  • Size 100 µg
Applications
Western Blot (0.1-0.3 µg/ml)
Alternate names:
Solute carrier family 47 member 2; Kidney-specific H(+)/organic cation antiporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537990 309.00
Check your price
  • CD28 Monoclonal Antibody for Western Blot, ICC, IHC (F), Flow, IP and FN
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA110166
  • Size 100 ug
Applications
Flow Cytometry (2-8 ug/ml), Functional Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
TP44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110166 159.00
Check your price
  • Phospho-Tau (Thr181) Monoclonal Antibody for Western Blot, IF and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MN1050
  • Size 100 ug
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1050 550.00
Check your price
  • Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP conjugate for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate HRP
  • Catalog # 626520
  • Size 1 mL
Applications
ELISA (1:2,000-1:4,000), Immunohistochemistry (1:2,000-1:4,000), Western Blot (1:2,000-1:4,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
626520 149.00
Check your price
  • Integrin-Linked Kinase (human) for Western Blot and Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75638
  • Size 10 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75638 225.00
Check your price
  • Chondroitin Sulfate Monoclonal Antibody for IF and IHC (F)
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA183055
  • Size 100 µL
Applications
Immunofluorescence (1/100-1/400), Immunohistochemistry (Frozen) (1/100-1/400)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183055 309.00
Check your price
  • PGD Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521376
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
6PGD, 6-phosphogluconate dehydrogenase, decarboxylating
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521376 349.00
Check your price
  • alpha Tubulin Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # A11126
  • Size 50 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:1000-1:5000)
Alternate names:
Tubulin alpha-1 chain; Alpha-tubulin 1; Tubulin alpha-4 chain; Alpha-tubulin isotype M-alpha-4; Alpha-tubulin 4; Tubulin H2-alpha; Testis-specific alpha-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11126 251.00
Check your price
  • Goat anti-Human IgA Cross-Adsorbed Secondary Antibody for ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # A18789
  • Size 1 mg
Applications
ELISA (1:500-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18789 73.00
Check your price