Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
391 - 420 of 2000 products displayed
  • Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody (Monoclonal, D12H7L17)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 700031
  • Size 100 µg
Applications
ChIP assay (1 ul), ELISA (1-5 ug/ml), Flow Cytometry (0.5-5 ug x 10^6 cells), Immunocytochemistry (4-6 ug/ml), Immunofluorescence (4-6 ug/ml), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (0.1-1 ug/ml)
Alternate names:
c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; Stress-activated protein kinase 1c; JNK-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
700031 305.00
Check your price
  • NG2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517199
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
HSN tumor-specific antigen; Chondroitin sulfate proteoglycan NG2; Proteoglycan AN2; Melanoma-associated chondroitin sulfate proteoglycan; Melanoma chondroitin sulfate proteoglycan
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517199 275.00
Check your price
  • ORAI1 Antibody (Monoclonal, 6D11A11)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515777
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Transmembrane protein 142A; Protein orai-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515777 349.00
Check your price
  • Goat anti-Rat IgM Heavy Chain Cross-Adsorbed, Alexa Fluor® 488
  • Host Goat
  • Target Species Rat
  • Conjugate Alexa Fluor® 488
  • Catalog # A21212
  • Size 250 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21212 149.00
Check your price
  • Bovine Coronavirus Antibody (Monoclonal, 5A4)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA123055
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
BCV, Calf Enteritis, Enzootic Pneumonia complex, Winter Dysentery
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123055 349.00
Check your price
  • WNT11 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547362
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
WNT11, HWNT11, Protein Wnt-11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547362 349.00
Check your price
  • Mouse anti-Monkey IgG
  • Host Mouse
  • Target Species Non-human primate
  • Conjugate Unconjugated
  • Catalog # SA110654
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
SA110654 409.00
Check your price
  • c-Fos Antibody
  • Host Sheep
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSC00007W
  • Size 100 µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
G0/G1 switch regulatory protein 7; Cellular oncogene fos
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSC00007W 375.00
Check your price
  • HIV1 gp120 Antibody (Monoclonal, H81E)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA17042
  • Size 1mL
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50), Western Blot (1:10-1:50)
Alternate names:
Human immunodeficiency virus, HIV Glycoprotein 120, HIV-1 Glycoprotein 120
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17042 375.00
Check your price
  • Calprotectin Antibody (Monoclonal, MAC 387)
  • Host Mouse
  • Target Species Dog, Cat, Guinea pig, Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512213
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:1000)
Alternate names:
S100 calcium-binding protein A9; Migration inhibitory factor-related protein 14; Leukocyte L1 complex heavy chain; Calgranulin-B; S100 calcium-binding protein A8; Migration inhibitory factor-related protein 8; Calgranulin-A; Urinary stone protein band A; Leukocyte L1 complex light chain; Cystic fibrosis antigen; Calprotectin L1L subunit; Myeloid-related protein 14; Calprotectin L1H subunit; Pro-inflammatory S100 cytokine; Chemotactic cytokine CP-10; JNK-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512213 269.00
Check your price
  • Brucella abortus Antibody (Monoclonal, I86D)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA17391
  • Size 100 µg
Applications
ELISA (1:20), Western Blot (Assay Dependent)
Alternate names:
B. abortus, B. abortus Lipopolysaccharide, Brucella abortus lipopolysaccharide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17391 299.00
Check your price
  • CXCL12 Antibody (Monoclonal, 79018)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523759
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (8-25 µg/ml)
Alternate names:
Pre-B cell growth-stimulating factor; Intercrine reduced in hepatomas; C-X-C motif chemokine 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523759 349.00
Check your price
  • Dengue Virus Type 2 NS1 Antibody (Monoclonal, D2N1)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA171253
  • Size 1 mL
Applications
Immunofluorescence (5-10X), Western Blot (5-10X)
Alternate names:
NS1, Dengue NS1, NS1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA171253 375.00
Check your price
  • Human Insulin Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10908
  • Size 25 mg
Applications
Control (Assay Dependent)
Alternate names:
Insulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10908 307.00
Check your price
  • Goat anti-Rat IgM Heavy Chain, Alexa Fluor® 647
  • Host Goat
  • Target Species Rat
  • Conjugate Alexa Fluor® 647
  • Catalog # A21248
  • Size 250 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21248 149.00
Check your price
  • alpha Synuclein Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA516738
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:800)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516738 309.00
Check your price
  • beta Tubulin Loading Control Antibody (Monoclonal, BT7R)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA516308
  • Size 100 µg
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:2000-1:5000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308 225.00
Check your price
  • GAD65/GAD67 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538102
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Glutamate decarboxylase 65 kDa isoform; 65 kDa glutamic acid decarboxylase; Glutamate decarboxylase 67 kDa isoform; 67 kDa glutamic acid decarboxylase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538102 309.00
Check your price
  • GFAP Antibody
  • Host Chicken
  • Target Species Bovine, Chicken, Horse, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA110004
  • Size 100 µL
Applications
Immunocytochemistry (1:1000-1:5000), Immunofluorescence (1:1000-1:5000), Immunohistochemistry (1:1000-1:5000), Western Blot (1:50,000)
Alternate names:
GFAP, FLJ45472
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110004 375.00
Check your price
  • PAR3 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA545056
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Thrombin receptor-like 2; Coagulation factor II receptor-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545056 309.00
Check your price
  • RSV strain A2 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA123413
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
human respiratory syncytial virus, RSV, RSV A2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA123413 399.00
Check your price
  • SIK2 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA141208
  • Size 100 µg
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Serine/threonine-protein kinase SNF1-like kinase 2; Salt-inducible kinase 2; Qin-induced kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141208 309.00
Check your price
  • CD64 Antibody, PE (Monoclonal, 10.1)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # CD6404
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Fc-gamma RIA; Fc-gamma RI
Catalog # Size Price (AUD) Your Price (AUD) Qty
CD6404 263.00
Check your price
  • Mycobacterium tuberculosis Antibody, FITC
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate FITC
  • Catalog # PA173134
  • Size 1 mL
Applications
Immunohistochemistry (1:10-50)
Alternate names:
M. tuberculosis, myobacter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173134 349.00
Check your price
  • TCR gamma/delta Antibody, FITC (Monoclonal, 5A6.E9)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MHGD014
  • Size 2 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
T-cell receptor gamma, t-cell receptor delta, TCRd, TCRg, TCR delta gamma, TCR-CD3 complex
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHGD014 876.00
Check your price
  • Chlamydia trachomatis Antibody (Monoclonal, 103)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA190448
  • Size 500 µL
Applications
ELISA (Assay dependent)
Alternate names:
C. trachomatis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190448 449.00
Check your price
  • EpCAM Antibody (Monoclonal, Ber-EP4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512604
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:10)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512604 309.00
Check your price
  • Goat anti-Rabbit IgG (H+L) Cross-Adsorbed, Texas Red®-X
  • Host Goat
  • Target Species Rabbit
  • Conjugate Texas Red®-X
  • Catalog # T6391
  • Size 500 µL
Applications
Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
T6391 138.00
Check your price
  • Sheep IgG Isotype Control
  • Host Sheep
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # 31243
  • Size 10 mg
Applications
Blocking Assay (Assay Dependent), Control (Assay-Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent)
Alternate names:
Negative Control, Isotype Control, Immunoglobulin Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
31243 127.00
Check your price
  • MMP2 Antibody (Monoclonal, MMP2/8B4)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA116641
  • Size 100 µL
Applications
ELISA (0.03 µg/ml), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (2 µg/ml), Immunohistochemistry (Paraffin) (2 µg/ml), Immunoprecipitation (1-2 µg/100-500 µg total protein), Western Blot (1-2 µg/ml)
Alternate names:
TBE-1; Matrix metalloproteinase-2; Gelatinase A; 72 kDa gelatinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116641 375.00
Check your price