Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
451 - 480 of 2000 products displayed
  • Goat anti-Human IgG (H+L) Cross-Adsorbed, Alexa Fluor® 568
  • Host Goat
  • Target Species Human
  • Conjugate Alexa Fluor® 568
  • Catalog # A21090
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21090 169.00
Check your price
  • C-Peptide Antibody, Biotin (Monoclonal, C-PEP-01)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA110317
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (2-5 ug/ml)
Alternate names:
IDDM2, ILPR, IRDN, MODY10, proinsulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110317 249.00
Check your price
  • Human IgE Isotype Control
  • Host Human
  • Target Species Not Applicable
  • Conjugate Unconjugated
  • Catalog # DIAHE101
  • Size 100 µL
Applications
Control (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
DIAHE101 375.00
Check your price
  • LRP4 Antibody
  • Host Sheep
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA547795
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Multiple epidermal growth factor-like domains 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547795 349.00
Check your price
  • Human MMP9 Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75655
  • Size 1 mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75655 2
Check your price
  • Phospho-HSP27 (Ser82) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 44534G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-2 µg/ml)
Alternate names:
Stress-responsive protein 27; Heat shock 27 kDa protein; Estrogen-regulated 24 kDa protein; 28 kDa heat shock protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44534G 304.00
Check your price
  • Complex IV Antibody (Monoclonal, 20E8C12)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # A21348
  • Size 100 µL
Applications
Western Blot (1.0 ug/ml)
Alternate names:
COX4, COX4-1, COXIV, COX IV-1, cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1 mitochondrial
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21348 328.00
Check your price
  • CD53 Antibody, PE (Monoclonal, MEM-53)
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA110248
  • Size 100 Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Tetraspanin-25; Cell surface glycoprotein CD53
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110248 275.00
Check your price
  • GAPDH Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # TAB1001
  • Size 100 ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH
Catalog # Size Price (AUD) Your Price (AUD) Qty
TAB1001 249.00
Check your price
  • Phospho-(Ser/Thr) ATM + ATR Substrate Antibody (Monoclonal, G.770.10)
  • Host Rabbit
  • Target Species All
  • Conjugate Unconjugated
  • Catalog # MA514872
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514872 309.00
Check your price
  • Phospho-4E-BP1 (Ser65) Antibody (Monoclonal, T.730.9)
  • Host Rabbit
  • Target Species Chicken, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA514948
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
Phosphorylated heat- and acid-stable protein regulated by insulin 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514948 309.00
Check your price
  • PDGFRA Antibody (Oligoclonal, 7HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710169
  • Size 100 µg
Applications
ChIP assay (1 ul), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (2-5 ug/ml)
Alternate names:
Platelet-derived growth factor receptor 2; Platelet-derived growth factor alpha receptor; CD140a antigen; CD140 antigen-like family member A; Alpha-type platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710169 309.00
Check your price
  • Acetyl-Histone H2A.Z (Lys5, Lys7, Lys11) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540095
  • Size 50 µg
Applications
ChIP assay (1 µg), ChIP-sequencing (Assay dependant), Immunocytochemistry (1:500), Immunofluorescence (1:500), Peptide Array (1:20,000)
Alternate names:
H2A.ZK5ac, H2AZK7ac, H2AZK11ac, H2A.ZK5,7,11ac, H2AFZac
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540095 309.00
Check your price
  • Bcl-2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130411
  • Size 1 mL
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:200)
Alternate names:
Bcl-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130411 475.00
Check your price
  • CD138 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516918
  • Size 500 µL
Applications
Immunohistochemistry (1:25-1:50)
Alternate names:
SDC1, SYND1, SDC, CD antigen 138
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516918 309.00
Check your price
  • TRPC4 Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518987
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
TRPC4; transient receptor potential cation channel, subfamily C, member 4; HTRP4; MGC119570; MGC119571; MGC119572; MGC119573; TRP4; transient receptor potential 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518987 349.00
Check your price
  • Goat anti-Human IgA Fc, FITC
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # H14001
  • Size 2 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
H14001 213.00
Check your price
  • Lamin A Antibody (Monoclonal, 133A2)
  • Host Mouse
  • Target Species Mouse, Rat, Bovine, Dog, Human
  • Conjugate Unconjugated
  • Catalog # MA106101
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106101 309.00
Check your price
  • Mouse anti-Human Lambda Light Chain
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511590
  • Size 500 µL
Applications
ELISA (1ug/ml), Flow Cytometry (1ug/ml), Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (1ug/ml)
Alternate names:
Immunoglobulin lambda light chain, Ig lambda light chain, Ig lambda, lambda light chain, Igl
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511590 249.00
Check your price
  • Fibromodulin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526250
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Keratan sulfate proteoglycan fibromodulin; Collagen-binding 59 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526250 349.00
Check your price
  • CD4 Antibody (Monoclonal, GK1.5)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA11461MG
  • Size 1 mg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:20), Immunofluorescence (1:20), Western Blot (1:1000)
Alternate names:
T-cell surface antigen T4/Leu-3; T-cell differentiation antigen L3T4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA11461MG 900.00
Check your price
  • Human Insulin Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10935
  • Size 250 mg
Applications
Control (Assay Dependent)
Alternate names:
Insulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10935 532.00
Check your price
  • Prolactin Receptor Antibody (Monoclonal, B6.2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA511955
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
PRL-R, delta 4-SF1b truncated prolactin receptor, delta 4-delta 7/11 truncated prolactin receptor, hPRL receptor, prolactin receptor delta 7/11, secreted prolactin binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511955 349.00
Check your price
  • FGFR1 Antibody (Monoclonal, VBS-7)
  • Host Mouse
  • Target Species Bovine, Chicken, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 133100
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (2-5 ug), Western Blot (2 ug/ml)
Alternate names:
Proto-oncogene c-Fgr; MFR; Basic fibroblast growth factor receptor 1; Tyrosine kinase receptor CEK1; N-sam; Fms-like tyrosine kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
133100 304.00
Check your price
  • CXCL10 Antibody (Monoclonal, 134013)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523774
  • Size 500 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (10-40 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine B10; Interferon-gamma induced protein CRG-2; C7; 10 kDa interferon gamma-induced protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523774 375.00
Check your price
  • CD44 Antibody, APC (Monoclonal, MEM-263)
  • Host Mouse
  • Target Species Dog, Human, Pig
  • Conjugate APC
  • Catalog # MA110227
  • Size 100 Tests
Applications
Flow Cytometry (10 ul/10^6 cells)
Alternate names:
Phagocytic glycoprotein I; Phagocytic glycoprotein 1; Hyaluronate receptor; Hermes antigen; Heparan sulfate proteoglycan; HUTCH-I; GP90 lymphocyte homing/adhesion receptor; Extracellular matrix receptor III; Epican; CDw44
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110227 309.00
Check your price
  • SAP102 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA529116
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Synapse-associated protein 102; XLMR; Neuroendocrine-DLG
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529116 349.00
Check your price
  • IFN gamma Antibody, PE-Alexa Fluor® 610 (Monoclonal, XMG1.2)
  • Host Rat
  • Target Species Mouse
  • Conjugate PE-Alexa Fluor® 610
  • Catalog # RM90022
  • Size 1 mL
Applications
Flow Cytometry (0.1-0.25 µg per 1x10^6 cells)
Alternate names:
IFN-gamma, IFN-y, Interferon y, IFNG
Catalog # Size Price (AUD) Your Price (AUD) Qty
RM90022 284.00
Check your price
  • N-cadherin Antibody (Monoclonal, SP90)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516324
  • Size 500 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Neural cadherin; CDw325
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516324 499.00
Check your price
  • Cytokeratin 8 Antibody (Monoclonal, TS1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514428
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (0.5-1.0 µg/ml), Immunoprecipitation (2µg/mg protein lysate)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514428 309.00
Check your price