Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  117 
121 - 150 of 3500 products displayed
  • beta Actin Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1183
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (2 µg), Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1183 225.00
Check your price
  • HK1 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515680
  • Size 100µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1/200-1/1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Hexokinase type I; Brain form hexokinase; Hexokinase, tumor isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515680 349.00
Check your price
  • Phospho-PIK3R1 (Tyr607) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538905
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538905 309.00
Check your price
  • eNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA516887
  • Size 500µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:50-1:500)
Alternate names:
NOS type III; Endothelial NOS; EC-NOS; Constitutive NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516887 309.00
Check your price
  • CD8 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532334
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
T-lymphocyte differentiation antigen T8/Leu-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532334 375.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody for Western Blot and ELISA
  • Host Rat
  • Target Species Tag
  • Conjugate Unconjugated
  • Catalog # MA1142
  • Size 100ug
Applications
ELISA (100ng/ml), Western Blot (1ug/ml)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1142 159.00
Check your price
  • SERPINF2 Polyclonal Antibody for Western Blot, IHC (F) and IP
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547101
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Serpin F2; Alpha-2-plasmin inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547101 349.00
Check your price
  • HSV Type 1 gD Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA130233
  • Size 200µg
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:400)
Alternate names:
HSV1, Herpes Simplex I, HSV Type 1, Herpes Simplex 1, HHV, HSV, Herpes Simplex Type 1, Glycoprotein D
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130233 475.00
Check your price
  • CD5 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # A15813
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15813 129.00
Check your price
  • FXR Monoclonal Antibody for Western Blot, IHC, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 417200
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Retinoid X receptor-interacting protein 14; Nuclear receptor subfamily 1 group H member 4; Farnesol receptor HRR-1; Farnesoid X-activated receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
417200 427.00
Check your price
  • PRMT2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511114
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Histone-arginine N-methyltransferase PRMT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511114 349.00
Check your price
  • CD3e Monoclonal Antibody, FITC conjugate for Flow
  • Host Hamster
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # HM3401
  • Size 1mL
Applications
Flow Cytometry (0.1-0.25 µg per 1x10^6 cells)
Alternate names:
T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
HM3401 136.00
Check your price
  • UGGT1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA543360
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
UDP-glucose ceramide glucosyltransferase-like 1; UDP--Glc:glycoprotein glucosyltransferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543360 309.00
Check your price
  • Lamin A/C (mutant 453W) Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518063
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:32,000)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32; mutant 453W; Lamin A+C mutant
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518063 399.00
Check your price
  • VDR Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186623
  • Size 100µg
Applications
ELISA (1:16,000), Western Blot (0.3-1 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group I member 1; 1,25-dihydroxyvitamin D3 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186623 375.00
Check your price
  • SNAP91 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA125067
  • Size 100µL
Applications
Immunocytochemistry (1:10,000), Immunofluorescence (1:10,000), Immunoprecipitation (1:10,000), Western Blot (1:10,000)
Alternate names:
Clathrin coat-associated protein AP180; 91 kDa synaptosomal-associated protein; Phosphoprotein F1-20
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125067 309.00
Check your price
  • Phospho-MSK2 (Ser196) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA548109
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.5 µg/ml)
Alternate names:
Ribosomal protein kinase B; Nuclear mitogen- and stress-activated protein kinase 2; 90 kDa ribosomal protein S6 kinase 4; RSK-like protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548109 309.00
Check your price
  • Arg3.1 Polyclonal Antibody for Western Blot and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519513
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (1:300), Western Blot (1 µg/ml)
Alternate names:
Activity-regulated gene 3.1 protein homolog; ARC/ARG3.1; Activity-regulated gene 3.1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519513 349.00
Check your price
  • SAE2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141110
  • Size 100µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Ubiquitin-like modifier-activating enzyme 2; Ubiquitin-like 1-activating enzyme E1B; Anthracycline-associated resistance ARX
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141110 375.00
Check your price
  • EAR2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523321
  • Size 100ug
Applications
Western Blot (0.5-1 µg/ml)
Alternate names:
V-erbA-related protein 2; COUP transcription factor 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523321 349.00
Check your price
  • Goat anti-Rabbit IgG Secondary Antibody, WesternDot® 625 conjugate for Western Blot
  • Host Goat
  • Target Species Rabbit
  • Conjugate WesternDot® 625
  • Catalog # W10809
  • Size 25blots
Applications
Western Blot (1:500)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
W10809 183.00
Check your price
  • Rat anti-Mouse Lambda Light Chain Secondary Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MLAMBDA04
  • Size 1mL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Immunoglobulin lambda light chain, Ig lambda light chain, Ig lambda, lambda light chain, Igl
Catalog # Size Price (AUD) Your Price (AUD) Qty
MLAMBDA04 202.00
Check your price
  • NPY2R Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA141576
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
NPY-Y2 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141576 375.00
Check your price
  • Claudin 10 Polyclonal Antibody for Western Blot, IF, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 388400
  • Size 100µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (Assay Dependent)
Alternate names:
CPETRL3, OSP-L, OSP-like protein, claudin-10
Catalog # Size Price (AUD) Your Price (AUD) Qty
388400 304.00
Check your price
  • TTBK1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520686
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Brain-derived tau kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520686 349.00
Check your price
  • PTEN Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520418
  • Size 100ug
Applications
Immunocytochemistry (2 ug/ml), Immunofluorescence (2 ug/ml), Immunoprecipitation (Assay Dependent), Western Blot (1-2 µg/ml)
Alternate names:
Phosphatase and tensin homolog; Mutated in multiple advanced cancers 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520418 349.00
Check your price
  • FosB Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA141121
  • Size 100µg
Applications
Western Blot (2 µg/ml)
Alternate names:
G0/G1 switch regulatory protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA141121 309.00
Check your price
  • Fibrillarin Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527529
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Nucleolar protein 1; Histone-glutamine methyltransferase; 34 kDa nucleolar scleroderma antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527529 349.00
Check your price
  • ACE Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA514902
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Kininase II; Dipeptidyl carboxypeptidase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514902 349.00
Check your price
  • CD55 Monoclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182066
  • Size 100µg
Applications
Flow Cytometry (1:10-1:25), Immunohistochemistry (Frozen) (1:100-1:1000), Western Blot (Assay Dependent)
Alternate names:
RP11-357P18.1, CR, CROM, DAF, TC
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182066 275.00
Check your price