Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  70  80  90  100  110  117 
301 - 330 of 3500 products displayed
  • RFX2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA541423
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Regulatory factor X 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA541423 309.00
Check your price
  • AOX1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536922
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Azaheterocycle hydroxylase 1; Azaheterocycle hydroxylase; Aldehyde oxidase 1; Retinal oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536922 309.00
Check your price
  • STEP Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA1158
  • Size 100µg
Applications
Immunoprecipitation (2 µg), Western Blot (1:200-1:1000)
Alternate names:
Striatum-enriched protein-tyrosine phosphatase; Neural-specific protein-tyrosine phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1158 309.00
Check your price
  • RANKL Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA141318
  • Size 100µg
Applications
Western Blot (0.5 µg/ml)
Alternate names:
TNF-related activation-induced cytokine; Receptor activator of nuclear factor kappa-B ligand; Osteoprotegerin ligand; Osteoclast differentiation factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141318 309.00
Check your price
  • Proinsulin Monoclonal Antibody for IF, ICC, IHC (F), IHC (P), ELISA and RIA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA122710
  • Size 100µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Radioimmune Assays (Assay-dependent)
Alternate names:
ILPR, IRDN
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122710 309.00
Check your price
  • Phospho-S6 (Ser235, Ser236) Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701363
  • Size 100µg
Applications
Flow Cytometry (1-3µg x 10^6 cells), Immunocytochemistry (0.5 ug/ml), Immunofluorescence (0.5 ug/ml), Western Blot (0.5-1µg/ml)
Alternate names:
Phosphoprotein NP33
Catalog # Size Price (AUD) Your Price (AUD) Qty
701363 309.00
Check your price
  • Nuclear Matrix Protein p84 Monoclonal Antibody for Western Blot, IF, ICC, IHC, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA123261
  • Size 100µg
Applications
ChIP assay (Assay Dependent), Immunocytochemistry (0.5-2 ug/ml), Immunofluorescence (0.5-2 µg/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (1:100-1:500), Western Blot (0.3-2 µg/ml)
Alternate names:
Nuclear matrix protein p84; hTREX84
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123261 309.00
Check your price
  • MT-ND1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539276
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
NADH dehydrogenase subunit 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539276 309.00
Check your price
  • eNOS Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA516887
  • Size 500µL
Applications
Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:50-1:500)
Alternate names:
NOS type III; Endothelial NOS; EC-NOS; Constitutive NOS
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516887 309.00
Check your price
  • Zfp110 Polyclonal Antibody for IHC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSZ00002G
  • Size 500µg
Applications
Immunohistochemistry (10-50 µg/ml)
Alternate names:
Zinc finger protein 110; Neurotrophin receptor-interacting factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSZ00002G 309.00
Check your price
  • COPD Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSC00036G
  • Size 500µg
Applications
Immunohistochemistry (10-50 µg/ml), Western Blot (10-50 µg/ml)
Alternate names:
Delta-coat protein; Archain
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSC00036G 309.00
Check your price
  • GSR Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA170099
  • Size 250ug
Applications
Flow Cytometry (1 ug/1 million cells), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
polymorphonuclear leukocytes, PMN, PML
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170099 309.00
Check your price
  • Goat anti-Human IgG (Kappa light chain) Secondary Antibody, FITC conjugate for IF, IHC (F) and Flow
  • Host Goat
  • Target Species Human
  • Conjugate FITC
  • Catalog # PA174405
  • Size 1mg
Applications
Flow Cytometry (Neat-1/10), Immunofluorescence (Neat-1/10), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA174405 309.00
Check your price
  • TFB2M Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Horse, Human, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540897
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 2; Mitochondrial transcription factor B2; Mitochondrial 12S rRNA dimethylase 2; Hepatitis C virus NS5A-transactivated protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540897 309.00
Check your price
  • Phospho-ILK (Ser246) Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537609
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
p59ILK; ILK-2; ILK-1; 59 kDa serine/threonine-protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537609 309.00
Check your price
  • Phospho-Estrogen Receptor alpha (Ser118) Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515206
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 3 group A member 1; Estradiol receptor; ER-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515206 309.00
Check your price
  • PEA15 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544950
  • Size 100µl
Applications
Western Blot (1:1000)
Alternate names:
15 kDa phosphoprotein enriched in astrocytes; Phosphoprotein enriched in diabetes
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544950 309.00
Check your price
  • MKL1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535929
  • Size 100µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Myocardin-related transcription factor A; Megakaryocytic acute leukemia protein; Megakaryoblastic leukemia 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535929 309.00
Check your price
  • MAP2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # OSM00030W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Microtubule-associated protein 2, MAP 2, MAP-2, MAP2R
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSM00030W 309.00
Check your price
  • KLF2 Polyclonal Antibody for Western Blot, IHC and ChIP
  • Host Rabbit
  • Target Species Guinea pig, Human, Mouse, Pig, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540591
  • Size 100µl
Applications
ChIP assay (Assay-Dependent), Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Lung krueppel-like factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540591 309.00
Check your price
  • GABRA3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA546829
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
GABA(A) receptor subunit alpha-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546829 309.00
Check your price
  • CLN6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536442
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Ceroid-lipofuscinosis neuronal protein 6, Protein CLN6, CLN6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536442 309.00
Check your price
  • Phospho-TERT (Ser824) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538820
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Telomerase-associated protein 2; Telomerase catalytic subunit; HEST2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538820 309.00
Check your price
  • PBK Monoclonal Antibody for IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 730013
  • Size 100µg
Applications
Immunoprecipitation (2 ug)
Alternate names:
T-LAK cell-originated protein kinase; Spermatogenesis-related protein kinase; PDZ-binding kinase; Nori-3; MAPKK-like protein kinase; Cancer/testis antigen 84
Catalog # Size Price (AUD) Your Price (AUD) Qty
730013 309.00
Check your price
  • NFkB p50 Oligoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ChIP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710450
  • Size 100µg
Applications
ChIP assay (1-5 µg x 10^6 cells), Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1-2 ug/ml), Immunofluorescence (1-2 ug/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (2-3 ug/ml)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1; NF-kappa-B1 p84/NF-kappa-B1 p98; EBP-1; DNA-binding factor KBF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710450 309.00
Check your price
  • IFI16 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539180
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Interferon-inducible myeloid differentiation transcriptional activator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539180 309.00
Check your price
  • CD14 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA119514
  • Size 100µg
Applications
Flow Cytometry (1:1000)
Alternate names:
Myeloid cell-specific leucine-rich glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119514 309.00
Check your price
  • alpha Tubulin Monoclonal Antibody for Western Blot, ICC, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA119209
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Paraffin) (10 µg/ml), Immunoprecipitation (Assay Dependent), Western Blot (1-2 µg/ml, 60 min)
Alternate names:
Tubulin alpha-4 chain; Alpha-tubulin 4; Tubulin alpha-1 chain; Alpha-tubulin 1; Alpha-tubulin isotype M-alpha-4; Tubulin H2-alpha; Testis-specific alpha-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119209 309.00
Check your price
  • CD45 Monoclonal Antibody, PE-Cyanine5 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE-Cyanine5
  • Catalog # MA112467
  • Size 100Tests
Applications
Flow Cytometry (5 µl/test)
Alternate names:
T200; Leukocyte common antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112467 309.00
Check your price
  • Ubiquitin Monoclonal Antibody for Western Blot, IF, IHC, IP and ELISA
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # 131600
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (2-5 ug/ml), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1-3 ug/ml)
Alternate names:
Ubiquitin-like product Chap1/Dsk2; Protein linking IAP with cytoskeleton 2; DSK2 homolog; Chap1
Catalog # Size Price (AUD) Your Price (AUD) Qty
131600 306.00
Check your price