Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
361 - 390 of 2000 products displayed
  • CACNA2D1 Antibody (Monoclonal, 20A)
  • Host Mouse
  • Target Species Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3921
  • Size 100 µL
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (1:250), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:500), Western Blot (1:500)
Alternate names:
Voltage-gated calcium channel subunit alpha-2/delta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3921 375.00
Check your price
  • MIP-1 delta Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546965
  • Size 100 µg
Applications
Neutralization (0.5-2.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A15; NCC-3; Mrp-2b; Macrophage inflammatory protein 5; MIP-1 delta; Leukotactin-1; Chemokine CC-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546965 375.00
Check your price
  • IL-26 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546973
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Protein AK155
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546973 375.00
Check your price
  • CD1d Antibody (Monoclonal, NOR3.2 (NOR3.2/13.17))
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA120223
  • Size 100 µg
Applications
Flow Cytometry (Assay-Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (Assay-dependent)
Alternate names:
R3G1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA120223 375.00
Check your price
  • CD3/CD25 Antibody, FITC, PE (Monoclonal, M2AB, ITYV)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC , PE
  • Catalog # MA112091
  • Size 100 Tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; p55; TAC antigen; Growth arrest-specific protein 3; T-cell surface antigen T3/Leu-4 epsilon chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112091 375.00
Check your price
  • Streptococcus pneumoniae Antibody, HRP
  • Host Rabbit
  • Target Species Bacteria
  • Conjugate HRP
  • Catalog # PA173012
  • Size 1 mL
Applications
ELISA (1:200-1:1,000), Immunohistochemistry (1:20-200), Western Blot (1:20-200)
Alternate names:
S. pneumoniae, Pneumococcus
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173012 375.00
Check your price
  • HA Tag Antibody, Agarose (Monoclonal, 12CA5)
  • Host Mouse
  • Target Species Tag
  • Conjugate Agarose
  • Catalog # MA112455
  • Size 1 mL
Applications
Immunoprecipitation (Assay Dependent)
Alternate names:
human influenza hemagglutinin, HA-tag, HA, HA Epitope Tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112455 375.00
Check your price
  • Caspase 10 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532293
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
ICE-like apoptotic protease 4; FAS-associated death domain protein interleukin-1B-converting enzyme 2; Apoptotic protease Mch-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532293 375.00
Check your price
  • Chymotrypsin Antibody (Monoclonal, CHY 10-104.5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110623
  • Size 500 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Caldecrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110623 375.00
Check your price
  • Phospho-beta Catenin (Ser33, Ser37) Antibody (Monoclonal, BC-22)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA125084
  • Size 50 µL
Applications
ELISA (5-10 µg/ml), Western Blot (5-10 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125084 375.00
Check your price
  • LASP1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131450
  • Size 50 µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Metastatic lymph node gene 50 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131450 375.00
Check your price
  • Phalloidin, DyLight® 350
  • Host
  • Target Species Many
  • Conjugate DyLight® 350
  • Catalog # 21830
  • Size 300 units
Applications
Immunocytochemistry (1-5 units/ml), Immunofluorescence (1-5 units/ml), Immunohistochemistry (Assay dependent)
Alternate names:
DY350-Phalloidin, F-actin toxin, Amanita phalloides
Catalog # Size Price (AUD) Your Price (AUD) Qty
21830 373.00
Check your price
  • GM-CSF Antibody, Biotin (Monoclonal, 31G6)
  • Host Rat
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # MM500DB
  • Size 250 ug
Applications
Western Blot (Assay Dependent)
Alternate names:
Colony-stimulating factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MM500DB 375.00
Check your price
  • ITGA2 Antibody (Monoclonal, AK7)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516571
  • Size 200 ug
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (1:500-1:1000)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516571 375.00
Check your price
  • Tomato Spotted Wilt Virus Antibody
  • Host Chicken
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA120368
  • Size 125 µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120368 375.00
Check your price
  • TRPM2 Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522883
  • Size 100 µL
Applications
Western Blot (2 µg/ml)
Alternate names:
EREG1, KNP3, LTRPC2, NUDT9H, NUDT9L1, TRPC7, LTrpC-2, estrogen-responsive element-associated gene 1 protein, long transient receptor potential channel 2, transient receptor potential cation channel subfamily M member 2, transient receptor potential channel 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522883 375.00
Check your price
  • rRNA Antibody, DyLight® 550 (Monoclonal, Y10b)
  • Host Mouse
  • Target Species All
  • Conjugate DyLight® 550
  • Catalog # MA516064
  • Size 100 µL
Applications
Immunocytochemistry (1:2500), Immunofluorescence (1:2500), Immunoprecipitation (10 µl/lysate)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516064 375.00
Check your price
  • Phospho-NHE3 (Ser552) Antibody, DyLight® 650 (Monoclonal, 14D5)
  • Host Mouse
  • Target Species Human, Marsupial, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516093
  • Size 100 µL
Applications
ELISA (1:100-1:2000), Immunocytochemistry (1:50-1:1000), Immunofluorescence (1:50-1:1000), Immunohistochemistry (1:50-1:1000), Immunoprecipitation (1:10-1:500), Western Blot (1-5 µg/ml)
Alternate names:
Solute carrier family 9 member 3; Na(+)/H(+) exchanger 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516093 375.00
Check your price
  • NFATC1 Antibody (Monoclonal, 7A6)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA3024
  • Size 100 µL
Applications
ChIP assay (Assay dependent), Gel Shift (Assay dependent), Immunocytochemistry (1:250), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:20-200), Immunohistochemistry (Paraffin) (Assay Dependent), Immunohistochemistry (PFA fixed) (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blo
Alternate names:
NFAT transcription complex cytosolic component
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3024 375.00
Check your price
  • mDIC Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA3840
  • Size 100 µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Western Blot (1:500)
Alternate names:
Solute carrier family 25 member 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3840 375.00
Check your price
  • LC3A/LC3B Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA116931
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:300), Immunofluorescence (1:100-1:300), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (20 µg/500 µg of lysate), Western Blot (2 µg/ml)
Alternate names:
Microtubule-associated protein 1 light chain 3 alpha; MAP1A/MAP1B light chain 3 A; MAP1 light chain 3-like protein 1; Autophagy-related ubiquitin-like modifier LC3 A; Autophagy-related protein LC3 A; Microtubule-associated protein 1 light chain 3 beta; MAP1A/MAP1B light chain 3 B; MAP1 light chain 3-like protein 2; Autophagy-related ubiquitin-like modifier LC3 B; Autophagy-related protein LC3 B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116931 375.00
Check your price
  • Laminin Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # PA522900
  • Size 100 µL
Applications
Immunohistochemistry (Frozen) (1:500-1:2000), Immunohistochemistry (Paraffin) (1:500-1:2000), Western Blot (1:100-1:5000)
Alternate names:
S-laminin subunit alpha; Laminin-3 subunit alpha; Laminin-1 subunit alpha; Laminin A chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522900 375.00
Check your price
  • HIF3A Antibody, HRP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # PA522750
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Neonatal and embryonic PAS protein; Member of PAS protein 7; Inhibitory PAS domain protein; HIF3-alpha-1; Basic-helix-loop-helix-PAS protein MOP7; PAS domain-containing protein 7; Class E basic helix-loop-helix protein 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522750 375.00
Check your price
  • HIF1A Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Goat, Hamster, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522778
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (1:500-1:2000)
Alternate names:
ARNT-interacting protein; PAS domain-containing protein 8; Member of PAS protein 1; Class E basic helix-loop-helix protein 78; Basic-helix-loop-helix-PAS protein MOP1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522778 375.00
Check your price
  • CYR61 Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Rabbit
  • Conjugate Biotin
  • Catalog # PA522762
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:200)
Alternate names:
Protein GIG1; Insulin-like growth factor-binding protein 10; Cysteine-rich angiogenic inducer 61; CCN family member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522762 375.00
Check your price
  • ChREBP Antibody, Biotin
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Biotin
  • Catalog # PA522926
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500)
Alternate names:
Williams-Beuren syndrome chromosomal region 14 protein homolog; MLX-interacting protein-like; MLX interactor; Williams-Beuren syndrome chromosomal region 14 protein; WS basic-helix-loop-helix leucine zipper protein; Class D basic helix-loop-helix protein 14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522926 375.00
Check your price
  • CHOP Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA124697
  • Size 100 µL
Applications
Western Blot (1:250)
Alternate names:
Growth arrest and DNA-damage-inducible protein GADD153; CCAAT/enhancer-binding protein homologous protein; C/EBP-homologous protein 10; C/EBP-homologous protein; C/EBP zeta; Growth arrest and DNA damage-inducible protein GADD153
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA124697 375.00
Check your price
  • 53BP1 Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # PA522791
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
53BP1, p202, p53-binding protein 1, p53BP1, tumor protein 53-binding protein, 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522791 375.00
Check your price
  • ULBP1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547131
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
UL16-binding protein 1; Retinoic acid early transcript 1I; ALCAN-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547131 375.00
Check your price
  • RSV strain B1 Antibody
  • Host Rabbit
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA14182
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
human respiratory syncytial virus, RSV, RSV B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14182 375.00
Check your price