Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  40  50  60  70  80  90  100  110  117 
541 - 570 of 3500 products displayed
  • CXCL1 Monoclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523707
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (3-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Neutrophil-activating protein 3; Melanoma growth stimulatory activity; GRO-alpha(1-73); C-X-C motif chemokine 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523707 375.00
Check your price
  • Chymotrypsin Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110623
  • Size 500µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Caldecrin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110623 375.00
Check your price
  • CA125 Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA124603
  • Size 500µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ovarian carcinoma antigen CA125; Ovarian cancer-related tumor marker CA125
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124603 375.00
Check your price
  • Cyclophilin A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Hamster, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1025
  • Size 400µL
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Western Blot (1:300)
Alternate names:
Rotamase A; Cyclosporin A-binding protein; Cyclophilin A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1025 375.00
Check your price
  • LAMP2 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517868
  • Size 300ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Lysosomal membrane glycoprotein type B; CD107 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517868 375.00
Check your price
  • Erythrocytes Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517824
  • Size 300ug
Applications
Flow Cytometry (0.5 ug/10^6 cells)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517824 375.00
Check your price
  • CD4 Monoclonal Antibody for Flow and IP
  • Host Mouse
  • Target Species Horse
  • Conjugate Unconjugated
  • Catalog # MA181196
  • Size 2mL
Applications
Flow Cytometry (Neat), Immunoprecipitation (Assay dependent)
Alternate names:
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181196 375.00
Check your price
  • GPR78 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132777
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132777 375.00
Check your price
  • GPR133 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132849
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
G-protein coupled receptor PGR25; G-protein coupled receptor 133
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132849 375.00
Check your price
  • GCNF Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132593
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Retinoid receptor-related testis-specific receptor; Germ cell nuclear factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132593 375.00
Check your price
  • P-Glycoprotein Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ELISA
  • Host Mouse
  • Target Species Human, Mammal
  • Conjugate Unconjugated
  • Catalog # MA126530
  • Size 250µL
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin) (1:40), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
P-glycoprotein 1; ATP-binding cassette sub-family B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126530 375.00
Check your price
  • HSP70 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IP and GS
  • Host Mouse
  • Target Species Amphibian, Avian, Fruit fly, Fish, Flatworm, Human, Mouse, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA3007
  • Size 250µL
Applications
Gel Shift (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (1:250-1:1000)
Alternate names:
Hsp68; Heat shock 70 kDa protein 3; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3007 375.00
Check your price
  • Caspase 6 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191593
  • Size 250µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
Apoptotic protease Mch-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191593 375.00
Check your price
  • MCP-1 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Mouse
  • Target Species Human, Rabbit
  • Conjugate Unconjugated
  • Catalog # MA121383
  • Size 250µg
Applications
ELISA (Assay dependent), Neutralization (0.5 µg/ml), Western Blot (0.1-5 µg/ml)
Alternate names:
Small-inducible cytokine A2; Monocyte secretory protein JE; Monocyte chemotactic protein 1; Monocyte chemotactic and activating factor; Monocyte chemoattractant protein 1; HC11
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121383 375.00
Check your price
  • IL-10 Monoclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rat
  • Target Species Human
  • Conjugate Biotin
  • Catalog # M011B
  • Size 250µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cytokine synthesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
M011B 375.00
Check your price
  • Haloperidol Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA191883
  • Size 250µg
Applications
ELISA (Assay dependent)
Alternate names:
Aloperidin, Bioperidolo, Brotopon, Dozic, Duraperidol, Einalon S, Eukystol, Haldol, Halosten, Keselan, Linton, Peluces, Serenace, Serenase, Sigaperido
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191883 375.00
Check your price
  • G-CSF Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121362
  • Size 250µg
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Pluripoietin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121362 375.00
Check your price
  • CD169 Monoclonal Antibody for IF, IHC (F), Flow and IP
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA180164
  • Size 250µg
Applications
Flow Cytometry (1/500), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:50-1:250), Immunoprecipitation (Assay Dependent)
Alternate names:
CD169, SIGLEC-1, SN, dJ1009E24.1, sialic acid-binding Ig-like lectin 1, sialic acid-binding immunoglobulin-like lectin 1, sialoadhesin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180164 375.00
Check your price
  • Follicular Dendritic Cells Monoclonal Antibody for IHC (F)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA516663
  • Size 250ug
Applications
Immunohistochemistry (Frozen) (1:100-1:200)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516663 375.00
Check your price
  • C5AR1 Monoclonal Antibody for IHC (F) and Flow
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA170060
  • Size 250ug
Applications
Flow Cytometry (1 ug/1 million cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
C5a anaphylatoxin chemotactic receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170060 375.00
Check your price
  • RPE65 Monoclonal Antibody, DyLight® 488 conjugate for Western Blot, IF, ICC, IHC and IHC (F)
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Pig, Rat, Xenopus
  • Conjugate DyLight® 488
  • Catalog # MA516042
  • Size 200µl
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:250), Western Blot (1:5000-1:10,000)
Alternate names:
Retinol isomerase; Retinal pigment epithelium-specific 65 kDa protein; All-trans-retinyl-palmitate hydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516042 375.00
Check your price
  • NMDAR1 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA3102
  • Size 200µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunohistochemistry (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:600)
Alternate names:
N-methyl-D-aspartate receptor subunit NR1; Glutamate [NMDA] receptor subunit zeta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3102 375.00
Check your price
  • Staphylococcus Enterotoxin pan Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183468
  • Size 200µg
Applications
ELISA (1/5,000-1/20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183468 375.00
Check your price
  • Salmonella typhimurium LPS Monoclonal Antibody for IF and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183451
  • Size 200µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Salmonellae, S. typhimurium lipopolysaccharide
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183451 375.00
Check your price
  • Mycobacterium avium Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183339
  • Size 200µg
Applications
ELISA (1:5000)
Alternate names:
M. avium
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183339 375.00
Check your price
  • MMP1 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA112891
  • Size 200µg
Applications
Immunohistochemistry (Paraffin) (1-5 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Matrix metalloproteinase-1; Fibroblast collagenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112891 375.00
Check your price
  • Leu-Enkephalin Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # MA183138
  • Size 200µg
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183138 375.00
Check your price
  • ITGA5 Monoclonal Antibody for IHC (F), Flow, IP and FN
  • Host Mouse
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # MA181134
  • Size 200µg
Applications
Flow Cytometry (Neat-1:10), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (0.5-1 ug/ml), Immunoprecipitation (1-5 ug/ml)
Alternate names:
VLA-5; Integrin alpha-F; Fibronectin receptor subunit alpha; CD49 antigen-like family member E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181134 375.00
Check your price
  • HMOX1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1112
  • Size 200µg
Applications
Flow Cytometry (10 ug/ml), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1,000)
Alternate names:
P32 protein; HSP32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1112 375.00
Check your price
  • Cholera Toxin beta Monoclonal Antibody for IHC (F) and ELISA
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA183519
  • Size 200µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
beta subunit cholera toxin, CTB, VC1456
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183519 375.00
Check your price