Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  50  60  67 
751 - 780 of 2000 products displayed
  • Nhe-1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520444
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20µg/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Solute carrier family 9 member 1; Na(+)/H(+) exchanger 1; Na(+)/H(+) antiporter, amiloride-sensitive; APNH
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520444 349.00
Check your price
  • MUC1 Antibody (Monoclonal, GP1.4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA106503
  • Size 100 µg
Applications
Flow Cytometry (1:25-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:25-1:200), Western Blot (1:100-1:1000)
Alternate names:
Tumor-associated mucin; Tumor-associated epithelial membrane antigen; Polymorphic epithelial mucin; Peanut-reactive urinary mucin; PEMT; Krebs von den Lungen-6; H23AG; Episialin; Carcinoma-associated mucin; Cancer antigen 15-3; Breast carcinoma-associated antigen DF3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106503 349.00
Check your price
  • MUC1 Antibody (Monoclonal, 126E7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA135688
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (Assay dependent.)
Alternate names:
Tumor-associated mucin; Tumor-associated epithelial membrane antigen; Polymorphic epithelial mucin; Peanut-reactive urinary mucin; PEMT; Krebs von den Lungen-6; H23AG; Episialin; Carcinoma-associated mucin; Cancer antigen 15-3; Breast carcinoma-associated antigen DF3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA135688 349.00
Check your price
  • IL1RL2 Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547770
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-1 receptor-related protein 2; IL-36 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547770 349.00
Check your price
  • ID1 Antibody
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547617
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Inhibitor of differentiation 1; Inhibitor of DNA binding 1; Class B basic helix-loop-helix protein 24
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547617 349.00
Check your price
  • HSV Type 2 gE Antibody, FITC (Monoclonal, F54C)
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17260
  • Size 100 µg
Applications
ELISA (Assay Dependent)
Alternate names:
HSV2, HSV-2, Human herpes virus 2, HHV-2, Herpes Simplex II, HSV Type 2, Herpes Simplex 2, HSV Type II, Glycoprotein E
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17260 349.00
Check your price
  • HCMV IL-10 Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA547082
  • Size 100 µg
Applications
Neutralization (0.15-0.9 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547082 349.00
Check your price
  • GRP78 Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1014A
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Western Blot (2 µg/ml)
Alternate names:
Steroidogenesis-activator polypeptide; Immunoglobulin heavy chain-binding protein; Heat shock 70 kDa protein 5; Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1014A 349.00
Check your price
  • Ghrelin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA11046
  • Size 100 µg
Applications
Western Blot (1 ug/ml)
Alternate names:
Protein M46; Motilin-related peptide; Growth hormone-releasing peptide; Growth hormone secretagogue
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11046 349.00
Check your price
  • Galectin 9 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547503
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
HUAT, LGALS9A, ecalectin, gal-9, galectin 9, galectin-9, tumor antigen HOM-HD-21, urate transporter/channel protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547503 349.00
Check your price
  • FZD1 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547072
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
FZD1, Chfz-1, Fz1, Frizzled 1, Fz receptor 1, HFz1, Frizzled homolog 1, Fz-1, FzE1, Rfz1, Xfrizzled-1, Wnt receptor, Dfrizzled-1, Frizzled family receptor 1, Frizzled-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547072 349.00
Check your price
  • EPCR Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547382
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Endothelial cell protein C receptor; Centrosomal protein CCD41; Centrocyclin; Activated protein C receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547382 349.00
Check your price
  • Desmocollin 2 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547651
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Desmosomal glycoprotein III; Desmosomal glycoprotein II; Desmocollin-3; Cadherin family member 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547651 349.00
Check your price
  • CtIP Antibody (Monoclonal, 19E8)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA123304
  • Size 100 µg
Applications
ChIP assay (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:500-1:1000)
Alternate names:
Sporulation in the absence of SPO11 protein 2 homolog; Retinoblastoma-interacting protein and myosin-like; Retinoblastoma-binding protein 8; CtBP-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123304 349.00
Check your price
  • beta-3 Tubulin Antibody (Monoclonal, 2G10)
  • Host Mouse
  • Target Species Bovine, Guinea pig, Hamster, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1118
  • Size 100 µg
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (5 µg), Western Blot (1:500-1:2000)
Alternate names:
Tubulin beta-III; Tubulin beta-4 chain; Neuron-specific class III beta-tubulin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1118 349.00
Check your price
  • TRIM21 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518147
  • Size 100 ug
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Tripartite motif-containing protein 21; Sjoegren syndrome type A antigen; Ro(SS-A); RING finger protein 81; 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa Ro protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518147 349.00
Check your price
  • Tankyrase Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA517931
  • Size 100 ug
Applications
Western Blot (0.5-1.5 ug/ml)
Alternate names:
Tankyrase I; TRF1-interacting ankyrin-related ADP-ribose polymerase; TNKS-1; Poly [ADP-ribose] polymerase 5A; ADP-ribosyltransferase diphtheria toxin-like 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517931 349.00
Check your price
  • STING Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520782
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Transmembrane protein 173; Mediator of IRF3 activation; Endoplasmic reticulum interferon stimulator
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520782 349.00
Check your price
  • PITPNM1 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518266
  • Size 100 ug
Applications
Immunocytochemistry (Assay Dependent), Western Blot (2 ug/ml)
Alternate names:
Pyk2 N-terminal domain-interacting receptor 2; Phosphatidylinositol transfer protein, membrane-associated 1; Mpt-1; Drosophila retinal degeneration B homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518266 349.00
Check your price
  • Phospho-IkB alpha (Ser32, Ser36) Antibody (Monoclonal, 39A1413)
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA516161
  • Size 100 ug
Applications
Immunoprecipitation (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Major histocompatibility complex enhancer-binding protein MAD3; I-kappa-B-alpha; ECI-6; RL/IF-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516161 349.00
Check your price
  • PD-1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520350
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 ug/ml), Western Blot (1 µg/ml)
Alternate names:
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, programmed cell death protein 1, protein PD-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520350 349.00
Check your price
  • OTUB1 Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518960
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Ubiquitin-specific-processing protease OTUB1; Otubain-1; OTU domain-containing ubiquitin aldehyde-binding protein 1; Deubiquitinating enzyme OTUB1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518960 349.00
Check your price
  • Noggin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522795
  • Size 100 ug
Applications
Western Blot (1:1000-1:2000)
Alternate names:
SYM1, SYNS1, symphalangism 1 (proximal)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522795 349.00
Check your price
  • NLRP3 Antibody (Monoclonal, 25N10E9)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA516274
  • Size 100 ug
Applications
Western Blot (2-5 µg/ml)
Alternate names:
PYRIN-containing APAF1-like protein 1; Cryopyrin; Cold-induced autoinflammatory syndrome 1 protein; Caterpiller protein 1.1; Angiotensin/vasopressin receptor AII/AVP-like; Mast cell maturation-associated-inducible protein 1; Cold autoinflammatory syndrome 1 protein homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516274 349.00
Check your price
  • Nanog Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520889
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Homeobox transcription factor Nanog; Early embryo specific expression NK-type homeobox protein; ES cell-associated protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520889 349.00
Check your price
  • JMJD1B Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520803
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Jumonji domain-containing protein 1B; JmjC domain-containing histone demethylation protein 2B; Nuclear protein 5qNCA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520803 349.00
Check your price
  • Intestinal Alkaline Phosphatase Antibody (Monoclonal, P4A10E4)
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # MA514742
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Radioimmune Assays (Assay Dependent)
Alternate names:
PLAP-1, Regan isozyme, alkaline phosphatase Regan isozyme, alkaline phosphatase, placental type, alkaline phosphomonoesterase, glycerophosphatase, placental alkaline phosphatase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514742 349.00
Check your price
  • IKK alpha Antibody (Monoclonal, 14A231)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516157
  • Size 100 ug
Applications
Flow Cytometry (0.25-0.5 µg/10^6 cells), Immunocytochemistry (1:10), Immunofluorescence (1:10), Immunohistochemistry (Paraffin) (5 µg/ml), Immunoprecipitation (1-2 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516157 349.00
Check your price
  • Dectin 1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534382
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (5 ug/mL), Immunohistochemistry (5 ug/mL), Western Blot (1-2 ug/mL)
Alternate names:
Dendritic cell-associated C-type lectin 1; C-type lectin superfamily member 12; Beta-glucan receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534382 349.00
Check your price
  • BACE2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519950
  • Size 100 ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Theta-secretase; Membrane-associated aspartic protease 1; Memapsin-1; Beta-site amyloid precursor protein cleaving enzyme 2; Aspartyl protease 1; Down region aspartic protease; Aspartic-like protease 56 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519950 349.00
Check your price