Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
91 - 120 of 2000 products displayed
  • IL-2 Polyclonal Antibody for Western Blot, IF, ICC and ELISA
  • Host Goat
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # PA546923
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
T-cell growth factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546923 375.00
Check your price
  • EpCAM Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA110197
  • Size 100 Tests
Applications
Flow Cytometry (20 ul/10^6 cells)
Alternate names:
Tumor-associated calcium signal transducer 1; Major gastrointestinal tumor-associated protein GA733-2; KSA; KS 1/4 antigen; Epithelial glycoprotein 314; Epithelial glycoprotein; Epithelial cell surface antigen; Cell surface glycoprotein Trop-1; Adenocarcinoma-associated antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110197 249.00
Check your price
  • CD8 Monoclonal Antibody for IF, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA181180
  • Size 250 µg
Applications
Flow Cytometry (1:50-1:100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
T-cell surface glycoprotein Lyt-3; T-cell membrane glycoprotein Ly-3; Lymphocyte antigen 3; T-cell surface glycoprotein Lyt-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181180 249.00
Check your price
  • CD45R Monoclonal Antibody, PE-Texas Red® conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE-Texas Red®
  • Catalog # RM2617
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
B220, CD45R, Cd45, Ly-5, Lyt-4, T200, Cluster designation 45R
Catalog # Size Price (AUD) Your Price (AUD) Qty
RM2617 363.00
Check your price
  • IFN beta Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520390
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 ug/ml), Western Blot (5 ug/ml)
Alternate names:
Fibroblast interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520390 349.00
Check your price
  • GLUT1 Monoclonal Antibody for IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA511315
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Glucose transporter type 1, erythrocyte/brain; HepG2 glucose transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511315 349.00
Check your price
  • CD14 Monoclonal Antibody, Alexa Fluor® 700 conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 700
  • Catalog # MHCD1429
  • Size 0.5 ml
Applications
Flow Cytometry (Assay-Dependent)
Alternate names:
Myeloid cell-specific leucine-rich glycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MHCD1429 352.00
Check your price
  • CD32 Monoclonal Antibody for IHC (F), Flow, IP and FN
  • Host Mouse
  • Target Species Dog, Human, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # MA181191
  • Size 0.5 mg
Applications
Flow Cytometry (20 µg/ml), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (1:500-1:1000), Immunoprecipitation (20 µg/ml)
Alternate names:
Fc-gamma RII-c; CDw32; Fc-gamma RII-a; Fc-gamma RII-b
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181191 700.00
Check your price
  • Parathyroid Hormone for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10937
  • Size 500 µg
Applications
Control (Assay Dependent)
Alternate names:
PTH
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10937 700.00
Check your price
  • B7-H3 Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515693
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunohistochemistry (Paraffin) (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Costimulatory molecule; B7 homolog 3; 4Ig-B7-H3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515693 349.00
Check your price
  • Bovine Coronavirus Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA123055
  • Size 500 µg
Applications
ELISA (Assay dependent)
Alternate names:
BCV, Calf Enteritis, Enzootic Pneumonia complex, Winter Dysentery
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123055 349.00
Check your price
  • IFN alpha Monoclonal Antibody for Neu
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # 271051
  • Size 500 ug
Applications
Neutralization (Assay Dependent)
Alternate names:
IFN-alpha-6; interferon alpha 1; interferon-alpha-6
Catalog # Size Price (AUD) Your Price (AUD) Qty
271051 349.00
Check your price
  • Parkin Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513398
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Parkinson juvenile disease protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513398 349.00
Check your price
  • HMGA2 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521320
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:5000-1:20,000)
Alternate names:
High mobility group AT-hook protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521320 349.00
Check your price
  • Podoplanin Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516270
  • Size 100 ug
Applications
Flow Cytometry (0.5 µg/10^5 cells), Western Blot (3-5 µg/ml)
Alternate names:
Transmembrane glycoprotein E11; T1-alpha; PA2.26 antigen; OTS-8; Glycoprotein 38; Aggrus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516270 349.00
Check your price
  • RDH16 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525370
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Sterol/retinol dehydrogenase; Short chain dehydrogenase/reductase family 9C member 8; Microsomal NAD(+)-dependent retinol dehydrogenase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525370 349.00
Check your price
  • p21 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514456
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
p21; Melanoma differentiation-associated protein 6; CDK-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514456 349.00
Check your price
  • CD31 Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514372
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514372 349.00
Check your price
  • PSMD4 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1966
  • Size 200 µL
Applications
Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Multiubiquitin chain-binding protein; Antisecretory factor 1; 26S proteasome regulatory subunit S5A; 26S proteasome regulatory subunit RPN10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1966 349.00
Check your price
  • Oval Cell Marker Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516137
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:100), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:100), Western Blot (1:100-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516137 349.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Alexa Fluor® 647
  • Catalog # MA516308A647
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308A647 225.00
Check your price
  • c-Myc Tag Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Tag
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA121316D800
  • Size 50 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Transcription factor p64; Proto-oncogene c-Myc; Class E basic helix-loop-helix protein 39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121316D800 225.00
Check your price
  • DYKDDDDK Tag Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot, IF and ICC
  • Host Rat
  • Target Species Tag
  • Conjugate Alexa Fluor® 488
  • Catalog # MA1142A488
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500)
Alternate names:
FLAG Tag, DYKDDDDK tag, FLAG antibody, ECS epitope tag, DDDDK epitope tag, DDDDK, DYKDDDDK epitope tag, Enterokinase Cleavage Site epitope tag, Enterokinase Cleavage Site tag, ECS tag
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1142A488 219.00
Check your price
  • CD204 Monoclonal Antibody for Western Blot, IHC (F), Flow, IP, ELISA and FN
  • Host Rat
  • Target Species Fish, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # MA516494
  • Size 500 ug
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Scavenger receptor type A; Macrophage acetylated LDL receptor I and II
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516494 650.00
Check your price
  • Laminin alpha-5 Monoclonal Antibody for IF, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183286
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Laminin-15 subunit alpha; Laminin-11 subunit alpha; Laminin-10 subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183286 650.00
Check your price
  • TLR1 Monoclonal Antibody for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516691
  • Size 200 ug
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Toll/interleukin-1 receptor-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516691 650.00
Check your price
  • Leucomalachite Green Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA175233
  • Size 100 µL
Applications
ELISA (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175233 650.00
Check your price
  • SLA Class II DR Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Bovine, Pig
  • Conjugate FITC
  • Catalog # MA516490
  • Size 100 ug
Applications
Flow Cytometry (Neat)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516490 650.00
Check your price
  • CD8 alpha Monoclonal Antibody, Alexa Fluor® 405 conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 405
  • Catalog # MCD0826
  • Size 1 mL
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
T-cell surface glycoprotein Lyt-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MCD0826 326.00
Check your price
  • CD45 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Dog
  • Conjugate APC
  • Catalog # MA516995
  • Size 100 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516995 650.00
Check your price