Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  40  50  60  67 
691 - 720 of 2000 products displayed
  • PKLR Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA522178
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Red cell/liver pyruvate kinase; R-type/L-type pyruvate kinase; Pyruvate kinase isozymes L/R; Pyruvate kinase 1; L-PK
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522178 349.00
Check your price
  • PKC lambda/iota Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527811
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
nPKC-iota; Atypical protein kinase C-lambda/iota
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527811 349.00
Check your price
  • PKA gamma Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528148
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
KAPG, PKACg, PKA C-gamma, cAMP-dependent protein kinase catalytic subunit gamma, serine(threonine) protein kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528148 349.00
Check your price
  • PKA alpha Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528604
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PKACA, PKA C-alpha, cAMP-dependent protein kinase catalytic subunit alpha, protein kinase A catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528604 349.00
Check your price
  • PITPNB Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531358
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
PI-TP-beta, PtdInsTP, VIB1B, PtdIns transfer protein beta, phosphatidylinositol transfer protein beta isoform, phosphotidylinositol transfer protein, beta, ptdInsTP beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531358 349.00
Check your price
  • PIN1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529675
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Rotamase Pin1; Peptidyl-prolyl cis-trans isomerase Pin1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529675 349.00
Check your price
  • PIASy Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530628
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein inhibitor of activated STAT protein gamma; Protein inhibitor of activated STAT protein 4; PIASy
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530628 349.00
Check your price
  • Phospho-STAT5 alpha (Ser780) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536688
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Mammary gland factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536688 349.00
Check your price
  • Phospho-NFkB p52 (Ser869) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536675
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; DNA-binding factor KBF2; Oncogene Lyt-10; Lymphocyte translocation chromosome 10 protein; H2TF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536675 349.00
Check your price
  • Phospho-JNK1/JNK2/JNK3 (Thr183) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536832
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
p54 gamma; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; SAPK gamma; c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; Stress-activated protein kinase 1c; JNK-46; p54-beta; c-Jun N-terminal kinase 3; Stress-activated protein kinase JNK3; SAPK-beta; p54-alpha; SAPK-alpha; Stress-activated protein kinase 1b; MAP kinase p49 3F12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536832 349.00
Check your price
  • Phospho-IKK alpha (Thr23) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536652
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor 16; Nuclear factor NF-kappa-B inhibitor kinase alpha; I-kappa-B kinase 1; Conserved helix-loop-helix ubiquitous kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536652 349.00
Check your price
  • Phospho-FOXO1/FOXO3/FOXO4 (Thr24, Thr32) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536737
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200)
Alternate names:
Fork head domain transcription factor AFX1; Forkhead in rhabdomyosarcoma; Forkhead box protein O1A; Afxh; Forkhead in rhabdomyosarcoma-like 1; AF6q21 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536737 349.00
Check your price
  • Phospho-ERK1/ERK2 (Tyr204) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536776
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536776 349.00
Check your price
  • Phospho-C/EBP beta (Thr188, Thr235) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536629
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein; Silencer factor B; Liver-enriched transcriptional activator; Interleukin-6-dependent-binding protein; C/EBP-related protein 2; AGP/EBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536629 349.00
Check your price
  • Phospho-Bcl-2 (Thr56) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536625
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Immunoprecipitation (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Apoptosis regulator Bcl-2, Bcl 2, BCL2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536625 349.00
Check your price
  • Phospho-Aurora B/C (Thr236, Thr202) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536030
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Serine/threonine-protein kinase aurora-B; Serine/threonine-protein kinase 5; Serine/threonine-protein kinase 12; STK-1; Aurora/IPL1-related kinase 2; Aurora- and IPL1-like midbody-associated protein 1; Aurora 1; Serine/threonine-protein kinase aurora-C; Serine/threonine-protein kinase 13; Aurora/IPL1/Eg2 protein 2; Aurora/IPL1-related kinase 3; Aurora 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536030 349.00
Check your price
  • Phospho-ATF2 (Thr52) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535391
  • Size 100 µL
Applications
Western Blot (1:250)
Alternate names:
cAMP response element-binding protein CRE-BP1; Histone acetyltransferase ATF2; HB16; Cyclic AMP-responsive element-binding protein 2; Activating transcription factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535391 349.00
Check your price
  • Phospho-ASK1 (Ser83) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536618
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
MAPK/ERK kinase kinase 5; Apoptosis signal-regulating kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536618 349.00
Check your price
  • Phospho-Aquaporin 2 (Ser264) Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA535387
  • Size 100 µL
Applications
Immunohistochemistry (1:100), Western Blot (1:1000)
Alternate names:
Water channel protein for renal collecting duct; WCH-CD; Collecting duct water channel protein; Aquaporin-CD; ADH water channel
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535387 349.00
Check your price
  • Phospho-AMPK alpha-1,2 (Thr172, Thr183) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536764
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Hydroxymethylglutaryl-CoA reductase kinase; Acetyl-CoA carboxylase kinase; Tau-protein kinase PRKAA1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536764 349.00
Check your price
  • PFKFB3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521931
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
iPFK-2; Renal carcinoma antigen NY-REN-56; 6PF-2-K/Fru-2,6-P2ase brain/placenta-type isozyme
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521931 349.00
Check your price
  • PDE6D Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522008
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Protein p17
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522008 349.00
Check your price
  • PD-1 Antibody (Monoclonal, DHIC2-4A10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516134
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:100), Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (Frozen) (1:100), Western Blot (1:100-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516134 349.00
Check your price
  • PCCB Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521647
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Propanoyl-CoA:carbon dioxide ligase subunit beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521647 349.00
Check your price
  • PAX8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521368
  • Size 100 µL
Applications
ChIP assay (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:500-1:1000), Western Blot (1:500-1:3000)
Alternate names:
paired box protein Pax-8, paired domain gene 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521368 349.00
Check your price
  • PAX6 Antibody (Monoclonal, 1C8)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515657
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Western Blot (1/500-1/2000)
Alternate names:
Oculorhombin; Aniridia type II protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515657 349.00
Check your price
  • PARP Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527219
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Poly[ADP-ribose] synthase 1; NAD(+) ADP-ribosyltransferase 1; ADP-ribosyltransferase diphtheria toxin-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527219 349.00
Check your price
  • PARP Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534802
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Poly[ADP-ribose] synthase 1; NAD(+) ADP-ribosyltransferase 1; ADP-ribosyltransferase diphtheria toxin-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534802 349.00
Check your price
  • PAI2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527857
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Urokinase inhibitor; Serpin B2; Placental plasminogen activator inhibitor; Monocyte Arg-serpin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527857 349.00
Check your price
  • PAG3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530287
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Pyk2 C-terminus-associated protein; Paxillin-associated protein with ARF GAP activity 3; Development and differentiation-enhancing factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530287 349.00
Check your price