Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  50  60  67 
721 - 750 of 2000 products displayed
  • PABP Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527443
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
PAB1, PABP, PABP1, PABPC2, PABPL1, poly(A) binding protein, cytoplasmic 2, polyadenylate-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527443 349.00
Check your price
  • p16INK4a Antibody (Monoclonal, 1E12E10)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517142
  • Size 100 µL
Applications
Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
p16-INK4a; Cyclin-dependent kinase 4 inhibitor A; p14ARF; Cyclin-dependent kinase inhibitor 2A; Alternative reading frame; p19ARF; Multiple tumor suppressor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517142 349.00
Check your price
  • Oval Cell Marker Antibody (Monoclonal, MIC1-1C3)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA516136
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:100), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
macrophage inhibitory cytokine-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516136 349.00
Check your price
  • OLIG2 Antibody (Monoclonal, 1G11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515810
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Protein kinase C-binding protein RACK17; Protein kinase C-binding protein 2; Class E basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515810 349.00
Check your price
  • NRG1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527331
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
ARIA, GGF, GGF2, HGL, HRG, HRG1, HRGA, MST131, NDF, SMDF, MSTP131, glial growth factor, heregulin, alpha (45kD, ERBB2 p185-activator), neu differentiation factor, neuregulin 1 type IV beta 1a, neuregulin 1 type IV beta 3, pro-NRG1, pro-neuregulin-1, membrane-bound isoform, sensory and motor neuron derived factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527331 349.00
Check your price
  • NQO1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521290
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Quinone reductase 1; Phylloquinone reductase; NAD(P)H:quinone oxidoreductase 1; Menadione reductase; DT-diaphorase; Azoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521290 349.00
Check your price
  • NOTCH3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528082
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
CADASIL, CASIL, Notch homolog 3, neurogenic locus notch homolog protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528082 349.00
Check your price
  • NME2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530602
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
nm23-M2; P18; Histidine protein kinase NDKB; nm23-H2; C-myc purine-binding transcription factor PUF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530602 349.00
Check your price
  • Nkx2.5 Antibody (Monoclonal, 2E1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515551
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Homeobox protein NK-2 homolog E; Homeobox protein CSX; Cardiac-specific homeobox
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515551 349.00
Check your price
  • NHP2L1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522010
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
U4/U6.U5 tri-snRNP 15.5 kDa protein; U4/U6.U5 small nuclear ribonucleoprotein SNU13; SNU13 homolog; OTK27; High mobility group-like nuclear protein 2 homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522010 349.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527617
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527617 349.00
Check your price
  • NFkB p65 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534737
  • Size 100 µL
Applications
Gel Shift (Assay-dependent), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:500-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3; Nuclear factor NF-kappa-B p65 subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534737 349.00
Check your price
  • NFkB p52 Antibody (Monoclonal, 9D2)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517217
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (0.5 µg/ml)
Alternate names:
Nuclear factor of kappa light polypeptide gene enhancer in B-cells 2; DNA-binding factor KBF2; Oncogene Lyt-10; Lymphocyte translocation chromosome 10 protein; H2TF1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517217 349.00
Check your price
  • Neutrophil elastase Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529659
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
PMN elastase; Medullasin; Human leukocyte elastase; Elastase-2; Bone marrow serine protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529659 349.00
Check your price
  • NDRG1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523158
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Rit42; Reducing agents and tunicamycin-responsive protein; Nickel-specific induction protein Cap43; N-myc downstream-regulated gene 1 protein; Differentiation-related gene 1 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523158 349.00
Check your price
  • NBS1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522972
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (2 µg/ml)
Alternate names:
Nijmegen breakage syndrome protein 1; Cell cycle regulatory protein p95; Nijmegen breakage syndrome protein 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522972 349.00
Check your price
  • MYH9 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527506
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Non-muscle myosin heavy chain IIa; Non-muscle myosin heavy chain A; Myosin heavy chain, non-muscle IIa; Myosin heavy chain 9; Cellular myosin heavy chain, type A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527506 349.00
Check your price
  • MX1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522149
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:500), Western Blot (1:1000-1:10,000)
Alternate names:
Myxovirus resistance protein 1; Myxoma resistance protein 1; Interferon-regulated resistance GTP-binding protein MxA; Interferon-induced protein p78
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522149 349.00
Check your price
  • MURF1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522112
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Tripartite motif-containing protein 63; Striated muscle RING zinc finger protein; RING finger protein 28; Muscle-specific RING finger protein 1; Iris RING finger protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522112 349.00
Check your price
  • MUC4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523077
  • Size 100 µL
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunohistochemistry (Paraffin) (1:400), Western Blot (1:5000)
Alternate names:
Tracheobronchial mucin; Testis mucin; Pancreatic adenocarcinoma mucin; Ascites sialoglycoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523077 349.00
Check your price
  • MTMR2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527748
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Phosphatidylinositol-3-phosphate phosphatase; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527748 349.00
Check your price
  • MTHFR Antibody (Monoclonal, 5D3)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515844
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
RP11-56N19.4, 5,10-methylenetetrahydrofolate reductase (NADPH); methylenetetrahydrofolate reductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515844 349.00
Check your price
  • MRPL3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521892
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MRL3, RPML3, 39S ribosomal protein L3, mitochondrial, L3mt, MRP-L3, mitochondrial 60S ribosomal protein L3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521892 349.00
Check your price
  • MPP5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530966
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
PALS1, MAGUK p55 subfamily member 5, protein associated with lin seven 1, stardust
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530966 349.00
Check your price
  • MOXD1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531526
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Monooxygenase X
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531526 349.00
Check your price
  • MMP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527210
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Matrix metalloproteinase-1; Fibroblast collagenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527210 349.00
Check your price
  • MLH1 Antibody (Monoclonal, 4C9C7)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA515431
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
MutL protein homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515431 349.00
Check your price
  • MIOX Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA530650
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Renal-specific oxidoreductase; Myo-inositol oxygenase; Kidney-specific protein 32; Aldehyde reductase-like 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530650 349.00
Check your price
  • MFN2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527606
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transmembrane GTPase MFN2; Mitochondrial assembly regulatory factor; Hypertension-related protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527606 349.00
Check your price
  • MDMX Antibody (Monoclonal, 2D10F4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515432
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
p53-binding protein Mdm4; Protein Mdmx; Mdm2-like p53-binding protein; Double minute 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515432 349.00
Check your price