Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  50  60  67 
781 - 810 of 2000 products displayed
  • Helicobacter pylori Urease B Antibody (Monoclonal, GT1263)
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # MA516311
  • Size 100 µL
Applications
ELISA (Assay-dependent), Western Blot (1:5000-1:20,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516311 349.00
Check your price
  • HADH Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA528203
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Short-chain 3-hydroxyacyl-CoA dehydrogenase; Medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528203 349.00
Check your price
  • HABP2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528396
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Plasma hyaluronan-binding protein; Hepatocyte growth factor activator-like protein; Factor seven-activating protease; Factor VII-activating protease
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528396 349.00
Check your price
  • GTF2H1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529379
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
TFIIH basal transcription factor complex p62 subunit; General transcription factor IIH polypeptide 1; Basic transcription factor 2 62 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529379 349.00
Check your price
  • GRP78 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522968
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (0.5 µg/ml)
Alternate names:
Steroidogenesis-activator polypeptide; Immunoglobulin heavy chain-binding protein; Heat shock 70 kDa protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522968 349.00
Check your price
  • GRP78 Antibody
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522905
  • Size 100 µL
Applications
Immunocytochemistry (1:20), Immunofluorescence (1:20), Western Blot (2 µg/ml)
Alternate names:
Steroidogenesis-activator polypeptide; Immunoglobulin heavy chain-binding protein; Heat shock 70 kDa protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522905 349.00
Check your price
  • GRK2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528609
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
G-protein coupled receptor kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528609 349.00
Check your price
  • GRAMD1B Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531387
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
GRAM domain-containing protein 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531387 349.00
Check your price
  • GRAF Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528703
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Rho-type GTPase-activating protein 26; Oligophrenin-1-like protein; GTPase regulator associated with focal adhesion kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528703 349.00
Check your price
  • GPNMB Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527874
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Transmembrane glycoprotein HGFIN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527874 349.00
Check your price
  • GPI Antibody (Monoclonal, 1B7D7)
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA515396
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (Paraffin) (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Phosphohexose isomerase; Phosphoglucose isomerase; Neuroleukin; Autocrine motility factor; Sperm antigen 36
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515396 349.00
Check your price
  • GPD1 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA531051
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
GPD-C, GPDH-C, glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic, glycerol-3-phosphate dehydrogenase, soluble
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531051 349.00
Check your price
  • GNAS Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521702
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Extra large alphas protein; Adenylate cyclase-stimulating G alpha protein; Alternative gene product encoded by XL-exon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521702 349.00
Check your price
  • GLUT9 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA522971
  • Size 100 µL
Applications
Western Blot (0.5 µg/ml)
Alternate names:
Glucose transporter type 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522971 349.00
Check your price
  • GLUT1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA146152
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (0.5-2.0 µg/ml)
Alternate names:
Glucose transporter type 1, erythrocyte/brain; HepG2 glucose transporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146152 349.00
Check your price
  • GFI1 Antibody (Monoclonal, 5D7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515688
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Zinc finger protein 163; Growth factor independent protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515688 349.00
Check your price
  • GEF-H1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA532213
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
Rhobin; Oncogene LFC; Lymphoid blast crisis-like 1; LBC'S first cousin; Guanine nucleotide exchange factor H1; Proliferating cell nucleolar antigen p40; Microtubule-regulated Rho-GEF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532213 349.00
Check your price
  • GDF5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528815
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Radotermin; Lipopolysaccharide-associated protein 4; Cartilage-derived morphogenetic protein 1; Bone morphogenetic protein 14
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528815 349.00
Check your price
  • GDA Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530322
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
p51-nedasin; Guanine aminohydrolase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530322 349.00
Check your price
  • GBF1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527751
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
ARF1GEF, BFA-resistant GEF 1, Golgi-specific brefeldin A-resistance guanine nucleotide exchange factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527751 349.00
Check your price
  • GATA1 Antibody (Monoclonal, 4G1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515637
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
NF-E1 DNA-binding protein; GATA-binding factor 1; Eryf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515637 349.00
Check your price
  • FXR2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528979
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
FMR1L2, FXR2P, fragile X mental retardation syndrome-related protein 2, fragile X-mental retardation 1-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528979 349.00
Check your price
  • FUS Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527531
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Translocated in liposarcoma protein; POMp75; Oncogene TLS; Oncogene FUS; 75 kDa DNA-pairing protein; Protein pigpen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527531 349.00
Check your price
  • FOXP4 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530908
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (1:500-1:3000)
Alternate names:
Fork head-related protein-like A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530908 349.00
Check your price
  • FIS1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA522142
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Tetratricopeptide repeat protein 11; FIS1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522142 349.00
Check your price
  • Fibulin 5 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528843
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Urine p50 protein; Developmental arteries and neural crest EGF-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528843 349.00
Check your price
  • FGFR1 Antibody (Monoclonal, 3D4F7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515530
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Proto-oncogene c-Fgr; N-sam; Fms-like tyrosine kinase 2; Basic fibroblast growth factor receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515530 349.00
Check your price
  • FBXO43 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521622
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Endogenous meiotic inhibitor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521622 349.00
Check your price
  • ERK1 Antibody (Monoclonal, 12D11)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA113041
  • Size 100 µL
Applications
ChIP assay (1-3 ul), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:100), Western Blot (1:1000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA113041 349.00
Check your price
  • ENPP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527905
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Plasma-cell membrane glycoprotein PC-1; Phosphodiesterase I/nucleotide pyrophosphatase 1; Membrane component chromosome 6 surface marker 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527905 349.00
Check your price